Product References
CCL19+ dendritic cells potentiate clinical benefit of anti-PD-(L)1 immunotherapy in triple-negative breast cancer.
Med (New York, N.Y.)
Wu SY,Zhang SW,Ma D,Xiao Y,Liu Y,Chen L,Song XQ,Ma XY,Xu Y,Chai WJ,Jin X,Shao ZM,Jiang YZ
Fri Jun 09 00:00:00 UTC 2023
Invasion characteristics and clinical significance of tumor-associated macrophages in gastrointestinal Krukenberg tumors.
Frontiers in oncology
Huang Z,Li B,Qin H,Mo X
Sat Sep 14 00:00:00 UTC 2024
Opposing Roles of Sphingosine 1-Phosphate Receptors 1 and 2 in Fat Deposition and Glucose Tolerance in Obese Male Mice.
Endocrinology
Asano M,Kajita K,Fuwa M,Kajita T,Mori I,Akahoshi N,Ishii I,Morita H
Mon Jan 09 00:00:00 UTC 2023
Lidocaine Ameliorates Psoriasis by Obstructing Pathogenic CGRP Signaling‒Mediated Sensory Neuron‒Dendritic Cell Communication.
The Journal of investigative dermatology
Yin Q,Sun L,Cai X,Lou F,Sun Y,Wang B,Jiang B,Bao L,Li X,Song N,Tang S,Bai J,Wang Z,Wu Y,Zhou H,Wang H,Yu B,Li Q,Wang H
Mon Aug 01 00:00:00 UTC 2022
Tissue-resident FOLR2+ macrophages associate with CD8+ T cell infiltration in human breast cancer.
Cell
Nalio Ramos R,Missolo-Koussou Y,Gerber-Ferder Y,Bromley CP,Bugatti M,Núñez NG,Tosello Boari J,Richer W,Menger L,Denizeau J,Sedlik C,Caudana P,Kotsias F,Niborski LL,Viel S,Bohec M,Lameiras S,Baulande S,Lesage L,Nicolas A,Meseure D,Vincent-Salomon A,Reyal F,Dutertre CA,Ginhoux F,Vimeux L,Donnadieu E,Buttard B,Galon J,Zelenay S,Vermi W,Guermonprez P,Piaggio E,Helft J
Thu Mar 31 00:00:00 UTC 2022
Morphological analysis of cell distribution and network structure via gap junctions in swine corneal stroma.
The Journal of veterinary medical science
Hoshino N,Takehana K,Hosotani M,Kametani K,Iwasaki T,Hasegawa Y,Ueda H,Watanabe T
Wed Dec 14 00:00:00 UTC 2022
AIM2 regulates anti-tumor immunity and is a viable therapeutic target for melanoma.
The Journal of experimental medicine
Fukuda K,Okamura K,Riding RL,Fan X,Afshari K,Haddadi NS,McCauley SM,Guney MH,Luban J,Funakoshi T,Yaguchi T,Kawakami Y,Khvorova A,Fitzgerald KA,Harris JE
Mon Sep 06 00:00:00 UTC 2021
Tumor β-catenin expression is associated with immune evasion in non-small cell lung cancer with high tumor mutation burden.
Oncology letters
Muto S,Ozaki Y,Yamaguchi H,Mine H,Takagi H,Watanabe M,Inoue T,Yamaura T,Fukuhara M,Okabe N,Matsumura Y,Hasegawa T,Osugi J,Hoshino M,Higuchi M,Shio Y,Nanamiya H,Imai JI,Isogai T,Watanabe S,Suzuki H
Mon Mar 01 00:00:00 UTC 2021
Case Report: Dendritic Cells and Macrophages Capture Sperm in Chronically Inflamed Human Epididymis.
Frontiers in immunology
Zheng W,Zhang S,Chen X,Jiang S,Li Z,Li M
Mon Sep 20 00:00:00 UTC 2021
Growth differentiation factor-15 promotes immune escape of ovarian cancer via targeting CD44 in dendritic cells.
Experimental cell research
Gao Y,Xu Y,Zhao S,Qian L,Song T,Zheng J,Zhang J,Chen B
Sat May 01 00:00:00 UTC 2021
Growth differentiation factor-15 promotes immune escape of ovarian cancer via targeting CD44 in dendritic cells.
Experimental cell research
Gao Y,Xu Y,Zhao S,Qian L,Song T,Zheng J,Zhang J,Chen B
Sat May 01 00:00:00 UTC 2021
In Situ Characterization of Human Lymphoid Tissue Immune Cells by Multispectral Confocal Imaging and Quantitative Image Analysis; Implications for HIV Reservoir Characterization.
Frontiers in immunology
Moysi E,Del Rio Estrada PM,Torres-Ruiz F,Reyes-Terán G,Koup RA,Petrovas C
Mon Nov 01 00:00:00 UTC 2021
Glioma stem-like cells evade interferon suppression through MBD3/NuRD complex-mediated STAT1 downregulation.
The Journal of experimental medicine
Zhan X,Guo S,Li Y,Ran H,Huang H,Mi L,Wu J,Wang X,Xiao D,Chen L,Li D,Zhang S,Yan X,Yu Y,Li T,Han Q,He K,Cui J,Li T,Zhou T,Rich JN,Bao S,Zhang X,Li A,Man J
Mon May 04 00:00:00 UTC 2020
The Extracellular Domain of the β2 Integrin β Subunit (CD18) Is Sufficient for Escherichia coli Hemolysin and Aggregatibacter actinomycetemcomitans Leukotoxin Cytotoxic Activity.
mBio
Ristow LC,Tran V,Schwartz KJ,Pankratz L,Mehle A,Sauer JD,Welch RA
Tue Jul 09 00:00:00 UTC 2019
Food hypersensitivity-induced chronic gastrointestinal inflammation in a non-human primate model of diet-induced obesity.
PloS one
Mustafa T,Li Q,Kelly LE,Gibbon A,Ryan I,Roffey K,Simonds S,Cowley MA,Sleeman MW
Mon Jan 20 00:00:00 UTC 2020
Loss of IDO1 Expression From Human Pancreatic β-Cells Precedes Their Destruction During the Development of Type 1 Diabetes.
Diabetes
Anquetil F,Mondanelli G,Gonzalez N,Rodriguez Calvo T,Zapardiel Gonzalo J,Krogvold L,Dahl-Jørgensen K,Van den Eynde B,Orabona C,Grohmann U,von Herrath MG
Sat Sep 01 00:00:00 UTC 2018
Loss of IDO1 Expression From Human Pancreatic β-Cells Precedes Their Destruction During the Development of Type 1 Diabetes.
Diabetes
Anquetil F,Mondanelli G,Gonzalez N,Rodriguez Calvo T,Zapardiel Gonzalo J,Krogvold L,Dahl-Jørgensen K,Van den Eynde B,Orabona C,Grohmann U,von Herrath MG
Sat Sep 01 00:00:00 UTC 2018
Lacrimal Gland Inflammation Deregulates Extracellular Matrix Remodeling and Alters Molecular Signature of Epithelial Stem/Progenitor Cells.
Investigative ophthalmology & visual science
Umazume T,Thomas WM,Campbell S,Aluri H,Thotakura S,Zoukhri D,Makarenkova HP
Tue Dec 01 00:00:00 UTC 2015
Lacrimal Gland Inflammation Deregulates Extracellular Matrix Remodeling and Alters Molecular Signature of Epithelial Stem/Progenitor Cells.
Investigative ophthalmology & visual science
Umazume T,Thomas WM,Campbell S,Aluri H,Thotakura S,Zoukhri D,Makarenkova HP
Tue Dec 01 00:00:00 UTC 2015