Product References
Adipose‑derived stem cell‑mediated alphastatin targeting delivery system inhibits angiogenesis and tumor growth in glioma.
Molecular medicine reports
Liang C,Wei T,Zhang T,Niu C
17-1338-42 was used in Flow Cytometry to target glioma tissue and inhibit glioma angiogenesis and tumor growth.
Wed Nov 01 00:00:00 UTC 2023
HERC2 promotes inflammation-driven cancer stemness and immune evasion in hepatocellular carcinoma by activating STAT3 pathway.
Journal of experimental & clinical cancer research : CR
Liu Y,Xu Q,Deng F,Zheng Z,Luo J,Wang P,Zhou J,Lu X,Zhang L,Chen Z,Zhang Q,Chen Q,Zuo D
17-1338-42 was used in Flow cytometry/Cell sorting to identify HERC2 functions as a previously unknown modulator of the JAK2/STAT3 pathway, thereby promoting inflammation-induced stemness and immune evasion in HCC.
Wed Feb 01 00:00:00 UTC 2023
MCM7 supports the stemness of bladder cancer stem-like cells by enhancing autophagic flux.
iScience
Mo L,Su B,Xu L,Hu Z,Li H,Du H,Li J
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 17-1338-42) in Flow Cytometry
Fri Sep 16 00:00:00 UTC 2022
BML-260 promotes the growth of cord blood and mobilized peripheral blood hematopoietic stem and progenitor cells with improved reconstitution ability.
Journal of cellular biochemistry
Albayrak E,Akgol S,Turan RD,Uslu M,Kocabas F
17-1338-42 was used in Flow cytometry/Cell sorting to show that BML-260 molecules can be used for human HSC expansion and regulation of function.
Thu Dec 01 00:00:00 UTC 2022
Circ_RNF13 Regulates the Stemness and Chemosensitivity of Colorectal Cancer by Transcriptional Regulation of DDX27 Mediated by TRIM24 Stabilization.
Cancers
Guo Y,Hu G,Tian B,Ma M,Long F,Chen M
17-1338-42 was used in Flow cytometry/Cell sorting to characterize circ_RNF13 and investigate its role in regulating stemness and chemosensitivity in colorectal cancer (CRC), revealing that circ_RNF13 modulates CRC stemness by regulating DDX27 expression through TRIM24-mediated transcriptional regulation and stabilizing TRIM24 by suppressing FBXW7-mediated TRIM24 degradation.
Fri Dec 16 00:00:00 UTC 2022
Astragaloside IV - mediated endothelial progenitor cell exosomes promote autophagy and inhibit apoptosis in hyperglycemic damaged endothelial cells via miR-21/PTEN axis.
Folia histochemica et cytobiologica
Xiong W,Huang XL,Li CY,Wang XL,Lan HW,Wang TT,Chen ZL,Yang QP,Hu AL,Xia YF,Zhu FR,Zhou ZZ
17-1338-42 was used in Flow cytometry/Cell sorting to demonstrate that AS-IV-mediated EPC-exos-miR-21 enhances autophagy and depresses apoptosis in HG-damaged endothelial cells via the miR-21/PTEN axis.
Mon Jan 02 00:00:00 UTC 2023
EMP3 negatively modulates breast cancer cell DNA replication, DNA damage repair, and stem-like properties.
Cell death & disease
Zhou K,Sun Y,Dong D,Zhao C,Wang W
17-1338-42 was used in Flow cytometry/Cell sorting to indicate that EMP3 is a putative tumor suppressor in breast cancer, and EMP3 downregulation may be responsible for breast cancer chemoresistance.
Sun Sep 12 00:00:00 UTC 2021
Small molecule-mediated modulation of ubiquitination and neddylation improves HSC function ex vivo.
Journal of cellular physiology
Albayrak E,Uslu M,Akgol S,Tuysuz EC,Kocabas F
17-1338-42 was used in flow cytometry to shed light on the contribution of SKP2, and associated ubiquitination and neddylation in HSC maintenance, self-renewal, and expansion.
Wed Dec 01 00:00:00 UTC 2021
IGF2BP2 regulates DANCR by serving as an N6-methyladenosine reader.
Cell death and differentiation
Hu X,Peng WX,Zhou H,Jiang J,Zhou X,Huang D,Mo YY,Yang L
17-1338-42 was used in Flow Cytometry to promote cancer stemness-like properties and pancreatic cancer pathogenesis.
Mon Jun 01 00:00:00 UTC 2020
PIM protein kinases regulate the level of the long noncoding RNA H19 to control stem cell gene transcription and modulate tumor growth.
Molecular oncology
Singh N,Padi SKR,Bearss JJ,Pandey R,Okumura K,Beltran H,Song JH,Kraft AS,Olive V
17-1338 was used in Flow cytometry/Cell sorting to study the mechanisms through which PIM kinases regulate tumor growth.
Fri May 01 00:00:00 UTC 2020
Curcumin Enhances Radiosensitization of Nasopharyngeal Carcinoma via Mediating Regulation of Tumor Stem-like Cells by a CircRNA Network.
Journal of Cancer
Zhu D,Shao M,Yang J,Fang M,Liu S,Lou D,Gao R,Liu Y,Li A,Lv Y,Mo Z,Fan Q
17-1338-42 was used in Flow Cytometry to explore the potential relationships among curcumin, circRNAs, and nasopharyngeal carcinoma (NPC).
Mon Sep 28 00:00:00 UTC 2020
High expression of endothelial progenitor cell-induced angiogenic markers is associated with bile acid levels in HCC.
Oncology letters
Chen JL,Wang L,Li R,Jiao YF,Yu WF
17-1338 was used in Flow cytometry/Cell sorting to investigate the factors influencing the number of EPCs and circulating progenitor cells (CPCs), as well as the expression levels of vascular endothelial growth factor receptor 2 (VEGFR-2) and CD34, in patients with hepatocellular carcinoma.
Tue Sep 01 00:00:00 UTC 2020
The Deubiquitinase USP4 Stabilizes Twist1 Protein to Promote Lung Cancer Cell Stemness.
Cancers
Li F,Hu Q,He T,Xu J,Yi Y,Xie S,Ding L,Fu M,Guo R,Xiao ZJ,Niu M
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 17-1338-42) in Flow Cytometry
Mon Jun 15 00:00:00 UTC 2020
Profiling and Targeting of Energy and Redox Metabolism in Grade 2 Bladder Cancer Cells with Different Invasiveness Properties.
Cells
Pasquale V,Ducci G,Campioni G,Ventrici A,Assalini C,Busti S,Vanoni M,Vago R,Sacco E
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 17-1338-42) in Flow Cytometry
Fri Dec 11 00:00:00 UTC 2020
Chemerin enhances the adhesion and migration of human endothelial progenitor cells and increases lipid accumulation in mice with atherosclerosis.
Lipids in health and disease
Jia J,Yu F,Xiong Y,Wei W,Ma H,Nisi F,Song X,Yang L,Wang D,Yuan G,Zhou H
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 17-1338-42) in Flow Cytometry
Sun Sep 20 00:00:00 UTC 2020
FZD5 contributes to TNBC proliferation, DNA damage repair and stemness.
Cell death & disease
Sun Y,Wang Z,Na L,Dong D,Wang W,Zhao C
17-1338-42 was used in Flow Cytometry to show that FZD5 is a novel target for the development of therapeutic strategies to overcome chemoresistance and prevent recurrence in TNBC.
Sat Dec 12 00:00:00 UTC 2020
Overexpression of Aiolos promotes epithelial-mesenchymal transition and cancer stem cell-like properties in lung cancer cells.
Scientific reports
Hung JJ,Kao YS,Huang CH,Hsu WH
17-1338 was used in Flow cytometry/Cell sorting to study the role of Aiolos/Ikaros family zinc finger 3 in epithelial-mesenchymal transition and cancer stem cell-like properties in lung cancer.
Thu Feb 28 00:00:00 UTC 2019
Loss of Disabled-2 Expression in Pancreatic Cancer Progression.
Scientific reports
Hocevar BA
17-1338 was used in Flow cytometry/Cell sorting to identify Disabled-2 as a tumor suppressor by modulation of TGFβ-stimulated epithelial-to-mesenchymal transition and cancer stem cell phenotype.
Fri May 17 00:00:00 UTC 2019
ING5 activity in self-renewal of glioblastoma stem cells via calcium and follicle stimulating hormone pathways.
Oncogene
Wang F,Wang AY,Chesnelong C,Yang Y,Nabbi A,Thalappilly S,Alekseev V,Riabowol K
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 17-1338-42) in Flow Cytometry
Thu Jan 18 00:00:00 UTC 2018
Inhibition of Fas associated phosphatase 1 (Fap1) facilitates apoptosis of colon cancer stem cells and enhances the effects of oxaliplatin.
Oncotarget
Huang W,Bei L,Eklund EA
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 17-1338-42) in Flow Cytometry
Fri May 25 00:00:00 UTC 2018
Targeting of BMI-1 with PTC-209 inhibits glioblastoma development.
Cell cycle (Georgetown, Tex.)
Kong Y,Ai C,Dong F,Xia X,Zhao X,Yang C,Kang C,Zhou Y,Zhao Q,Sun X,Wu X
17-1338-42 was used in Flow Cytometry to provide proof-of-concept for inhibitors targeting BMI-1 in potential applications as an anti-GBM therapy.
Fri Nov 01 00:00:00 UTC 2019
Immunophenotypic comparison of heterogenous non-sorted versus sorted mononuclear cells from human umbilical cord blood: a novel cell enrichment approach.
Cytotechnology
Indumathi S,Harikrishnan R,Rajkumar JS,Dhanasekaran M
17-1338 was used in Flow cytometry/Cell sorting to enrich the stem/progenitor cell population from human umbilical cord blood.
Thu Jan 01 00:00:00 UTC 2015
Evaluation of cancer stem cell markers CD133, CD44, CD24: association with AKT isoforms and radiation resistance in colon cancer cells.
PloS one
Sahlberg SH,Spiegelberg D,Glimelius B,Stenerlöw B,Nestor M
Fri Jan 09 00:00:00 UTC 2015
Comparative evaluation of differentiation potential of menstrual blood- versus bone marrow-derived stem cells into hepatocyte-like cells.
PloS one
Khanjani S,Khanmohammadi M,Zarnani AH,Akhondi MM,Ahani A,Ghaempanah Z,Naderi MM,Eghtesad S,Kazemnejad S
Sat Nov 08 00:00:00 UTC 2014
Detection and validation of circulating endothelial cells, a blood-based diagnostic marker of acute myocardial infarction.
PloS one
Li C,Wu Q,Liu B,Yao Y,Chen Y,Zhang H,Wang C,Cao J,Ge S
17-1338-42 was used in Flow cytometry/Cell sorting to investigate the diagnostic and prognostic value of circulating endothelial cell monitoring in acute myocardial infarction patients.
Tue Sep 03 00:00:00 UTC 2013
Inhibition of p38 MAPK activity promotes ex vivo expansion of human cord blood hematopoietic stem cells.
Annals of hematology
Zou J,Zou P,Wang J,Li L,Wang Y,Zhou D,Liu L
17-1338 was used in Flow cytometry/Cell sorting to examine the effect of inhibition of p38 mitogen-activated protein kinase on expansion of human umbilical cord blood CD133(+) cells.
Fri Jun 01 00:00:00 UTC 2012