Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Specificity: Recognizes human CD200R, a receptor for the CD200/OX2.
CD200R (OX2R) is a receptor for the OX-2 membrane glycoprotein, and both the receptor and substrate are cell surface glycoproteins containing two immunoglobulin-like domains. CD200R is restricted to the surfaces of myeloid lineage cells and the receptor-substrate interaction may function as a myeloid downregulatory signal. In mouse, CD200R is expressed primarily by peripheral blood monocytes and neutophils but also by other leucocytes including T-lymphocytes and mast cells. Further mouse studies of a gene related to CD200R suggests that the receptor may control myeloid function in a tissue-specific manner. Alternative splicing of CD200R results in multiple transcript variants. Diseases associated with CD200R dysfunction include primary hypertrophic osteoarthropathy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD200 cell surface glycoprotein receptor; CD200 R1; Cell surface glycoprotein CD200 receptor 1; Cell surface glycoprotein OX2 receptor 1; cell surface glycoprotein receptor CD200; MOX2 receptor
Gene Aliases: CD200R; CD200R1; CRTR2; HCRTR2; MOX2R; OX2R; UNQ2522/PRO6015
UniProt ID: (Human) Q8TD46
Entrez Gene ID: (Human) 131450
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support