Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Description: The M1/69 monoclonal antibody reacts with the mouse CD24 molecule, also known as Heat Stable Antigen (HSA). This 35-50 kDa molecule is anchored in the plasma membrane via phosphatidylinositol and is expressed by erythrocytes, thymocytes, peripheral lymphocytes and myeloid lineage. CD24 is a variably glycosylated molecule resulting in heterogeneity of molecular mass of this antigen on cells of different lineages and antibodies to CD24 exhibit subtle differences in staining level on lymphocyte populations. The expression of CD24 has been used to resolve stages of B lymphopoiesis in mouse bone marrow. It has been reported that P-selectin (CD62P) binds to CD24.
Applications Reported: This M1/69 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This M1/69 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Brilliant Violet™ 711 (BV711) is a tandem dye that emits at 713 nm and is intended for use on cytometers equipped with a violet (405 nm) laser. Please make sure that your instrument is capable of detecting this fluorochrome.
When using two or more Super Bright, Brilliant Violet™, Brilliant Ultra Violet™, or other polymer dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401-42) or Brilliant Stain Buffer™ (Product # 00-4409-75) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer or Brilliant Stain Buffer for more information.
Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.
Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222-49) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-54) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone-specific performance should be determined empirically.
Our internal testing suggests that Brilliant Violet™ 711 (BV711) is not compatible with methanol-based fixation.
Excitation: 407 nm; Emission: 713 nm; Laser: Violet Laser.
BRILLIANT VIOLET™ is a trademark or registered trademark of Becton, Dickinson and Company or its affiliates, and is used under license. Powered by Sirigen™.
CD24 is a 35-50 kDa glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed on a variety of cell types, including erythrocytes, thymocytes, peripheral lymphocytes, and cells of the myeloid lineage. It is variably glycosylated, resulting in heterogeneity in molecular mass across different cell lineages, which can affect antibody staining levels on lymphocyte populations. In the context of B-cell development, CD24 is expressed from the pro-B-cell stage in the bone marrow through to mature, surface Ig-positive B cells, with very low or negative expression on plasma cells. It is also present on the majority of B-lineage acute lymphoblastic leukemias, B-cell chronic lymphocytic leukemias (CCLs), and B-cell non-Hodgkin's lymphomas. CD24 is thought to play a role in regulating B-cell proliferation and maturation, as well as in controlling autoimmunity. Additionally, CD24 has been reported to interact with P-selectin (CD62P), indicating a potential role in cell adhesion and signaling. CD24 is a valuable marker for studying B-cell development, leukemia, lymphoma, and autoimmune regulation, as well as its interactions with other molecules like P-selectin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: CD24; cluster of differentiation 24; FLJ22950; FLJ43543; heat stable antigen; HSA; Ly-52; Lymphocyte antigen 52; M1/69-J11D heat stable antigen; MGC75043; Nectadrin; R13-Ag; Signal transducer CD24; X62 heat stable antigen
Gene Aliases: Cd24; Cd24a; HSA; Ly-52; nectadrin
UniProt ID: (Mouse) P24807
Entrez Gene ID: (Mouse) 12484
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support