Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CD24 Antibodies

          Invitrogen

          CD24 Monoclonal Antibody (eBioSN3 (SN3 A5-2H10)), Super Bright™ 436, eBioscience™

          13 Published Figures
          10 References
          View all (108) CD24 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CD24 Monoclonal Antibody (eBioSN3 (SN3 A5-2H10)), Super Bright™ 436, eBioscience™

          • Antibody Testing Data (1)
          • Published Figures (13)
          CD24 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD24 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 14

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD24 Antibody (62-0247-42) in Flow

          Normal human peripheral blood cells were stained with Mouse IgG1 kappa Isotype Control, Super Bright 436 (Product # 62-4714-82) (blue histogram) or CD24 Monoclonal Antibody, Super Bright 436 (purple histogram). Cells in the granulocyte gate were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD24 Antibody in Flow Cytometry (Flow)
          CD24 Antibody in Flow Cytometry (Flow)
          CD24 Antibody in Flow Cytometry (Flow)
          CD24 Antibody in Flow Cytometry (Flow)
          CD24 Antibody in Flow Cytometry (Flow)
          CD24 Antibody in Flow Cytometry (Flow)
          CD24 Antibody in Flow Cytometry (Flow)
          CD24 Antibody in Flow Cytometry (Flow)
          CD24 Antibody in Flow Cytometry (Flow)
          CD24 Antibody in Flow Cytometry (Flow)
          CD24 Antibody in Flow Cytometry (Flow)
          CD24 Antibody in Flow Cytometry (Flow)
          CD24 Antibody in Flow Cytometry (Flow)
          CD24 Antibody in Flow Cytometry (Flow)

          Product Details

          62-0247-42

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          5 µL (0.25 µg)/test
          View 10 publications 10 publications
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG1, kappa

          Recommended Isotype Control

          Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), Super Bright™ 436, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          eBioSN3 (SN3 A5-2H10)

          Conjugate

          Super Bright™ 436 Super Bright™ 436 Super Bright™ 436
          • Unconjugated
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • eFluor 450
          • FITC
          • NovaFluor Blue 510
          • NovaFluor Blue 555
          • NovaFluor Blue 585
          • NovaFluor Blue 690
          • NovaFluor Blue 725
          • NovaFluor Blue 760
          • NovaFluor Red 725
          • NovaFluor UV 765
          • NovaFluor Violet 800
          • NovaFluor Yellow 570
          • NovaFluor Yellow 590
          • NovaFluor Yellow 660
          • NovaFluor Yellow 755
          • PE 
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-eFluor 710
          • Super Bright 600
          • Super Bright 645
          • Request custom conjugation

          Excitation/Emission Max

          413/431 nm View spectra spectra

          Form

          Liquid

          Concentration

          5 µL/Test

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2, with BSA

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2802403

          Product Specific Information

          Description: The SN3 monoclonal antibody reacts with the human CD24 molecule, also known as Heat Stable Antigen (HSA). This 35-50 kDa molecule is anchored in the plasma membrane via phosphatidylinositol and is expressed by erythrocytes, thymocytes, peripheral lymphocytes and myeloid lineage. It has been reported that P-selectin (CD62P) binds to CD24.

          Applications Reported: This eBioSN3 (SN3 A5-2H10) antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This eBioSN3 (SN3 A5-2H10) antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 µL (0.25 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

          Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome.

          When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information.

          Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser

          Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.

          Target Information

          CD24 is a 35-50 kDa glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed on a variety of cell types, including erythrocytes, thymocytes, peripheral lymphocytes, and cells of the myeloid lineage. It is variably glycosylated, resulting in heterogeneity in molecular mass across different cell lineages, which can affect antibody staining levels on lymphocyte populations. In the context of B-cell development, CD24 is expressed from the pro-B-cell stage in the bone marrow through to mature, surface Ig-positive B cells, with very low or negative expression on plasma cells. It is also present on the majority of B-lineage acute lymphoblastic leukemias, B-cell chronic lymphocytic leukemias (CCLs), and B-cell non-Hodgkin's lymphomas. CD24 is thought to play a role in regulating B-cell proliferation and maturation, as well as in controlling autoimmunity. Additionally, CD24 has been reported to interact with P-selectin (CD62P), indicating a potential role in cell adhesion and signaling. CD24 is a valuable marker for studying B-cell development, leukemia, lymphoma, and autoimmune regulation, as well as its interactions with other molecules like P-selectin.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: CD24; CD24 antigen (small cell lung carcinoma cluster 4 antigen); FLJ22950; FLJ43543; MGC75043; Signal transducer CD24; Small cell lung carcinoma cluster 4 antigen

          View more View less

          Gene Aliases: CD24; CD24A

          View more View less

          UniProt ID: (Human) P25063

          View more View less

          Entrez Gene ID: (Human) 100133941

          View more View less

          Function(s)
          signal transducer activity protein binding protein kinase binding protein tyrosine kinase activator activity
          Process(es)
          response to hypoxia cell activation regulation of cytokine-mediated signaling pathway response to molecule of bacterial origin immune response-regulating cell surface receptor signaling pathway positive regulation of cytosolic calcium ion concentration Wnt signaling pathway single organismal cell-cell adhesion cell migration regulation of epithelial cell differentiation T cell costimulation B cell receptor transport into membrane raft chemokine receptor transport out of membrane raft negative regulation of transforming growth factor beta3 production positive regulation of activated T cell proliferation regulation of phosphorylation cholesterol homeostasis positive regulation of MAP kinase activity regulation of MAPK cascade response to estrogen respiratory burst positive regulation of protein tyrosine kinase activity glomerular visceral epithelial cell differentiation glomerular parietal epithelial cell differentiation intrinsic apoptotic signaling pathway positive regulation of nephron tubule epithelial cell differentiation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-2w6sl:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0