Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • RANKL (RANK Ligand) Antibodies

          Invitrogen

          CD254 (RANK Ligand) Monoclonal Antibody (IK22/5), Biotin, eBioscience™

          5 Published Figures
          7 References
          View all (77) RANKL (RANK Ligand) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CD254 (RANK Ligand) Monoclonal Antibody (IK22/5), Biotin, eBioscience™

          • Antibody Testing Data (1)
          • Published Figures (5)
          CD254 (RANK Ligand) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD254 (RANK Ligand) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD254 (RANK Ligand) Antibody (13-5952-82) in Flow

          Staining of 3-day unstimulated (left) and 3-day ConA activated (right) BALB/c splenocytes with 0.125 µg of Biotin Rat IgG2a Isotype Control (Product # 13-4321-82) (open histogram) or 0.125 µg of Anti-Mouse CD254 Biotin (filled histogram) followed by Streptavidin PE (Product # 12-4317-87). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD254 (RANK Ligand) Antibody in Flow Cytometry (Flow)
          CD254 (RANK Ligand) Antibody in Immunocytochemistry (ICC/IF)
          CD254 (RANK Ligand) Antibody in Immunocytochemistry (ICC/IF)
          CD254 (RANK Ligand) Antibody in Immunohistochemistry (IHC)
          CD254 (RANK Ligand) Antibody in Flow Cytometry (Flow)
          CD254 (RANK Ligand) Antibody in Immunomicroscopy (IM)

          Product Details

          13-5952-82

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          -
          View 2 publications 2 publications

          Flow Cytometry (Flow)

          0.25 µg/test
          View 2 publications 2 publications

          Immunomicroscopy (IM)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), Biotin, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          IK22/5

          Conjugate

          Biotin Biotin Biotin
          • Unconjugated
          • Functional Grade
          • PE 
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_466813

          Product Specific Information

          Description: The IK22/5 monoclonal antibody reacts with mouse CD254 also known as TRANCE (TNF-related activation-induced cytokine receptor) and RANKL (receptor activator of NF-kappa B ligand) and OPGL (Osteoprotegerin Ligand). TRANCE is a type II membrane protein with predicted molecular weight of 35 kDa. Its extracellular domain is most closely related to TRAIL, FasL and TNF. TRANCE mRNA is upregulated by TCR stimulation and is expressed abundantly by mouse T cells not B cells in mouse thymus and lymph nodes. TRANCE-deficient mouse lacks osteoclasts. In addition, TRANCE has been shown to be involved in the interactions between T-dendritic and T-B cells. It was also found to be critical in osteoclast differentiation. The TRANCE-RANK interaction regulates the expression of the anti-apoptotic molecule Bcl-xL.

          Applications Reported: This IK22/5 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: The IK22/5 antibody has been tested by flow cytometric analysis of mouse CD254 transfected cells. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          This gene encodes a member of the tumor necrosis factor (TNF) cytokine family which is a ligand for osteoprotegerin and functions as a key factor for osteoclast differentiation and activation. This protein was shown to be a dentritic cell survival factor and is involved in the regulation of T cell-dependent immune response. T cell activation was reported to induce expression of this gene and lead to an increase of osteoclastogenesis and bone loss. This protein was shown to activate antiapoptotic kinase AKT/PKB through a signaling complex involving SRC kinase and tumor necrosis factor receptor-associated factor (TRAF) 6, which indicated this protein may have a role in the regulation of cell apoptosis. Targeted disruption of the related gene in mice led to severe osteopetrosis and a lack of osteoclasts. The deficient mice exhibited defects in early differentiation of T and B lymphocytes, and failed to form lobulo-alveolar mammary structures during pregnancy. Two alternatively spliced transcript variants have been found.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CD254; H-RANK-L; M-Rank-L; ODF; OPG ligand; OPGL; Osteoclast differentiation factor; Osteoprotegerin ligand; rank-l; RANKL; receptor activator of NF-kappaB ligand; receptor activator of nuclear factor kappa B ligand; Receptor activator of nuclear factor kappa-B ligand; RP11-86N24.2; srankl; TNF-related activation-induced cytokine; TRANCE; Tumor necrosis factor ligand superfamily member 11; tumor necrosis factor-related activation-induced cytokine

          View more View less

          Gene Aliases: Ly109l; ODF; OPG; OPGL; RANKL; Tnfsf11; Trance

          View more View less

          UniProt ID: (Mouse) O35235

          View more View less

          Entrez Gene ID: (Mouse) 21943

          View more View less

          Function(s)
          cytokine activity tumor necrosis factor receptor binding protein binding tumor necrosis factor receptor superfamily binding
          Process(es)
          ossification osteoclast proliferation monocyte chemotaxis immune response activation of JUN kinase activity multicellular organism development organ morphogenesis cytokine-mediated signaling pathway cell differentiation osteoclast differentiation tumor necrosis factor-mediated signaling pathway mammary gland epithelial cell proliferation positive regulation of homotypic cell-cell adhesion paracrine signaling positive regulation of phosphorylation positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of MAP kinase activity bone resorption regulation of osteoclast differentiation positive regulation of osteoclast differentiation positive regulation of bone resorption positive regulation of transcription from RNA polymerase II promoter positive regulation of JNK cascade lymph node development positive regulation of T cell activation positive regulation of sequence-specific DNA binding transcription factor activity positive regulation of NF-kappaB transcription factor activity protein homooligomerization positive regulation of corticotropin-releasing hormone secretion positive regulation of protein kinase B signaling calcium ion homeostasis mammary gland alveolus development ERK1 and ERK2 cascade positive regulation of fever generation by positive regulation of prostaglandin secretion TNFSF11-mediated signaling pathway positive regulation of ERK1 and ERK2 cascade via TNFSF11-mediated signaling positive regulation of intracellular signal transduction positive regulation of osteoclast development
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0