Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Leinco Technologies
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Specificity: This non-therapeutic biosimilar antibody uses the same variable region sequence as the therapeutic antibody Nivolumab. Clone 5C4.B8 binds to the extracellular portion of Human/Cynomolgus PD-1 and does not bind to other IgG superfamily proteins. This product is for research use only.
Endotoxin Level: ≤ 1.0 EU/mg as determined by the LAL method. Purity: ≥95% monomer by analytical SEC.
Product Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using only In vitro protein free cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Pathogen Testing: To protect mouse colonies from infection by pathogens and to assure that experimental preclinical data is not affected by such pathogens, all of Leinco recombinant biosimilar antibodies are tested and guaranteed to be negative for all pathogens in the IDEXX IMPACT I Mouse Profile.
Storage and Handling: Functional grade biosimilar antibodies may be stored sterile as received at 2-8 C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80 C. Avoid Repeated Freeze Thaw Cycles.
This antibody has been tested in the following applications: Immunohistochemistry, Functional Assay, Flow Cytometry, Blocking.
Nivolumab (Opdivo) is a humanized monoclonal antibody that is approved by the U.S. Food and Drug Administration for the treatment of patients with unresectable or metastatic melanoma. Nivolumab binds to the human cell surface receptor PD-1 (Programmed Cell Death Protein 1) and blocks its interaction with PD-L1, resulting in the activation of T-cell-mediated immune responses against tumor cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support