Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MISIHNEEE NAFILDTLKK QWKGPDDILL GMFYDTDDAS FKWFDNSNMT FDKWTDQDDD EDLVDTCAFL HIKTGEWKKG NCEVSSVEGT LCKTAIPYKR KYLSDNHILI SALVIASTVI LTVLGAIIWF LYKKHSDSRF TTVFSTAPQS PYNEDCVLVV GEENEYPVQF D (63-232 aa encoded by BC020646)
CD302 is a C-type lectin receptor involved in cell adhesion and migration, as well as endocytosis and phagocytosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-type lectin BIMLEC; C-type lectin domain family 13 member A; C-type lectin domain family 13, member A; CD302; CD302 antigen; DEC205-associated C-type lectin 1; Type I transmembrane C-type lectin receptor DCL-1
Gene Aliases: 1110055L24Rik; AI159627; BIMLEC; CD302; CLEC13A; DCL-1; DCL1; KIAA0022
UniProt ID: (Human) Q8IX05, (Mouse) Q9DCG2
Entrez Gene ID: (Human) 9936, (Mouse) 66205
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support