Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Leinco Technologies
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Specificity: Clone 50946 recognizes human FcRL5 within the first 3 Ig domains. Clone 50946 does not cross-react with FcRL4.
Endotoxin Level: <1.0 EU/mg as determined by the LAL method.
Purity: ≥95% monomer by analytical SEC and >95% by SDS Page.
Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.
Fc receptor homologs (FcRH5), also known as IRTA2 proteins, are transmembrane proteins with sequence homology to classical Fc receptor families. FcRH5 function as activating or inhibitory receptors on discrete B cell populations and are involved in B-cell development and differentiation in peripheral lymphoid organs. FcRH5 expression is deregulated in multiple myeloma and Burkitt lymphoma cell lines with 1q21 abnormalities. FcRH5 may play a critical role for the pathogenesis of B-cell lymphoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BXMAS1; CD307e; Fc receptor homolog 5; Fc receptor-like 5; Fc receptor-like protein 5; FcR-like protein 5; FcRH5; Immune receptor translocation-associated protein 2; immunoglobulin superfamily receptor translocation associated 2 (IRTA2)
Gene Aliases: BXMAS1; CD307; CD307e; FCRH5; FCRL5; IRTA2; PRO820; UNQ503/PRO820
UniProt ID: (Human) Q96RD9
Entrez Gene ID: (Human) 83416
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support