Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • FLK1 (VEGF Receptor 2) Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.

          Invitrogen

          CD309 (FLK1) Monoclonal Antibody (Avas12a1), PE, eBioscience™

          40 Published Figures
          48 References
          View all (73) FLK1 (VEGF Receptor 2) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Datasheet
          Protocols

          Cite CD309 (FLK1) Monoclonal Antibody (Avas12a1), PE, eBioscience™

          • Antibody Testing Data (1)
          • Published Figures (40)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 41

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD309 (FLK1) Antibody (12-5821-81) in Flow

          Staining of bEnd-3 cell line with 0.5 µg of Rat IgG2a K Isotype Control PE (Product # 12-4321-80) (open histogram) or 0.5 µg of Anti-Mouse CD309 (FLK1) PE (filled histogram). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)

          Product Details

          12-5821-81

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.5 µg/test
          View 48 publications 48 publications
          Product Specifications

          Published species

          Human, Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), PE, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          Avas12a1

          Conjugate

          PE PE PE
          • Unconjugated
          • Alexa Fluor 700 
          • APC 
          • Biotin 
          • Super Bright 436
          • Super Bright 600
          • Request custom conjugation

          Excitation/Emission Max

          565/576 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Wet ice

          RRID

          AB_465966

          Product Specific Information

          Description: The Avas12a1 monoclonal antibody reacts with mouse Flk-1, also known as vascular endothelial growth factor receptor 2 (VEGFR2). Flk-1 is a receptor tyrosine kinase involved in vascular endothelial tissue development and is expressed on endothelial cells during embryonic stages and some endothelial tissues in the adult.

          Applications Reported: The Avas12a1 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: The Avas12a1 antibody has been tested by flow cytometric analysis of bEnd.3 cells. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          KDR (FLK1, VEGFR2) is a type III receptor tyrosine kinase. It binds vascular endothelial growth factor with high affinity in vitro, and is expressed early in development by endothelial cell precursors. FLK1 (VEGF Receptor 2) encodes one of the two receptors of the VEGF. This receptor, known as kinase insert domain receptor, is a type III receptor tyrosine kinase. The signalling and trafficking of this receptor are regulated by multiple factors, including Rab GTPase, P2Y purine nucleotide receptor, integrin alphaVbeta3, T-cell protein tyrosine phosphatase, etc. Mutations of FLK1 (VEGF Receptor 2) are implicated in infantile capillary hemangiomas.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0