Product References
Atf3 defines a population of pulmonary endothelial cells essential for lung regeneration.
eLife
Niethamer TK,Levin LI,Morley MP,Babu A,Zhou S,Morrisey EE
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Fri May 26 00:00:00 UTC 2023
Estradiol cycling drives female obesogenic adipocyte hyperplasia.
Cell reports
Saavedra-Peña RDM,Taylor N,Flannery C,Rodeheffer MS
25-0311-82 was used in Flow Cytometry to indicate that estrogen drives ERα-dependent obesogenic adipocyte hyperplasia in females, exacerbating obesity and contributing to the differential fat distribution between the sexes.
Tue Apr 25 00:00:00 UTC 2023
Estradiol cycling drives female obesogenic adipocyte hyperplasia.
Cell reports
Saavedra-Peña RDM,Taylor N,Flannery C,Rodeheffer MS
25-0311-82 was used in Flow Cytometry to indicate that estrogen drives ERα-dependent obesogenic adipocyte hyperplasia in females, exacerbating obesity and contributing to the differential fat distribution between the sexes.
Tue Apr 25 00:00:00 UTC 2023
Type H vessel/platelet-derived growth factor receptor β+ perivascular cell disintegration is involved in vascular injury and bone loss in radiation-induced bone damage.
Cell proliferation
Li J,Chen X,Ren L,Chen X,Wu T,Wang Y,Ren X,Cheng B,Xia J
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Sat Jul 01 00:00:00 UTC 2023
Transcriptomics and translatomics identify a robust inflammatory gene signature in brain endothelial cells after ischemic stroke.
Journal of neuroinflammation
Arbaizar-Rovirosa M,Gallizioli M,Lozano JJ,Sidorova J,Pedragosa J,Figuerola S,Chaparro-Cabanillas N,Boya P,Graupera M,Claret M,Urra X,Planas AM
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Mon Sep 11 00:00:00 UTC 2023
Plantamajoside attenuates cardiac fibrosis via inhibiting AGEs activated-RAGE/autophagy/EndMT pathway.
Phytotherapy research : PTR
Zhang L,Guo YN,Liu J,Wang LH,Wu HQ,Wang T,Deng B,Wang JY,Lu L,Chen ZX,He JQ,Liang BR,Li H,Huang YS,Yang ZQ,Xian SX,Wang LJ,Ye XH
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Wed Mar 01 00:00:00 UTC 2023
Suppression of preadipocyte determination by SOX4 limits white adipocyte hyperplasia in obesity.
iScience
He T,Wang S,Li S,Shen H,Hou L,Liu Y,Wei Y,Xie F,Zhang Z,Zhao Z,Mo C,Guo H,Huang Q,Zhang R,Shen D,Li B
25-0311-82 was used in Flow Cytometry to highlight the importance of SOX4 in regulating the adipocyte hyperplasia in obesity.
Fri Apr 21 00:00:00 UTC 2023
PD-L1 positive astrocytes attenuate inflammatory functions of PD-1 positive microglia in models of autoimmune neuroinflammation.
Nature communications
Linnerbauer M,Beyer T,Nirschl L,Farrenkopf D,Lößlein L,Vandrey O,Peter A,Tsaktanis T,Kebir H,Laplaud D,Oellinger R,Engleitner T,Alvarez JI,Rad R,Korn T,Hemmer B,Quintana FJ,Rothhammer V
25-0311-82 was used in Flow cytometry/Cell sorting to suggest the glial PD-L1/PD-1 axis as a potential therapeutic target for both acute and progressive MS stages.
Sat Sep 09 00:00:00 UTC 2023
The Wnt/TCF7L1 transcriptional repressor axis drives primitive endoderm formation by antagonizing naive and formative pluripotency.
Nature communications
Athanasouli P,Balli M,De Jaime-Soguero A,Boel A,Papanikolaou S,van der Veer BK,Janiszewski A,Vanhessche T,Francis A,El Laithy Y,Nigro AL,Aulicino F,Koh KP,Pasque V,Cosma MP,Verfaillie C,Zwijsen A,Heindryckx B,Nikolaou C,Lluis F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Fri Mar 03 00:00:00 UTC 2023
Isolation and in vitro characterization of murine young-adult long bone skeletal progenitors.
Frontiers in endocrinology
Loopmans S,Stockmans I,Carmeliet G,Stegen S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Fri Aug 19 00:00:00 UTC 2022
Lamin A/C-dependent chromatin architecture safeguards naïve pluripotency to prevent aberrant cardiovascular cell fate and function.
Nature communications
Wang Y,Elsherbiny A,Kessler L,Cordero J,Shi H,Serke H,Lityagina O,Trogisch FA,Mohammadi MM,El-Battrawy I,Backs J,Wieland T,Heineke J,Dobreva G
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Fri Nov 04 00:00:00 UTC 2022
Synovial Fibroblast Sialylation Regulates Cell Migration and Activation of Inflammatory Pathways in Arthritogenesis.
Frontiers in immunology
Wang Y,Pan P,Khan A,Çil Ç,Pineda MA
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Fri Apr 08 00:00:00 UTC 2022
Hematopoietic-Mesenchymal Signals Regulate the Properties of Mesenchymal Stem Cells.
International journal of molecular sciences
Kanazawa S,Okada H,Riu D,Mabuchi Y,Akazawa C,Iwata J,Hoshi K,Hikita A
25-0311-82 was used in Flow cytometry/Cell sorting to suggest that a hematopoietic-mesenchymal signal exists, and that the state of the HSCs is important for the stability of MSC properties.
Tue Jul 26 00:00:00 UTC 2022
Dynamic and adaptive cancer stem cell population admixture in colorectal neoplasia.
Cell stem cell
Vasquez EG,Nasreddin N,Valbuena GN,Mulholland EJ,Belnoue-Davis HL,Eggington HR,Schenck RO,Wouters VM,Wirapati P,Gilroy K,Lannagan TRM,Flanagan DJ,Najumudeen AK,Omwenga S,McCorry AMB,Easton A,Koelzer VH,East JE,Morton D,Trusolino L,Maughan T,Campbell AD,Loughrey MB,Dunne PD,Tsantoulis P,Huels DJ,Tejpar S,Sansom OJ,Leedham SJ
25-0311-82 was used in flow cytometry to propose a fitness landscape model where individual tumors have equilibrated stem cell population distributions along a CBC-RSC phenotypic axis.
Thu Aug 04 00:00:00 UTC 2022
PDGFRβ+ cells play a dual role as hematopoietic precursors and niche cells during mouse ontogeny.
Cell reports
Sá da Bandeira D,Kilpatrick AM,Marques M,Gomez-Salazar M,Ventura T,Gonzalez ZN,Stefancova D,Rossi F,Vermeren M,Vink CS,Beltran M,Henderson NC,Jung B,van der Linden R,van de Werken HJG,van Ijcken WFJ,Betsholtz C,Forbes SJ,Cuervo H,Crisan M
25-0311-82 was used in Flow cytometry/Cell sorting to demonstrate that PDGFRβ+ cells play a dual role in murine hematopoiesis.
Tue Jul 19 00:00:00 UTC 2022
Retrotransposon instability dominates the acquired mutation landscape of mouse induced pluripotent stem cells.
Nature communications
Gerdes P,Lim SM,Ewing AD,Larcombe MR,Chan D,Sanchez-Luque FJ,Walker L,Carleton AL,James C,Knaupp AS,Carreira PE,Nefzger CM,Lister R,Richardson SR,Polo JM,Faulkner GJ
25-0311-82 was used in flow cytometry to reveal the complete spectrum and potential significance of mutations acquired by miPSCs.
Sat Dec 03 00:00:00 UTC 2022
Inhibition of SRC-mediated integrin signaling in bone marrow niche enhances hematopoietic stem cell function.
iScience
Roy IM,Anu PV,Zaunz S,Reddi S,Giri AM,Sankar RS,Schouteden S,Huelsken J,Verfaillie CM,Khurana S
25-0311-82 was used in Flow Cytometry to establish a role of SRC-mediated integrin signaling in the transcriptional regulation of Sdf-1α.
Fri Oct 21 00:00:00 UTC 2022
MIF confers survival advantage to pancreatic CAFs by suppressing interferon pathway-induced p53-dependent apoptosis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Suresh V,Dash P,Suklabaidya S,Murmu KC,Sasmal PK,Jogdand GM,Parida D,Sethi M,Das B,Mohapatra D,Saha S,Prasad P,Satoskar A,Senapati S
25-0311-82 was used in Flow Cytometry to report that the PSCs of mouse or cancer-associated fibroblast cells (CAFs) of human expresses MIF and its receptors, whose expression gets upregulated upon LPS or TNF-α stimulation.
Mon Aug 01 00:00:00 UTC 2022
PI3K signaling specifies proximal-distal fate by driving a developmental gene regulatory network in SOX9+ mouse lung progenitors.
eLife
Khattar D,Fernandes S,Snowball J,Guo M,Gillen MC,Jain SS,Sinner D,Zacharias W,Swarr DT
25-0311-82 was used in Flow cytometry/Cell sorting to analyze paired gene expression (RNA-Seq) and chromatin accessibility (ATAC-Seq) data from SOX9+ epithelial progenitor cells (EPCs) during embryonic development in Mus musculus.
Wed Aug 17 00:00:00 UTC 2022
OCA-T1 and OCA-T2 are coactivators of POU2F3 in the tuft cell lineage.
Nature
Wu XS,He XY,Ipsaro JJ,Huang YH,Preall JB,Ng D,Shue YT,Sage J,Egeblad M,Joshua-Tor L,Vakoc CR
25-0311-82 was used in Flow cytometry/Cell sorting to reveal that the POU2F3-OCA-T complex is the master regulator of tuft cell identity and a molecular vulnerability of tuft-cell-like small-cell lung cancer.
Fri Jul 01 00:00:00 UTC 2022
Photoreceptor protection by mesenchymal stem cell transplantation identifies exosomal MiR-21 as a therapeutic for retinal degeneration.
Cell death and differentiation
Deng CL,Hu CB,Ling ST,Zhao N,Bao LH,Zhou F,Xiong YC,Chen T,Sui BD,Yu XR,Hu CH
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Mon Mar 01 00:00:00 UTC 2021
Anti-adipogenic signals at the onset of obesity-related inflammation in white adipose tissue.
Cellular and molecular life sciences : CMLS
Caputo T,Tran VDT,Bararpour N,Winkler C,Aguileta G,Trang KB,Giordano Attianese GMP,Wilson A,Thomas A,Pagni M,Guex N,Desvergne B,Gilardi F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Fri Jan 01 00:00:00 UTC 2021
SORLA is required for insulin-induced expansion of the adipocyte precursor pool in visceral fat.
The Journal of cell biology
Schmidt V,Horváth C,Dong H,Blüher M,Qvist P,Wolfrum C,Willnow TE
25-0311-82 was used in Flow Cytometry to identify the insulin-sensitizing receptor SORLA as a molecular factor explaining the distinct proliferative capacity of visceral white adipose tissue.
Mon Dec 06 00:00:00 UTC 2021
SORLA is required for insulin-induced expansion of the adipocyte precursor pool in visceral fat.
The Journal of cell biology
Schmidt V,Horváth C,Dong H,Blüher M,Qvist P,Wolfrum C,Willnow TE
25-0311-82 was used in Flow Cytometry to identify the insulin-sensitizing receptor SORLA as a molecular factor explaining the distinct proliferative capacity of visceral white adipose tissue.
Mon Dec 06 00:00:00 UTC 2021
Therapeutic Potential of Mesenchymal Stem Cells in a Pre-Clinical Model of Diabetic Kidney Disease and Obesity.
International journal of molecular sciences
Sávio-Silva C,Soinski-Sousa PE,Simplício-Filho A,Bastos RMC,Beyerstedt S,Rangel ÉB
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Thu Feb 04 00:00:00 UTC 2021
GRHL3 activates FSCN1 to relax cell-cell adhesions between migrating keratinocytes during wound reepithelialization.
JCI insight
Kashgari G,Venkatesh S,Refuerzo S,Pham B,Bayat A,Klein RH,Ramos R,Ta AP,Plikus MV,Wang PH,Andersen B
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Wed Sep 08 00:00:00 UTC 2021
The IRE1/XBP1 signaling axis promotes skeletal muscle regeneration through a cell non-autonomous mechanism.
eLife
Roy A,Tomaz da Silva M,Bhat R,Bohnert KR,Iwawaki T,Kumar A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Tue Nov 23 00:00:00 UTC 2021
Knockout RAGE alleviates cardiac fibrosis through repressing endothelial-to-mesenchymal transition (EndMT) mediated by autophagy.
Cell death & disease
Zhang L,He J,Wang J,Liu J,Chen Z,Deng B,Wei L,Wu H,Liang B,Li H,Huang Y,Lu L,Yang Z,Xian S,Wang L
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Tue May 11 00:00:00 UTC 2021
Tendon and motor phenotypes in the mouse model of recessive osteogenesis imperfecta.
eLife
Grol MW,Haelterman NA,Lim J,Munivez EM,Archer M,Hudson DM,Tufa SF,Keene DR,Lei K,Park D,Kuzawa CD,Ambrose CG,Eyre DR,Lee BH
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Wed May 26 00:00:00 UTC 2021
Loss of α2-6 sialylation promotes the transformation of synovial fibroblasts into a pro-inflammatory phenotype in arthritis.
Nature communications
Wang Y,Khan A,Antonopoulos A,Bouché L,Buckley CD,Filer A,Raza K,Li KP,Tolusso B,Gremese E,Kurowska-Stolarska M,Alivernini S,Dell A,Haslam SM,Pineda MA
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Tue Apr 20 00:00:00 UTC 2021
Interleukin-11-expressing fibroblasts have a unique gene signature correlated with poor prognosis of colorectal cancer.
Nature communications
Nishina T,Deguchi Y,Ohshima D,Takeda W,Ohtsuka M,Shichino S,Ueha S,Yamazaki S,Kawauchi M,Nakamura E,Nishiyama C,Kojima Y,Adachi-Akahane S,Hasegawa M,Nakayama M,Oshima M,Yagita H,Shibuya K,Mikami T,Inohara N,Matsushima K,Tada N,Nakano H
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Fri Apr 16 00:00:00 UTC 2021
Protocols for isolation and characterization of mouse placental hemogenic endothelial cells.
STAR protocols
Liang G,Huang B,Wang F,Liu F
25-0311-82 was used in Flow Cytometry to describe a protocol for isolation and characterization of the hemogenic endothelial (HE) cells from mouse placenta.
Fri Dec 17 00:00:00 UTC 2021
Protocols for isolation and characterization of mouse placental hemogenic endothelial cells.
STAR protocols
Liang G,Huang B,Wang F,Liu F
25-0311-82 was used in Flow Cytometry to describe a protocol for isolation and characterization of the hemogenic endothelial (HE) cells from mouse placenta.
Fri Dec 17 00:00:00 UTC 2021
SPARC regulation of PMN clearance protects from pristane-induced lupus and rheumatoid arthritis.
iScience
Sangaletti S,Botti L,Gulino A,Lecis D,Bassani B,Portararo P,Milani M,Cancila V,De Cecco L,Dugo M,Tripodo C,Colombo MP
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Fri Jun 25 00:00:00 UTC 2021
Restricted differentiative capacity of Wt1-expressing peritoneal mesothelium in postnatal and adult mice.
Scientific reports
Wilm TP,Tanton H,Mutter F,Foisor V,Middlehurst B,Ward K,Benameur T,Hastie N,Wilm B
25-0311-82 was used in Flow cytometry/Cell sorting to determine the spatio-temporal contribution of Wt1-expressing cells to the adult and postnatal intestine and peritoneal body wall, including the mesentery.
Thu Aug 05 00:00:00 UTC 2021
Evolution of fibroblasts in the lung metastatic microenvironment is driven by stage-specific transcriptional plasticity.
eLife
Shani O,Raz Y,Monteran L,Scharff Y,Levi-Galibov O,Megides O,Shacham H,Cohen N,Silverbush D,Avivi C,Sharan R,Madi A,Scherz-Shouval R,Barshack I,Tsarfaty I,Erez N
25-0311-82 was used in Flow Cytometry to identify Myc as a central regulator of fibroblast rewiring and found that stromal upregulation of Myc transcriptional networks is associated with disease progression in human breast cancer.
Fri Jun 25 00:00:00 UTC 2021
Protocols for endothelial cell isolation from mouse tissues: brain, choroid, lung, and muscle.
STAR protocols
Conchinha NV,Sokol L,Teuwen LA,Veys K,Dumas SJ,Meta E,García-Caballero M,Geldhof V,Chen R,Treps L,Borri M,de Zeeuw P,Falkenberg KD,Dubois C,Parys M,de Rooij LPMH,Rohlenova K,Goveia J,Schoonjans L,Dewerchin M,Eelen G,Li X,Kalucka J,Carmeliet P
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Fri Sep 17 00:00:00 UTC 2021
Testosterone metabolites differentially regulate obesogenesis and fat distribution.
Molecular metabolism
Sebo ZL,Rodeheffer MS
25-0311-81 was used in Flow Cytometry to clarify the role of testosterone and its downstream metabolites in the regulation of fat distribution.
Mon Feb 01 00:00:00 UTC 2021
Testosterone metabolites differentially regulate obesogenesis and fat distribution.
Molecular metabolism
Sebo ZL,Rodeheffer MS
25-0311-81 was used in Flow Cytometry to clarify the role of testosterone and its downstream metabolites in the regulation of fat distribution.
Mon Feb 01 00:00:00 UTC 2021
Mice with induced pulmonary morbidities display severe lung inflammation and mortality following exposure to SARS-CoV-2.
JCI insight
Falach R,Bar-On L,Lazar S,Kadar T,Mazor O,Aftalion M,Gur D,Evgy Y,Shifman O,Aminov T,Israeli O,Cohen-Gihon I,Zaide G,Gutman H,Vagima Y,Makdasi E,Stein D,Rosenfeld R,Alcalay R,Zahavy E,Levy H,Glinert I,Ben-Shmuel A,Israely T,Melamed S,Politi B,Achdout H,Yitzhaki S,Kronman C,Sabo T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Tue Jun 22 00:00:00 UTC 2021
The histone acetyltransferase HBO1 promotes efficient tip cell sprouting during angiogenesis.
Development (Cambridge, England)
Grant ZL,Hickey PF,Abeysekera W,Whitehead L,Lewis SM,Symons RCA,Baldwin TM,Amann-Zalcenstein D,Garnham AL,Naik SH,Smyth GK,Thomas T,Voss AK,Coultas L
25-0311-82 was used in Flow cytometry/Cell sorting to investigate the role of the histone acetyltransferase HBO1 (KAT7), which is important for activating genes during development and for histone H3 lysine 14 acetylation (H3K14ac).
Fri Oct 15 00:00:00 UTC 2021
Meningeal lymphatic vessels regulate brain tumor drainage and immunity.
Cell research
Hu X,Deng Q,Ma L,Li Q,Chen Y,Liao Y,Zhou F,Zhang C,Shao L,Feng J,He T,Ning W,Kong Y,Huo Y,He A,Liu B,Zhang J,Adams R,He Y,Tang F,Bian X,Luo J
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Sun Mar 01 00:00:00 UTC 2020
Therapy-Induced Senescence Drives Bone Loss.
Cancer research
Yao Z,Murali B,Ren Q,Luo X,Faget DV,Cole T,Ricci B,Thotala D,Monahan J,van Deursen JM,Baker D,Faccio R,Schwarz JK,Stewart SA
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Sun Mar 01 00:00:00 UTC 2020
The aging skin microenvironment dictates stem cell behavior.
Proceedings of the National Academy of Sciences of the United States of America
Ge Y,Miao Y,Gur-Cohen S,Gomez N,Yang H,Nikolova M,Polak L,Hu Y,Verma A,Elemento O,Krueger JG,Fuchs E
25-0311 was used in Flow cytometry/Cell sorting to highlight the importance of SC:niche interactions and favor a model where youthfulness of the niche microenvironment plays a dominant role in dictating the properties of its SCs and tissue health and fitness.
Tue Mar 10 00:00:00 UTC 2020
Membrane expression of the estrogen receptor ERα is required for intercellular communications in the mammary epithelium.
Development (Cambridge, England)
Gagniac L,Rusidzé M,Boudou F,Cagnet S,Adlanmerini M,Jeannot P,Gaide N,Giton F,Besson A,Weyl A,Gourdy P,Raymond-Letron I,Arnal JF,Brisken C,Lenfant F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Wed Mar 11 00:00:00 UTC 2020
DC-SCRIPT affects mammary organoids branching morphogenesis by modulating the FGFR1-pERK signaling axis.
Developmental biology
Tang C,van den Bijgaart RJE,Looman MWG,Triantis V,Nørskov Søndergaard J,Ansems M,Adema GJ
25-0311-82 was used in Flow Cytometry to investigated the molecular mechanism involved in the transcription regulator DC-SCRIPT mediated regulation of FGF2 induced mammary branching morphogenesis in a 3D organoid culture system.
Wed Jul 15 00:00:00 UTC 2020
Snail promotes the generation of vascular endothelium by breast cancer cells.
Cell death & disease
Chang Z,Zhang Y,Liu J,Zheng Y,Li H,Kong Y,Li P,Peng H,Shi Y,Cao B,Ran F,Chen Y,Song Y,Ye Q,Ding L
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Mon Jun 15 00:00:00 UTC 2020
Sierra: discovery of differential transcript usage from polyA-captured single-cell RNA-seq data.
Genome biology
Patrick R,Humphreys DT,Janbandhu V,Oshlack A,Ho JWK,Harvey RP,Lo KK
25-0311 was used in Flow cytometry/Cell sorting to present a computational pipeline, Sierra, that readily detects differential transcript usage from data generated by commonly used polyA-captured scRNA-seq technology.
Wed Jul 08 00:00:00 UTC 2020
Sierra: discovery of differential transcript usage from polyA-captured single-cell RNA-seq data.
Genome biology
Patrick R,Humphreys DT,Janbandhu V,Oshlack A,Ho JWK,Harvey RP,Lo KK
25-0311 was used in Flow cytometry/Cell sorting to present a computational pipeline, Sierra, that readily detects differential transcript usage from data generated by commonly used polyA-captured scRNA-seq technology.
Wed Jul 08 00:00:00 UTC 2020
Defining the role of pulmonary endothelial cell heterogeneity in the response to acute lung injury.
eLife
Niethamer TK,Stabler CT,Leach JP,Zepp JA,Morley MP,Babu A,Zhou S,Morrisey EE
25-0311 was used in Flow cytometry/Cell sorting to identify that different subtypes of endothelial cells exist in capillaries, and that some may be able to perform slightly different jobs during lung recovery.
Mon Feb 24 00:00:00 UTC 2020
Defining the role of pulmonary endothelial cell heterogeneity in the response to acute lung injury.
eLife
Niethamer TK,Stabler CT,Leach JP,Zepp JA,Morley MP,Babu A,Zhou S,Morrisey EE
25-0311 was used in Flow cytometry/Cell sorting to identify that different subtypes of endothelial cells exist in capillaries, and that some may be able to perform slightly different jobs during lung recovery.
Mon Feb 24 00:00:00 UTC 2020
Cardiac fibroblast proliferation rates and collagen expression mature early and are unaltered with advancing age.
JCI insight
Wu R,Ma F,Tosevska A,Farrell C,Pellegrini M,Deb A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Thu Dec 17 00:00:00 UTC 2020
Enhancer remodeling promotes tumor-initiating activity in NRF2-activated non-small cell lung cancers.
Nature communications
Okazaki K,Anzawa H,Liu Z,Ota N,Kitamura H,Onodera Y,Alam MM,Matsumaru D,Suzuki T,Katsuoka F,Tadaka S,Motoike I,Watanabe M,Hayasaka K,Sakurada A,Okada Y,Yamamoto M,Suzuki T,Kinoshita K,Sekine H,Motohashi H
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Fri Nov 20 00:00:00 UTC 2020
Myh11+ microvascular mural cells and derived mesenchymal stem cells promote retinal fibrosis.
Scientific reports
Ray HC,Corliss BA,Bruce AC,Kesting S,Dey P,Mansour J,Seaman SA,Smolko CM,Mathews C,Dey BK,Owens GK,Peirce SM,Yates PA
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Fri Sep 25 00:00:00 UTC 2020
Lymph Node Stromal Cells Generate Antigen-Specific Regulatory T Cells and Control Autoreactive T and B Cell Responses.
Cell reports
Nadafi R,Gago de Graça C,Keuning ED,Koning JJ,de Kivit S,Konijn T,Henri S,Borst J,Reijmers RM,van Baarsen LGM,Mebius RE
25-0311-82 was used in Flow Cytometry to uncover the ability of LNSCs to suppress the early activation of autoreactive immune cells and maintain peripheral tolerance.
Tue Mar 24 00:00:00 UTC 2020
FAM13A affects body fat distribution and adipocyte function.
Nature communications
Fathzadeh M,Li J,Rao A,Cook N,Chennamsetty I,Seldin M,Zhou X,Sangwung P,Gloudemans MJ,Keller M,Attie A,Yang J,Wabitsch M,Carcamo-Orive I,Tada Y,Lusis AJ,Shin MK,Molony CM,McLaughlin T,Reaven G,Montgomery SB,Reilly D,Quertermous T,Ingelsson E,Knowles JW
25-0311-82 was used in Flow Cytometry to indicate that GWAS-associated variants within the FAM13A locus alter adipose FAM13A expression, which in turn, regulates adipocyte differentiation and contribute to changes in body fat distribution.
Thu Mar 19 00:00:00 UTC 2020
Dynamics of Asymmetric and Symmetric Divisions of Muscle Stem Cells In Vivo and on Artificial Niches.
Cell reports
Evano B,Khalilian S,Le Carrou G,Almouzni G,Tajbakhsh S
25-0311 was used in Flow cytometry/Cell sorting to generate SNAP-tagged histone H3-reporter mice and find that, unlike fly germline stem cells, differential fate outcomes are associated with a symmetric distribution of the H3.1 and H3.3 histone variants in mouse muscle stem cells.
Tue Mar 10 00:00:00 UTC 2020
Dendritic Cells and Microglia Have Non-redundant Functions in the Inflamed Brain with Protective Effects of Type 1 cDCs.
Cell reports
Gallizioli M,Miró-Mur F,Otxoa-de-Amezaga A,Cugota R,Salas-Perdomo A,Justicia C,Brait VH,Ruiz-Jaén F,Arbaizar-Rovirosa M,Pedragosa J,Bonfill-Teixidor E,Gelderblom M,Magnus T,Cano E,Del Fresno C,Sancho D,Planas AM
25-0311 was used in Flow cytometry/Cell sorting to reveal aspects of the dynamics and functions of brain DCs in the regulation of inflammation and immunity.
Tue Oct 20 00:00:00 UTC 2020
The Lipid Handling Capacity of Subcutaneous Fat Is Programmed by mTORC2 during Development.
Cell reports
Hsiao WY,Jung SM,Tang Y,Haley JA,Li R,Li H,Calejman CM,Sanchez-Gurmaches J,Hung CM,Luciano AK,DeMambro V,Wellen KE,Rosen CJ,Zhu LJ,Guertin DA
25-0311-82 was used in Flow Cytometry to uncover a critical role for mTORC2 in setting SWAT lipid handling capacity.
Tue Oct 06 00:00:00 UTC 2020
Elucidating the fundamental fibrotic processes driving abdominal adhesion formation.
Nature communications
Foster DS,Marshall CD,Gulati GS,Chinta MS,Nguyen A,Salhotra A,Jones RE,Burcham A,Lerbs T,Cui L,King ME,Titan AL,Ransom RC,Manjunath A,Hu MS,Blackshear CP,Mascharak S,Moore AL,Norton JA,Kin CJ,Shelton AA,Januszyk M,Gurtner GC,Wernig G,Longaker MT
25-0311 was used in Flow cytometry/Cell sorting to present a systematic analysis of mouse and human adhesion tissues.
Thu Aug 13 00:00:00 UTC 2020
Engineering a haematopoietic stem cell niche by revitalizing mesenchymal stromal cells.
Nature cell biology
Nakahara F,Borger DK,Wei Q,Pinho S,Maryanovich M,Zahalka AH,Suzuki M,Cruz CD,Wang Z,Xu C,Boulais PE,Ma'ayan A,Greally JM,Frenette PS
25-0311-82 was used in Flow Cytometry to provide insight into the transcriptional regulation of the niche with implications for stem cell-based therapies.
Wed May 01 00:00:00 UTC 2019
Vascular endothelial growth factor pathway promotes osseointegration and CD31hiEMCNhi endothelium expansion in a mouse tibial implant model: an animal study.
The bone & joint journal
Ji G,Xu R,Niu Y,Li N,Ivashkiv L,Bostrom MPG,Greenblatt MB,Yang X
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Mon Jul 01 00:00:00 UTC 2019
LPS‑induced upregulation of the TLR4 signaling pathway inhibits osteogenic differentiation of human periodontal ligament stem cells under inflammatory conditions.
International journal of molecular medicine
Yu B,Li Q,Zhou M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Sat Jun 01 00:00:00 UTC 2019
Cell-specific ablation of Hsp47 defines the collagen-producing cells in the injured heart.
JCI insight
Khalil H,Kanisicak O,Vagnozzi RJ,Johansen AK,Maliken BD,Prasad V,Boyer JG,Brody MJ,Schips T,Kilian KK,Correll RN,Kawasaki K,Nagata K,Molkentin JD
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Thu Aug 08 00:00:00 UTC 2019
Single-cell expression profiling reveals dynamic flux of cardiac stromal, vascular and immune cells in health and injury.
eLife
Farbehi N,Patrick R,Dorison A,Xaymardan M,Janbandhu V,Wystub-Lis K,Ho JW,Nordon RE,Harvey RP
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Tue Mar 26 00:00:00 UTC 2019
Growth Factor Screening in Dystrophic Muscles Reveals PDGFB/PDGFRB-Mediated Migration of Interstitial Stem Cells.
International journal of molecular sciences
Camps J,Grosemans H,Gijsbers R,Maes C,Sampaolesi M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Tue Mar 05 00:00:00 UTC 2019
PDGFRα+ stromal adipocyte progenitors transition into epithelial cells during lobulo-alveologenesis in the murine mammary gland.
Nature communications
Joshi PA,Waterhouse PD,Kasaian K,Fang H,Gulyaeva O,Sul HS,Boutros PC,Khokha R
25-0311 was used in Flow cytometry/Cell sorting to identify the transition of a PDGFRα+ mesenchymal cell population into mammary epithelial progenitors in mice.
Mon Apr 15 00:00:00 UTC 2019
PDGFRα+ stromal adipocyte progenitors transition into epithelial cells during lobulo-alveologenesis in the murine mammary gland.
Nature communications
Joshi PA,Waterhouse PD,Kasaian K,Fang H,Gulyaeva O,Sul HS,Boutros PC,Khokha R
25-0311 was used in Flow cytometry/Cell sorting to identify the transition of a PDGFRα+ mesenchymal cell population into mammary epithelial progenitors in mice.
Mon Apr 15 00:00:00 UTC 2019
Global Transcriptomic Profiling of the Bone Marrow Stromal Microenvironment during Postnatal Development, Aging, and Inflammation.
Cell reports
Helbling PM,Piñeiro-Yáñez E,Gerosa R,Boettcher S,Al-Shahrour F,Manz MG,Nombela-Arrieta C
25-0311 was used in Flow cytometry/Cell sorting to reveal (1) molecular fingerprints defining cell-specific anatomical and functional features, (2) a radical reprogramming of pro-hematopoietic, immune, and matrisomic transcriptional programs during the transition from juvenile stages to adulthood, and (3) the aging-driven progressive upregulation of pro-inflammatory gene expression in stroma.
Tue Dec 03 00:00:00 UTC 2019
Inducing cancer indolence by targeting mitochondrial Complex I is potentiated by blocking macrophage-mediated adaptive responses.
Nature communications
Kurelac I,Iommarini L,Vatrinet R,Amato LB,De Luise M,Leone G,Girolimetti G,Umesh Ganesh N,Bridgeman VL,Ombrato L,Columbaro M,Ragazzi M,Gibellini L,Sollazzo M,Feichtinger RG,Vidali S,Baldassarre M,Foriel S,Vidone M,Cossarizza A,Grifoni D,Kofler B,Malanchi I,Porcelli AM,Gasparre G
25-0311-82 was used in Flow Cytometry to demonstrate that respiratory complex I-deficient tumours survive and carry out angiogenesis, despite their inability to stabilise Hypoxia Inducible Factor-1 alpha.
Fri Feb 22 00:00:00 UTC 2019
Nidogen-1 Contributes to the Interaction Network Involved in Pro-B Cell Retention in the Peri-sinusoidal Hematopoietic Stem Cell Niche.
Cell reports
Balzano M,De Grandis M,Vu Manh TP,Chasson L,Bardin F,Farina A,Sergé A,Bidaut G,Charbord P,Hérault L,Bailly AL,Cartier-Michaud A,Boned A,Dalod M,Duprez E,Genever P,Coles M,Bajenoff M,Xerri L,Aurrand-Lions M,Schiff C,Mancini SJC
25-0311 was used in Flow cytometry/Cell sorting to study pro-B cells in their support of stroll cells and their interaction network allowing their retention.
Tue Mar 19 00:00:00 UTC 2019
Omega-3 Fatty Acids Activate Ciliary FFAR4 to Control Adipogenesis.
Cell
Hilgendorf KI,Johnson CT,Mezger A,Rice SL,Norris AM,Demeter J,Greenleaf WJ,Reiter JF,Kopinke D,Jackson PK
25-0311-82 was used in Flow Cytometry to investigate whether preadipocytes use their cilia to sense and respond to external cues to remodel white adipose tissue.
Wed Nov 27 00:00:00 UTC 2019
Establishing a Link between Endothelial Cell Metabolism and Vascular Behaviour in a Type 1 Diabetes Mouse Model.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Silva C,Sampaio-Pinto V,Andrade S,Rodrigues I,Costa R,Guerreiro S,Carvalho E,Pinto-do-Ó P,Nascimento DS,Soares R
25-0311 was used in Flow cytometry/Cell sorting to study the angiogenic and metabolic gene expression profile of endothelial cells from a diabetes model.
Wed Apr 03 00:00:00 UTC 2019
Metastatic cancers promote cachexia through ZIP14 upregulation in skeletal muscle.
Nature medicine
Wang G,Biswas AK,Ma W,Kandpal M,Coker C,Grandgenett PM,Hollingsworth MA,Jain R,Tanji K,Lόpez-Pintado S,Borczuk A,Hebert D,Jenkitkasemwong S,Hojyo S,Davuluri RV,Knutson MD,Fukada T,Acharyya S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Fri Jun 01 00:00:00 UTC 2018
Targeting skeletal endothelium to ameliorate bone loss.
Nature medicine
Xu R,Yallowitz A,Qin A,Wu Z,Shin DY,Kim JM,Debnath S,Ji G,Bostrom MP,Yang X,Zhang C,Dong H,Kermani P,Lalani S,Li N,Liu Y,Poulos MG,Wach A,Zhang Y,Inoue K,Di Lorenzo A,Zhao B,Butler JM,Shim JH,Glimcher LH,Greenblatt MB
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Fri Jun 01 00:00:00 UTC 2018
Adrenergic nerve degeneration in bone marrow drives aging of the hematopoietic stem cell niche.
Nature medicine
Maryanovich M,Zahalka AH,Pierce H,Pinho S,Nakahara F,Asada N,Wei Q,Wang X,Ciero P,Xu J,Leftin A,Frenette PS
25-0311 was used in Flow cytometry/Cell sorting to determine whether haematopoietic stem cell ageing depends on bone marrow innervation by the sympathetic nervous system.
Fri Jun 01 00:00:00 UTC 2018
Regeneration of the lung alveolus by an evolutionarily conserved epithelial progenitor.
Nature
Zacharias WJ,Frank DB,Zepp JA,Morley MP,Alkhaleel FA,Kong J,Zhou S,Cantu E,Morrisey EE
25-0311 was used in Flow cytometry/Cell sorting to identify the AEP lineage as an evolutionarily conserved alveolar progenitor that represents a new target for human lung regeneration strategies.
Thu Mar 08 00:00:00 UTC 2018
YAP/TAZ-Dependent Reprogramming of Colonic Epithelium Links ECM Remodeling to Tissue Regeneration.
Cell stem cell
Yui S,Azzolin L,Maimets M,Pedersen MT,Fordham RP,Hansen SL,Larsen HL,Guiu J,Alves MRP,Rundsten CF,Johansen JV,Li Y,Madsen CD,Nakamura T,Watanabe M,Nielsen OH,Schweiger PJ,Piccolo S,Jensen KB
25-0311 was used in Flow cytometry/Cell sorting to demonstrate that the epithelium is transiently reprogrammed into a primitive state during colonic regeneration.
Thu Jan 04 00:00:00 UTC 2018
Endothelial cells in the innate response to allergens and initiation of atopic asthma.
The Journal of clinical investigation
Asosingh K,Weiss K,Queisser K,Wanner N,Yin M,Aronica M,Erzurum S
25-0311 was used in Flow cytometry/Cell sorting to identify that lung endothelium and proangiogenic haematopoietic progenitor cells in the airway sense allergen and elicit an angiogenic response that is central to the innate non-immune origins of Th2 inflammation.
Mon Jul 02 00:00:00 UTC 2018
Indian Hedgehog Suppresses a Stromal Cell-Driven Intestinal Immune Response.
Cellular and molecular gastroenterology and hepatology
Westendorp BF,Büller NVJA,Karpus ON,van Dop WA,Koster J,Versteeg R,Koelink PJ,Snel CY,Meisner S,Roelofs JJTH,Uhmann A,Ver Loren van Themaat E,Heijmans J,Hahn H,Muncan V,Wildenberg ME,van den Brink GR
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Thu Mar 31 00:00:00 UTC 2022
VASP regulates leukocyte infiltration, polarization, and vascular repair after ischemia.
The Journal of cell biology
Laban H,Weigert A,Zink J,Elgheznawy A,Schürmann C,Günther L,Abdel Malik R,Bothur S,Wingert S,Bremer R,Rieger MA,Brüne B,Brandes RP,Fleming I,Benz PM
25-0311 was used in Immunohistochemistry to investigate the role of vasodilator-stimulated phosphoprotein in vascular repair.
Mon Apr 02 00:00:00 UTC 2018
Stem cell factor is selectively secreted by arterial endothelial cells in bone marrow.
Nature communications
Xu C,Gao X,Wei Q,Nakahara F,Zimmerman SE,Mar J,Frenette PS
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Fri Jun 22 00:00:00 UTC 2018
Age-specific biological and molecular profiling distinguishes paediatric from adult acute myeloid leukaemias.
Nature communications
Chaudhury S,O'Connor C,Cañete A,Bittencourt-Silvestre J,Sarrou E,Prendergast Á,Choi J,Johnston P,Wells CA,Gibson B,Keeshan K
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Tue Dec 11 00:00:00 UTC 2018
Adipocyte hypertrophy and lipid dynamics underlie mammary gland remodeling after lactation.
Nature communications
Zwick RK,Rudolph MC,Shook BA,Holtrup B,Roth E,Lei V,Van Keymeulen A,Seewaldt V,Kwei S,Wysolmerski J,Rodeheffer MS,Horsley V
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Tue Sep 04 00:00:00 UTC 2018
A mesodermal fate map for adipose tissue.
Development (Cambridge, England)
Sebo ZL,Jeffery E,Holtrup B,Rodeheffer MS
25-0311-82 was used in Flow cytometry/Cell sorting to present evidence that 50% of interscapular brown adipocytes are derived from progenitors of the central dermomyotome.
Fri Aug 17 00:00:00 UTC 2018
Neonatally imprinted stromal cell subsets induce tolerogenic dendritic cells in mesenteric lymph nodes.
Nature communications
Pezoldt J,Pasztoi M,Zou M,Wiechers C,Beckstette M,Thierry GR,Vafadarnejad E,Floess S,Arampatzi P,Buettner M,Schweer J,Fleissner D,Vital M,Pieper DH,Basic M,Dersch P,Strowig T,Hornef M,Bleich A,Bode U,Pabst O,Bajénoff M,Saliba AE,Huehn J
25-0311 was used in Flow cytometry/Cell sorting to study the function of the neonatal imprinting of stromal cells in mesenteric lymph nodes.
Tue Sep 25 00:00:00 UTC 2018
Matrix stiffness controls lymphatic vessel formation through regulation of a GATA2-dependent transcriptional program.
Nature communications
Frye M,Taddei A,Dierkes C,Martinez-Corral I,Fielden M,Ortsäter H,Kazenwadel J,Calado DP,Ostergaard P,Salminen M,He L,Harvey NL,Kiefer F,Mäkinen T
25-0311 was used in Flow cytometry/Cell sorting to study whether extracellular matrix stiffness impacts lymphatic vascular development.
Tue Apr 17 00:00:00 UTC 2018
Collagenase-based Single Cell Isolation of Primary Murine Brain Endothelial Cells Using Flow Cytometry.
Bio-protocol
Czupalla CJ,Yousef H,Wyss-Coray T,Butcher EC
25-0311-82 was used in Flow Cytometry to report that the integrity of the brain endothelium is compromised in a plethora of neuropathologies.
Tue Nov 20 00:00:00 UTC 2018
Lrig1 marks a population of gastric epithelial cells capable of long-term tissue maintenance and growth in vitro.
Scientific reports
Schweiger PJ,Clement DL,Page ME,Schepeler T,Zou X,Sirokmány G,Watt FM,Jensen KB
25-0311 was used in Flow cytometry/Cell sorting to show that Lrig1 is expressed primarily by differentiated cells, but that these cells can be recruited to contribute to gastric epithelium maintenance.
Mon Oct 15 00:00:00 UTC 2018
Partial depletion of CD206-positive M2-like macrophages induces proliferation of beige progenitors and enhances browning after cold stimulation.
Scientific reports
Igarashi Y,Nawaz A,Kado T,Bilal M,Kuwano T,Yamamoto S,Sasahara M,Jiuxiang X,Inujima A,Koizumi K,Imura J,Shibahara N,Usui I,Fujisaka S,Tobe K
25-0311 was used in Flow cytometry/Cell sorting to examine the role of CD206+ M2-like macrophages in the cold-induced browning of white adipose tissue.
Mon Oct 01 00:00:00 UTC 2018
Gpr124 is essential for blood-brain barrier integrity in central nervous system disease.
Nature medicine
Chang J,Mancuso MR,Maier C,Liang X,Yuki K,Yang L,Kwong JW,Wang J,Rao V,Vallon M,Kosinski C,Zhang JJ,Mah AT,Xu L,Li L,Gholamin S,Reyes TF,Li R,Kuhnert F,Han X,Yuan J,Chiou SH,Brettman AD,Daly L,Corney DC,Cheshier SH,Shortliffe LD,Wu X,Snyder M,Chan P,Giffard RG,Chang HY,Andreasson K,Kuo CJ
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Sat Apr 01 00:00:00 UTC 2017
Tumour ischaemia by interferon-γ resembles physiological blood vessel regression.
Nature
Kammertoens T,Friese C,Arina A,Idel C,Briesemeister D,Rothe M,Ivanov A,Szymborska A,Patone G,Kunz S,Sommermeyer D,Engels B,Leisegang M,Textor A,Fehling HJ,Fruttiger M,Lohoff M,Herrmann A,Yu H,Weichselbaum R,Uckert W,Hübner N,Gerhardt H,Beule D,Schreiber H,Blankenstein T
Thu May 04 00:00:00 UTC 2017
Intratumoural heterogeneity generated by Notch signalling promotes small-cell lung cancer.
Nature
Lim JS,Ibaseta A,Fischer MM,Cancilla B,O'Young G,Cristea S,Luca VC,Yang D,Jahchan NS,Hamard C,Antoine M,Wislez M,Kong C,Cain J,Liu YW,Kapoun AM,Garcia KC,Hoey T,Murriel CL,Sage J
Thu May 18 00:00:00 UTC 2017
Stem Cell Lineage Infidelity Drives Wound Repair and Cancer.
Cell
Ge Y,Gomez NC,Adam RC,Nikolova M,Yang H,Verma A,Lu CP,Polak L,Yuan S,Elemento O,Fuchs E
25-0311 was used in Flow cytometry/Cell sorting to investigate the homeostatic mechanisms of tissue regeneration and wound repair by tissue stem cells in the skin, that can be hijacked by cancer cells.
Thu May 04 00:00:00 UTC 2017
Increased Vascular Permeability in the Bone Marrow Microenvironment Contributes to Disease Progression and Drug Response in Acute Myeloid Leukemia.
Cancer cell
Passaro D,Di Tullio A,Abarrategi A,Rouault-Pierre K,Foster K,Ariza-McNaughton L,Montaner B,Chakravarty P,Bhaw L,Diana G,Lassailly F,Gribben J,Bonnet D
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Mon Sep 11 00:00:00 UTC 2017
Differential cytokine contributions of perivascular haematopoietic stem cell niches.
Nature cell biology
Asada N,Kunisaki Y,Pierce H,Wang Z,Fernandez NF,Birbrair A,Ma'ayan A,Frenette PS
25-0311 was used in Flow cytometry/Cell sorting to demonstrate that the effects of cytokines regulating haematopoietic stem cell functions are dependent on the producing cell sources.
Wed Mar 01 00:00:00 UTC 2017
Adipocyte Accumulation in the Bone Marrow during Obesity and Aging Impairs Stem Cell-Based Hematopoietic and Bone Regeneration.
Cell stem cell
Ambrosi TH,Scialdone A,Graja A,Gohlke S,Jank AM,Bocian C,Woelk L,Fan H,Logan DW,Schürmann A,Saraiva LR,Schulz TJ
25-0311 was used in Flow cytometry/Cell sorting to specify the cellular identities of the adipogenic and osteogenic lineages of the bone.
Thu Jun 01 00:00:00 UTC 2017
Preexisting endothelial cells mediate cardiac neovascularization after injury.
The Journal of clinical investigation
He L,Huang X,Kanisicak O,Li Y,Wang Y,Li Y,Pu W,Liu Q,Zhang H,Tian X,Zhao H,Liu X,Zhang S,Nie Y,Hu S,Miao X,Wang QD,Wang F,Chen T,Xu Q,Lui KO,Molkentin JD,Zhou B
Tue Aug 01 00:00:00 UTC 2017
Obesity reversibly depletes the basal cell population and enhances mammary epithelial cell estrogen receptor alpha expression and progenitor activity.
Breast cancer research : BCR
Chamberlin T,D'Amato JV,Arendt LM
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Wed Nov 29 00:00:00 UTC 2017
Mesenchymal Stem Cell Therapy Protects Lungs from Radiation-Induced Endothelial Cell Loss by Restoring Superoxide Dismutase 1 Expression.
Antioxidants & redox signaling
Klein D,Steens J,Wiesemann A,Schulz F,Kaschani F,Röck K,Yamaguchi M,Wirsdörfer F,Kaiser M,Fischer JW,Stuschke M,Jendrossek V
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Mon Apr 10 00:00:00 UTC 2017
Cornea lymphatics drive the CD8+ T-cell response to herpes simplex virus-1.
Immunology and cell biology
Gurung HR,Carr MM,Carr DJ
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Sun Jan 01 00:00:00 UTC 2017
β3-Adrenergic Regulation of EPC Features Through Manipulation of the Bone Marrow MSC Niche.
Journal of cellular biochemistry
Vafaei R,Nassiri SM,Siavashi V
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Fri Dec 01 00:00:00 UTC 2017
Adamts1 responds to systemic cues and gates adipogenesis.
Adipocyte
Wong JC,Feldman BJ
25-0311 was used in Flow cytometry/Cell sorting to identify an Adamts 1 signal as a glucocorticoid and diet responsive regulator of an extracellular relay system that modulates the initiation of this intracellular adipogenesis program in adipocyte precursor cells.
Mon Oct 02 00:00:00 UTC 2017
Puberty is an important developmental period for the establishment of adipose tissue mass and metabolic homeostasis.
Adipocyte
Holtrup B,Church CD,Berry R,Colman L,Jeffery E,Bober J,Rodeheffer MS
25-0311-82 was used in Flow Cytometry to study the effects of exposure to obesogenic stimuli during critical developmental periods on adipose tissue function.
Mon Jul 03 00:00:00 UTC 2017
Etv2-miR-130a-Jarid2 cascade regulates vascular patterning during embryogenesis.
PloS one
Singh BN,Tahara N,Kawakami Y,Das S,Koyano-Nakagawa N,Gong W,Garry MG,Garry DJ
25-0311 was used in Flow cytometry/Cell sorting to examine the Etv2-Cre mediated ablation of DicerL/L, and characterise the perturbed vascular patterning during embryogenesis.
Thu Jan 04 00:00:00 UTC 2018
CD206+ M2-like macrophages regulate systemic glucose metabolism by inhibiting proliferation of adipocyte progenitors.
Nature communications
Nawaz A,Aminuddin A,Kado T,Takikawa A,Yamamoto S,Tsuneyama K,Igarashi Y,Ikutani M,Nishida Y,Nagai Y,Takatsu K,Imura J,Sasahara M,Okazaki Y,Ueki K,Okamura T,Tokuyama K,Ando A,Matsumoto M,Mori H,Nakagawa T,Kobayashi N,Saeki K,Usui I,Fujisaka S,Tobe K
25-0311 was used in Flow cytometry/Cell sorting to report that M2-like macrophages, create a microenvironment that inhibits proliferation of adipocyte progenitors due to the secretion of TGF-β1.
Fri Aug 18 00:00:00 UTC 2017
Single-cell profiling reveals GPCR heterogeneity and functional patterning during neuroinflammation.
JCI insight
Tischner D,Grimm M,Kaur H,Staudenraus D,Carvalho J,Looso M,Günther S,Wanke F,Moos S,Siller N,Breuer J,Schwab N,Zipp F,Waisman A,Kurschus FC,Offermanns S,Wettschureck N
25-0311 was used in Flow cytometry/Cell sorting to identify functionally distinct adaptive GPCR heterogeneity patternation in mice.
Thu Aug 03 00:00:00 UTC 2017
Brown adipocytes can display a mammary basal myoepithelial cell phenotype in vivo.
Molecular metabolism
Li L,Li B,Li M,Niu C,Wang G,Li T,Król E,Jin W,Speakman JR
Sun Oct 01 00:00:00 UTC 2017
Gli1 identifies osteogenic progenitors for bone formation and fracture repair.
Nature communications
Shi Y,He G,Lee WC,McKenzie JA,Silva MJ,Long F
25-0311 was used in Flow cytometry/Cell sorting to study how Gli1 marks mesenchymal progenitors responsible for both normal bone formation and fracture repair.
Mon Dec 11 00:00:00 UTC 2017
Identification of blood vascular endothelial stem cells by the expression of protein C receptor.
Cell research
Yu QC,Song W,Wang D,Zeng YA
25-0311 was used in Flow cytometry/Cell sorting to investigate the existence and identity of vascular endothelial stem cells, showing that these cells express the protein C receptor.
Sat Oct 01 00:00:00 UTC 2016
Strand-specific in vivo screen of cancer-associated miRNAs unveils a role for miR-21(∗) in SCC progression.
Nature cell biology
Ge Y,Zhang L,Nikolova M,Reva B,Fuchs E
25-0311 was used in Flow cytometry/Cell sorting to identify which of the microRNAs abundantly expressed in squamous cell carcinoma are key drivers in malignant progression.
Fri Jan 01 00:00:00 UTC 2016
Skin Adipocyte Stem Cell Self-Renewal Is Regulated by a PDGFA/AKT-Signaling Axis.
Cell stem cell
Rivera-Gonzalez GC,Shook BA,Andrae J,Holtrup B,Bollag K,Betsholtz C,Rodeheffer MS,Horsley V
25-0311 was used in Flow cytometry/Cell sorting to uncover PDGFA as a regulator of adipocyte stem cell self-renewal and proliferation.
Thu Dec 01 00:00:00 UTC 2016
OX40 ligand newly expressed on bronchiolar progenitors mediates influenza infection and further exacerbates pneumonia.
EMBO molecular medicine
Hirano T,Kikuchi T,Tode N,Santoso A,Yamada M,Mitsuhashi Y,Komatsu R,Kawabe T,Tanimoto T,Ishii N,Tanaka Y,Nishimura H,Nukiwa T,Watanabe A,Ichinose M
25-0311 was used in Flow cytometry/Cell sorting to show that the influenza-induced expression of OX40L on bronchiolar progenitors has pathogenic value to develop a novel therapeutic approach against influenza.
Fri Apr 01 00:00:00 UTC 2016
Dynamic Patterns of Clonal Evolution in Tumor Vasculature Underlie Alterations in Lymphocyte-Endothelial Recognition to Foster Tumor Immune Escape.
Cancer research
Corey DM,Rinkevich Y,Weissman IL
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Tue Mar 15 00:00:00 UTC 2016
Endothelial cell survival during angiogenesis requires the pro-survival protein MCL1.
Cell death and differentiation
Watson EC,Whitehead L,Adams RH,Dewson G,Coultas L
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Mon Aug 01 00:00:00 UTC 2016
DUX4 induces a transcriptome more characteristic of a less-differentiated cell state and inhibits myogenesis.
Journal of cell science
Knopp P,Krom YD,Banerji CR,Panamarova M,Moyle LA,den Hamer B,van der Maarel SM,Zammit PS
25-0311 was used in Flow cytometry/Cell sorting to investigate the role of DUX4 in the inhibition of myogenesis and in facioscapulohumeral muscular dystrophy.
Sat Oct 15 00:00:00 UTC 2016
Pdgfrb-Cre targets lymphatic endothelial cells of both venous and non-venous origins.
Genesis (New York, N.Y. : 2000)
Ulvmar MH,Martinez-Corral I,Stanczuk L,Mäkinen T
25-0311 was used in Flow cytometry/Cell sorting to show that Pdgfrb-Cre does not target YS HemEC or YS-derived erythro-myeloid progenitors.
Wed Jun 01 00:00:00 UTC 2016
Genetic lineage tracing defines myofibroblast origin and function in the injured heart.
Nature communications
Kanisicak O,Khalil H,Ivey MJ,Karch J,Maliken BD,Correll RN,Brody MJ,J Lin SC,Aronow BJ,Tallquist MD,Molkentin JD
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Fri Jul 22 00:00:00 UTC 2016
Endothelial to mesenchymal transition is common in atherosclerotic lesions and is associated with plaque instability.
Nature communications
Evrard SM,Lecce L,Michelis KC,Nomura-Kitabayashi A,Pandey G,Purushothaman KR,d'Escamard V,Li JR,Hadri L,Fujitani K,Moreno PR,Benard L,Rimmele P,Cohain A,Mecham B,Randolph GJ,Nabel EG,Hajjar R,Fuster V,Boehm M,Kovacic JC
25-0311 was used in Flow cytometry/Cell sorting to conclude that endothelial to mesenchymal transition contributes to atherosclerotic patho-biology and is associated with complex plaques.
Fri Jun 24 00:00:00 UTC 2016
A cell-autonomous tumour suppressor role of RAF1 in hepatocarcinogenesis.
Nature communications
Jeric I,Maurer G,Cavallo AL,Raguz J,Desideri E,Tarkowski B,Parrini M,Fischer I,Zatloukal K,Baccarini M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Wed Dec 21 00:00:00 UTC 2016
Self-renewing diploid Axin2(+) cells fuel homeostatic renewal of the liver.
Nature
Wang B,Zhao L,Fish M,Logan CY,Nusse R
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Thu Aug 13 00:00:00 UTC 2015
Host microbiota constantly control maturation and function of microglia in the CNS.
Nature neuroscience
Erny D,Hrabě de Angelis AL,Jaitin D,Wieghofer P,Staszewski O,David E,Keren-Shaul H,Mahlakoiv T,Jakobshagen K,Buch T,Schwierzeck V,Utermöhlen O,Chun E,Garrett WS,McCoy KD,Diefenbach A,Staeheli P,Stecher B,Amit I,Prinz M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Wed Jul 01 00:00:00 UTC 2015
Rapid depot-specific activation of adipocyte precursor cells at the onset of obesity.
Nature cell biology
Jeffery E,Church CD,Holtrup B,Colman L,Rodeheffer MS
25-0311 was used in Flow cytometry/Cell sorting to study how adipocyte precursor cells are activated depot-specifically at the onset of obesity.
Wed Apr 01 00:00:00 UTC 2015
Human definitive haemogenic endothelium and arterial vascular endothelium represent distinct lineages.
Nature cell biology
Ditadi A,Sturgeon CM,Tober J,Awong G,Kennedy M,Yzaguirre AD,Azzola L,Ng ES,Stanley EG,French DL,Cheng X,Gadue P,Speck NA,Elefanty AG,Keller G
25-0311 was used in Flow cytometry/Cell sorting to investigate whether haemogenic endothelium represents a subpopulation of arterial vascular endothelium, showing that they represent distinct lineages.
Fri May 01 00:00:00 UTC 2015
Nonvenous origin of dermal lymphatic vasculature.
Circulation research
Martinez-Corral I,Ulvmar MH,Stanczuk L,Tatin F,Kizhatil K,John SW,Alitalo K,Ortega S,Makinen T
25-0311 was used in Flow cytometry/Cell sorting to investigate the origin of murine lymphatic vasculature, showing a significant contribution from nonvenous cells.
Fri May 08 00:00:00 UTC 2015
Myogenic-specific ablation of Fgfr1 impairs FGF2-mediated proliferation of satellite cells at the myofiber niche but does not abolish the capacity for muscle regeneration.
Frontiers in aging neuroscience
Yablonka-Reuveni Z,Danoviz ME,Phelps M,Stuelsatz P
25-0311 was used in Flow cytometry/Cell sorting to investigate FGFR gene expression in quiescent SCs and their proliferating progeny, and its role in myogenesis.
Mon Jun 15 00:00:00 UTC 2015
Vascular endothelial growth factor c/vascular endothelial growth factor receptor 3 signaling regulates chemokine gradients and lymphocyte migration from tissues to lymphatics.
Transplantation
Iwami D,Brinkman CC,Bromberg JS
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Wed Apr 01 00:00:00 UTC 2015
Conditional immortalization of primary adipocyte precursor cells.
Adipocyte
Church C,Brown M,Rodeheffer MS
25-0311 was used in Flow cytometry/Cell sorting to assess the adipogenic capability of committed, SV40-immortalised adipocyte precursor cells.
Mon Aug 10 00:00:00 UTC 2015
Targeted Gene Correction in Osteopetrotic-Induced Pluripotent Stem Cells for the Generation of Functional Osteoclasts.
Stem cell reports
Neri T,Muggeo S,Paulis M,Caldana ME,Crisafulli L,Strina D,Focarelli ML,Faggioli F,Recordati C,Scaramuzza S,Scanziani E,Mantero S,Buracchi C,Sobacchi C,Lombardo A,Naldini L,Vezzoni P,Villa A,Ficara F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Tue Oct 13 00:00:00 UTC 2015
Distinct in vitro properties of embryonic and extraembryonic fibroblast-like cells are reflected in their in vivo behavior following grafting in the adult mouse brain.
Cell transplantation
Costa R,Bergwerf I,Santermans E,De Vocht N,Praet J,Daans J,Le Blon D,Hoornaert C,Reekmans K,Hens N,Goossens H,Berneman Z,Parolini O,Alviano F,Ponsaerts P
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Mon Nov 16 00:00:00 UTC 2015
Rejuvenation of the muscle stem cell population restores strength to injured aged muscles.
Nature medicine
Cosgrove BD,Gilbert PM,Porpiglia E,Mourkioti F,Lee SP,Corbel SY,Llewellyn ME,Delp SL,Blau HM
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Sat Mar 01 00:00:00 UTC 2014
Mesenchymal-endothelial transition contributes to cardiac neovascularization.
Nature
Ubil E,Duan J,Pillai IC,Rosa-Garrido M,Wu Y,Bargiacchi F,Lu Y,Stanbouly S,Huang J,Rojas M,Vondriska TM,Stefani E,Deb A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Thu Oct 30 00:00:00 UTC 2014
c-kit+ cells minimally contribute cardiomyocytes to the heart.
Nature
van Berlo JH,Kanisicak O,Maillet M,Vagnozzi RJ,Karch J,Lin SC,Middleton RC,Marbán E,Molkentin JD
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Thu May 15 00:00:00 UTC 2014
Transcriptional programs of lymphoid tissue capillary and high endothelium reveal control mechanisms for lymphocyte homing.
Nature immunology
Lee M,Kiefel H,LaJevic MD,Macauley MS,Kawashima H,O'Hara E,Pan J,Paulson JC,Butcher EC
25-0311 was used in Flow cytometry/Cell sorting to investigate the molecular signatures that distinguish HEVs from capillary endothelium, revealing control mechanisms for lymphocyte homing.
Wed Oct 01 00:00:00 UTC 2014
Developmental heterogeneity of cardiac fibroblasts does not predict pathological proliferation and activation.
Circulation research
Ali SR,Ranjbarvaziri S,Talkhabi M,Zhao P,Subat A,Hojjat A,Kamran P,Müller AM,Volz KS,Tang Z,Red-Horse K,Ardehali R
25-0311 was used in Flow cytometry/Cell sorting to study the developmental origins of cardiac fibroblasts and characterise their corresponding phenotypes, to aid in determining the proliferation rates of each developmental subset.
Fri Sep 12 00:00:00 UTC 2014
Cell-autonomous heparanase modulates self-renewal and migration in bone marrow-derived mesenchymal stem cells.
Journal of biomedical science
Cheng CC,Lee YH,Lin SP,Huangfu WC,Liu IH
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Thu Mar 13 00:00:00 UTC 2014
Laser microdissection of the alveolar duct enables single-cell genomic analysis.
Frontiers in oncology
Bennett RD,Ysasi AB,Belle JM,Wagner WL,Konerding MA,Blainey PC,Pyne S,Mentzer SJ
25-0311 was used in Flow cytometry/Cell sorting to measure the expression of selected markers in individual cells to characterise cell populations which contribute to the development of bronchioalveolar carcinoma.
Mon Oct 13 00:00:00 UTC 2014
Ancestral Myf5 gene activity in periocular connective tissue identifies a subset of fibro/adipogenic progenitors but does not connote a myogenic origin.
Developmental biology
Stuelsatz P,Shearer A,Yablonka-Reuveni Z
25-0311 was used in Flow cytometry/Cell sorting to identify and characterise the differences between myogenic progenitors (satellite cells) in extraocular muscles to those of the limb and diaphragm of adult mice.
Wed Jan 15 00:00:00 UTC 2014
Characterization of Cre recombinase activity for in vivo targeting of adipocyte precursor cells.
Stem cell reports
Krueger KC,Costa MJ,Du H,Feldman BJ
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Tue Dec 09 00:00:00 UTC 2014
Dominant lethal pathologies in male mice engineered to contain an X-linked DUX4 transgene.
Cell reports
Dandapat A,Bosnakovski D,Hartweck LM,Arpke RW,Baltgalvis KA,Vang D,Baik J,Darabi R,Perlingeiro RC,Hamra FK,Gupta K,Lowe DA,Kyba M
25-0311 was used in Flow cytometry/Cell sorting to demonstrate a murine model with potential use for testing anti-DUX4 therapies in facioscapulohumeral muscular dystrophy.
Thu Sep 11 00:00:00 UTC 2014
Adipocytes arise from multiple lineages that are heterogeneously and dynamically distributed.
Nature communications
Sanchez-Gurmaches J,Guertin DA
25-0311 was used in Flow cytometry/Cell sorting to establish a framework for adipose tissue development to explain body fat patterning variations in lipodystrophic or obese humans.
Thu Jun 19 00:00:00 UTC 2014
Distinct fibroblast lineages determine dermal architecture in skin development and repair.
Nature
Driskell RR,Lichtenberger BM,Hoste E,Kretzschmar K,Simons BD,Charalambous M,Ferron SR,Herault Y,Pavlovic G,Ferguson-Smith AC,Watt FM
25-0311 was used in Immunohistochemistry to explain why wounding is linked to formation of ECM-rich scar tissue that lacks hair follicles.
Thu Dec 12 00:00:00 UTC 2013
miR-126 and miR-126* repress recruitment of mesenchymal stem cells and inflammatory monocytes to inhibit breast cancer metastasis.
Nature cell biology
Zhang Y,Yang P,Sun T,Li D,Xu X,Rui Y,Li C,Chong M,Ibrahim T,Mercatali L,Amadori D,Lu X,Xie D,Li QJ,Wang XF
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Fri Mar 01 00:00:00 UTC 2013
The expression of Sox17 identifies and regulates haemogenic endothelium.
Nature cell biology
Clarke RL,Yzaguirre AD,Yashiro-Ohtani Y,Bondue A,Blanpain C,Pear WS,Speck NA,Keller G
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Wed May 01 00:00:00 UTC 2013
Notch2 genetic fate mapping reveals two previously unrecognized mammary epithelial lineages.
Nature cell biology
Šale S,Lafkas D,Artavanis-Tsakonas S
25-0311 was used in Flow cytometry/Cell sorting to study the fate of cells expressing the Notch2 receptor paralogue, uncovering two novel mammary epithelial cell linages.
Wed May 01 00:00:00 UTC 2013
Evidence for a multipotent mammary progenitor with pregnancy-specific activity.
Breast cancer research : BCR
Kaanta AS,Virtanen C,Selfors LM,Brugge JS,Neel BG
25-0311 was used in Flow cytometry/Cell sorting to provide data to show the existence of multi-potent mammary progenitor cells that have pregnancy-specific function.
Mon Dec 08 00:00:00 UTC 2014
Determination of mesenchymal stem cell fate by pigment epithelium-derived factor (PEDF) results in increased adiposity and reduced bone mineral content.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Gattu AK,Swenson ES,Iwakiri Y,Samuel VT,Troiano N,Berry R,Church CD,Rodeheffer MS,Carpenter TO,Chung C
25-0311 was used in Flow cytometry/Cell sorting to assess the contribution of pigment epithelium-derived factor to mesenchymal stem cell fate.
Fri Nov 01 00:00:00 UTC 2013
Etv2 rescues Flk1 mutant embryoid bodies.
Genesis (New York, N.Y. : 2000)
Rasmussen TL,Martin CM,Walter CA,Shi X,Perlingeiro R,Koyano-Nakagawa N,Garry DJ
25-0311 was used in Flow cytometry/Cell sorting to show that embryonic Flk1 mutants can be rescued by Etv2.
Mon Jul 01 00:00:00 UTC 2013
Mouse models and techniques for the isolation of the diabetic endothelium.
ISRN endocrinology
Darrow AL,Maresh JG,Shohet RV
25-0311 was used in Flow cytometry/Cell sorting to validate mouse models and endothelial isolation techniques for assessing diabetic endothelial dysfunction and vascular responses in vivo.
Wed Jul 10 00:00:00 UTC 2013
Endothelial cells provide an instructive niche for the differentiation and functional polarization of M2-like macrophages.
Blood
He H,Xu J,Warren CM,Duan D,Li X,Wu L,Iruela-Arispe ML
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Thu Oct 11 00:00:00 UTC 2012
Etv2 is expressed in the yolk sac hematopoietic and endothelial progenitors and regulates Lmo2 gene expression.
Stem cells (Dayton, Ohio)
Koyano-Nakagawa N,Kweon J,Iacovino M,Shi X,Rasmussen TL,Borges L,Zirbes KM,Li T,Perlingeiro RC,Kyba M,Garry DJ
Wed Aug 01 00:00:00 UTC 2012
ALDH1A isozymes are markers of human melanoma stem cells and potential therapeutic targets.
Stem cells (Dayton, Ohio)
Luo Y,Dallaglio K,Chen Y,Robinson WA,Robinson SE,McCarter MD,Wang J,Gonzalez R,Thompson DC,Norris DA,Roop DR,Vasiliou V,Fujita M
25-0311 was used in Flow cytometry/Cell sorting to demonstrate the existence of melanoma cells that fulfil the criteria for CSCs.
Mon Oct 01 00:00:00 UTC 2012
ER71 directs mesodermal fate decisions during embryogenesis.
Development (Cambridge, England)
Rasmussen TL,Kweon J,Diekmann MA,Belema-Bedada F,Song Q,Bowlin K,Shi X,Ferdous A,Li T,Kyba M,Metzger JM,Koyano-Nakagawa N,Garry DJ
25-0311 was used in Flow cytometry/Cell sorting to further define the functional role for ER71 in cell lineage decisions using genetically modified mouse models.
Tue Nov 01 00:00:00 UTC 2011
Intramuscular VEGF activates an SDF1-dependent progenitor cell cascade and an SDF1-independent muscle paracrine cascade for cardiac repair.
American journal of physiology. Heart and circulatory physiology
Zisa D,Shabbir A,Mastri M,Taylor T,Aleksic I,McDaniel M,Suzuki G,Lee T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Thu Dec 01 00:00:00 UTC 2011
Vegf regulates embryonic erythroid development through Gata1 modulation.
Blood
Drogat B,Kalucka J,Gutiérrez L,Hammad H,Goossens S,Farhang Ghahremani M,Bartunkova S,Haigh K,Deswarte K,Nyabi O,Naessens M,Ferrara N,Klingmüller U,Lambrecht BN,Nagy A,Philipsen S,Haigh JJ
25-0311 was used in Flow cytometry/Cell sorting to demonstrate that Vegf modulates Gata1 expression levels in vivo and provide new molecular insight into Vegf's ability to modulate erythropoiesis.
Thu Sep 23 00:00:00 UTC 2010
Auf1/Hnrnpd-deficient mice develop pruritic inflammatory skin disease.
The Journal of investigative dermatology
Sadri N,Schneider RJ
Sun Mar 01 00:00:00 UTC 2009
Heart failure therapy mediated by the trophic activities of bone marrow mesenchymal stem cells: a noninvasive therapeutic regimen.
American journal of physiology. Heart and circulatory physiology
Shabbir A,Zisa D,Suzuki G,Lee T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Mon Jun 01 00:00:00 UTC 2009
Intramuscular VEGF repairs the failing heart: role of host-derived growth factors and mobilization of progenitor cells.
American journal of physiology. Regulatory, integrative and comparative physiology
Zisa D,Shabbir A,Mastri M,Suzuki G,Lee T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0311-82) in Flow Cytometry
Sun Nov 01 00:00:00 UTC 2009