Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • E-cadherin Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.

          Invitrogen

          CD324 (E-Cadherin) Monoclonal Antibody (DECMA-1), Biotin, eBioscience™

          42 Published Figures
          49 References
          View all (152) E-cadherin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Datasheet
          Protocols

          Cite CD324 (E-Cadherin) Monoclonal Antibody (DECMA-1), Biotin, eBioscience™

          • Antibody Testing Data (1)
          • Published Figures (42)
          CD324 (E-Cadherin) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD324 (E-Cadherin) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 43

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD324 (E-Cadherin) Antibody (13-3249-80) in Flow

          Staining of the MDCK cell line with 0.125 µg of Rat IgG1 kappa Isotype Control Biotin (Product # 13-4301-82) (blue histogram) or 0.125 µg of Anti-Human CD324 (E-Cadherin) Biotin (purple histogram) followed by Streptavidin PE (Product # 12-4317-87). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD324 (E-Cadherin) Antibody in Flow Cytometry (Flow)
          CD324 (E-Cadherin) Antibody in Western Blot (WB)
          CD324 (E-Cadherin) Antibody in Western Blot (WB)
          CD324 (E-Cadherin) Antibody in Western Blot (WB)
          CD324 (E-Cadherin) Antibody in Western Blot (WB)
          CD324 (E-Cadherin) Antibody in Neutralization (Neu)
          CD324 (E-Cadherin) Antibody in Neutralization (Neu)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry, Immunohistochemistry (ICC/IF, IHC)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (IHC)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (IHC)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (IHC)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (IHC)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (IHC)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (IHC)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (IHC)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (IHC)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (IHC)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (IHC)
          CD324 (E-Cadherin) Antibody in Flow Cytometry (Flow)
          CD324 (E-Cadherin) Antibody in Flow Cytometry (Flow)
          CD324 (E-Cadherin) Antibody in Flow Cytometry (Flow)
          CD324 (E-Cadherin) Antibody in Flow Cytometry (Flow)
          CD324 (E-Cadherin) Antibody in Flow Cytometry (Flow)
          CD324 (E-Cadherin) Antibody in Flow Cytometry (Flow)
          CD324 (E-Cadherin) Antibody in Flow Cytometry (Flow)
          CD324 (E-Cadherin) Antibody in Flow Cytometry (Flow)
          CD324 (E-Cadherin) Antibody in Flow Cytometry (Flow)
          CD324 (E-Cadherin) Antibody in Neutralization (Neu)
          CD324 (E-Cadherin) Antibody in Neutralization (Neu)

          Product Details

          13-3249-80

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-Dependent
          View 5 publications 5 publications

          Immunohistochemistry (IHC)

          -
          View 7 publications 7 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-Dependent
          -

          Immunocytochemistry (ICC/IF)

          -
          View 12 publications 12 publications

          Flow Cytometry (Flow)

          0.25 µg/test
          View 20 publications 20 publications

          Neutralization (Neu)

          -
          View 2 publications 2 publications

          Miscellaneous PubMed (MISC)

          -
          View 1 publication 1 publication
          Product Specifications

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rat / IgG1, kappa

          Recommended Isotype Control

          Rat IgG1 kappa Isotype Control (eBRG1), Biotin, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          DECMA-1

          Conjugate

          Biotin Biotin Biotin
          • Unconjugated
          • Alexa Fluor 488 
          • eFluor 660
          • Functional Grade
          • PerCP-eFluor 710
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Wet ice

          RRID

          AB_1659689

          Product Specific Information

          Description: The monoclonal antibody DECMA-1 recognizes mouse, human and canine CD324 also known as E-cadherin (Epithelial cadherin) or uvomorulin. Like the other cadherin family members P and N cadherin, E-cadherin is a transmembrane glycoprotein involved in intercellular adhesion. These proteins share a common basic structure. The extracellular portions of the proteins are largely composed of repeating domains, each with two consensus Ca2+-binding motifs. The cytoplasmic domain interacts with a-, b-, and g-catenins and actinins. These catenins connect E-cadherin with the cytoskeleton.

          Expression is found in most epidermal cells including melanocytes and kerotinocytes. E-cadherin is localized at the intercellular boundaries of epithelial cells in several tissues, and is thought to play a role in maintenance of tissue integrity. Loss of E-cadherin function has been implicated in the progression of a variety of cancers.

          E-Cadherin protein is sensitive to trypsin treatment, so exposure to trypsin should be minimized or avoided.

          The monoclonal antibody DECMA-1 has been shown to have functional activity by disrupting adhesion in human, mouse and dog cells.

          Applications Reported: This DECMA-1 antibody has been reported for use in flow cytometric analysis, western blotting, and immunohistochemical staining of frozen tissue sections.

          Applications Tested: This DECMA-1 antibody has been tested by flow cytometric analysis of the MDCK cell line. Optimal staining is achieved by intracellular staining as protein turnover can result in variable surface staining. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          E-Cadherin (epithelial cadherin) is a classical cadherin from the cadherin (alcium dependent adhesion protein) superfamily. The encoded protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Mutations in this gene are correlated with gastric, breast, colorectal, thyroid and ovarian cancer. Loss of function is thought to contribute to progression in cancer by increasing proliferation, invasion, and/or metastasis. The ectodomain of this protein mediates bacterial adhesion to mammalian cells and the cytoplasmic domain is required for internalization. Identified transcript variants arise from mutation at consensus splice sites. E-cadherin plays a central role in the growth and development of cells by controlling tissue architecture, and maintenance of tissue integrity. In humans, E-cadherin is encoded by the CDH1 gene present on chromosome 16. Studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0