Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CD42d Antibodies

          Invitrogen

          CD42d Monoclonal Antibody (1C2), PE, eBioscience™

          2 Published Figures
          7 References
          View all (13) CD42d antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CD42d Monoclonal Antibody (1C2), PE, eBioscience™

          • Antibody Testing Data (1)
          • Published Figures (2)
          CD42d Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD42d Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD42d Antibody (12-0421-82) in Flow

          Staining of mouse platelets with Anti-Mouse CD41 FITC (Product # 11-0411-82) and 0.5 µg of Armenian Hamster IgG Isotype Control PE (Product # 12-4888-81) (left) or 0.5 µg of Anti-Mouse/Rat CD42d PE (right). Total events were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD42d Antibody in Flow Cytometry (Flow)
          CD42d Antibody in Flow Cytometry (Flow)
          CD42d Antibody in Flow Cytometry (Flow)

          Product Details

          12-0421-82

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          1 µg/test
          View 7 publications 7 publications
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Armenian hamster / IgG

          Recommended Isotype Control

          Armenian Hamster IgG Isotype Control (eBio299Arm), PE, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1C2

          Conjugate

          PE PE PE
          • Unconjugated
          • Alexa Fluor 488 
          • APC 
          • PerCP-eFluor 710
          • Request custom conjugation

          Excitation/Emission Max

          565/576 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10804771

          Product Specific Information

          Description: The monoclonal antibody 1C2 recognizes mouse and rat CD42d, a single transmembrane protein also known as platelet glycoprotein V (GPV). CD42d interacts with CD42a, CD42b and CD42c to form the receptor for von willebrand factor and thrombin. This functional complex plays a role in platelet adhesion. Expression of CD42d (unlike other markers such as CD41) is restricted to resting platelets and megakaryocytes and therefore is a good marker for development of megakaryocytes.

          Applications Reported: This 1C2 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This 1C2 antibody has been tested by flow cytometric analysis of mouse platelets. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: CD42d; Glycoprotein 5; glycoprotein 5, platelet; GPV; Platelet glycoprotein 5; Platelet glycoprotein V; Platelete glycoprotein 5; Platelets GP V

          View more View less

          Gene Aliases: Gp5; GPV; PLGPV

          View more View less

          UniProt ID: (Rat) O08770

          View more View less

          Entrez Gene ID: (Rat) 25259, (Mouse) 14729

          View more View less

          Function(s)
          collagen binding
          Process(es)
          cell adhesion cell-matrix adhesion blood coagulation negative regulation of platelet activation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-67f4cd694f-jv9gd:80/100.66.135.11:80.
          git-commit: 961b7666e63bcc533d6299fdd217d6a70bd96cf1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0