Product References
Pyrimidine de novo synthesis inhibition selectively blocks effector but not memory T cell development.
Nature immunology
Scherer S,Oberle SG,Kanev K,Gerullis AK,Wu M,de Almeida GP,Puleston DJ,Baixauli F,Aly L,Greco A,Nizharadze T,Becker NB,Hoesslin MV,Donhauser LV,Berner J,Chu T,McNamara HA,Esencan Z,Roelli P,Wurmser C,Kleiter I,Vehreschild MJGT,Mayer CA,Knolle P,Klingenspor M,Fumagalli V,Iannacone M,Prlic M,Korn T,Pearce EL,Höfer T,Schulz AM,Zehn D
25-0454-82 was used in Flow cytometry/Cell sorting to show memory T cell precursors are resistant to pyrimidine starvation.
Wed Mar 01 00:00:00 UTC 2023
Oxidative pentose phosphate pathway controls vascular mural cell coverage by regulating extracellular matrix composition.
Nature metabolism
Facchinello N,Astone M,Audano M,Oberkersch RE,Spizzotin M,Calura E,Marques M,Crisan M,Mitro N,Santoro MM
25-0454-82 was used in Flow cytometry/Cell sorting to show that the oxidative pentose phosphate pathway (oxPPP) in endothelial cells is required for establishing vMC coverage of the dorsal aorta during early vertebrate development in zebrafish and mice.
Sat Jan 01 00:00:00 UTC 2022
Smc3 dosage regulates B cell transit through germinal centers and restricts their malignant transformation.
Nature immunology
Rivas MA,Meydan C,Chin CR,Challman MF,Kim D,Bhinder B,Kloetgen A,Viny AD,Teater MR,McNally DR,Doane AS,Béguelin W,Fernández MTC,Shen H,Wang X,Levine RL,Chen Z,Tsirigos A,Elemento O,Mason CE,Melnick AM
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Mon Feb 01 00:00:00 UTC 2021
Niche-specific MHC II and PD-L1 regulate CD4+CD8αα+ intraepithelial lymphocyte differentiation.
The Journal of experimental medicine
Moon S,Park Y,Hyeon S,Kim YM,Kim JH,Kim H,Park S,Lee KJ,Koo BK,Ha SJ,Lee SW
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Mon Apr 05 00:00:00 UTC 2021
Brd4 modulates diet-induced obesity via PPARγ-dependent Gdf3 expression in adipose tissue macrophages.
JCI insight
Hu X,Dong X,Li G,Chen Y,Chen J,He X,Sun H,Kim DH,Kemper JK,Chen LF
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Thu Apr 08 00:00:00 UTC 2021
Differentiation of fetal hematopoietic stem cells requires ARID4B to restrict autocrine KITLG/KIT-Src signaling.
Cell reports
Young IC,Wu B,Andricovich J,Chuang ST,Li R,Tzatsos A,Wu RC,Wu MY
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Tue Nov 23 00:00:00 UTC 2021
Expansion of an Unusual Virtual Memory CD8+ Subpopulation Bearing Vα3.2 TCR in Themis-Deficient Mice.
Frontiers in immunology
Prasad M,Wojciech L,Brzostek J,Hu J,Chua YL,Tung DWH,Yap J,Rybakin V,Gascoigne NRJ
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Thu Sep 23 00:00:00 UTC 2021
The subunits of IL-12, originating from two distinct cells, can functionally synergize to protect against pathogen dissemination in vivo.
Cell reports
Gerber AN,Abdi K,Singh NJ
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Tue Oct 12 00:00:00 UTC 2021
Erythritol Ameliorates Small Intestinal Inflammation Induced by High-Fat Diets and Improves Glucose Tolerance.
International journal of molecular sciences
Kawano R,Okamura T,Hashimoto Y,Majima S,Senmaru T,Ushigome E,Asano M,Yamazaki M,Takakuwa H,Sasano R,Nakanishi N,Hamaguchi M,Fukui M
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Mon May 24 00:00:00 UTC 2021
Ddb1 Is Essential for the Expansion of CD4+ Helper T Cells by Regulating Cell Cycle Progression and Cell Death.
Frontiers in immunology
Yang L,Chen W,Li L,Xiao Y,Fan S,Zhang Q,Xia T,Li M,Hong Y,Zhao T,Li Q,Liu WH,Xiao N
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Mon Dec 20 00:00:00 UTC 2021
Abraxane-induced bone marrow CD11b+ myeloid cell depletion in tumor-bearing mice is visualized by μPET-CT with 64Cu-labeled anti-CD11b and prevented by anti-CSF-1.
Theranostics
Cao Q,Huang Q,Wang YA,Li C
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Fri Jul 23 00:00:00 UTC 2021
Visceral adipose tissue imparts peripheral macrophage influx into the hypothalamus.
Journal of neuroinflammation
Chen KE,Lainez NM,Nair MG,Coss D
25-0454-82 was used in Flow Cytometry to demonstrate a new connection between the adipose tissue and the hypothalamus in obesity that contributes to neuroinflammation and hypothalamic pathologies.
Mon Jun 21 00:00:00 UTC 2021
Heterogenous Populations of Tissue-Resident CD8+ T Cells Are Generated in Response to Infection and Malignancy.
Immunity
Milner JJ,Toma C,He Z,Kurd NS,Nguyen QP,McDonald B,Quezada L,Widjaja CE,Witherden DA,Crowl JT,Shaw LA,Yeo GW,Chang JT,Omilusik KD,Goldrath AW
25-0454 was used in Flow cytometry/Cell sorting to identify discrete lineages of intestinal antigen-specific CD8+ T cells, including a Blimp1hiId3lo tissue-resident effector cell population most prominent in the early phase of acute viral and bacterial infections and a molecularly distinct Blimp1loId3hi tissue-resident memory population that subsequently accumulated at later infection time points.
Tue May 19 00:00:00 UTC 2020
Mutant EZH2 Induces a Pre-malignant Lymphoma Niche by Reprogramming the Immune Response.
Cancer cell
Béguelin W,Teater M,Meydan C,Hoehn KB,Phillip JM,Soshnev AA,Venturutti L,Rivas MA,Calvo-Fernández MT,Gutierrez J,Camarillo JM,Takata K,Tarte K,Kelleher NL,Steidl C,Mason CE,Elemento O,Allis CD,Kleinstein SH,Melnick AM
25-0454 was used in Flow cytometry/Cell sorting to show that EZH2 mutations initiate FL by attenuating germinal center(GC) B cell requirement for T cell help and driving slow expansion of GC centrocytes that become enmeshed with and dependent on follicular dendritic cells.
Mon May 11 00:00:00 UTC 2020
Deregulated Notch and Wnt signaling activates early-stage myeloid regeneration pathways in leukemia.
The Journal of experimental medicine
Kang YA,Pietras EM,Passegué E
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Mon Mar 02 00:00:00 UTC 2020
Proenkephalin+ regulatory T cells expanded by ultraviolet B exposure maintain skin homeostasis with a healing function.
Proceedings of the National Academy of Sciences of the United States of America
Shime H,Odanaka M,Tsuiji M,Matoba T,Imai M,Yasumizu Y,Uraki R,Minohara K,Watanabe M,Bonito AJ,Fukuyama H,Ohkura N,Sakaguchi S,Morita A,Yamazaki S
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Tue Aug 25 00:00:00 UTC 2020
Interleukin 35 Delays Hindlimb Ischemia-Induced Angiogenesis Through Regulating ROS-Extracellular Matrix but Spares Later Regenerative Angiogenesis.
Frontiers in immunology
Fu H,Sun Y,Shao Y,Saredy J,Cueto R,Liu L,Drummer C,Johnson C,Xu K,Lu Y,Li X,Meng S,Xue ER,Tan J,Jhala NC,Yu D,Zhou Y,Bayless KJ,Yu J,Rogers TJ,Hu W,Snyder NW,Sun J,Qin X,Jiang X,Wang H,Yang X
25-0454 was used in Flow cytometry/Cell sorting to study the role and function of IL-35 in ischemia-induced angiogenesis in peripheral artery diseases.
Fri Jul 09 00:00:00 UTC 2021
Interleukin 35 Delays Hindlimb Ischemia-Induced Angiogenesis Through Regulating ROS-Extracellular Matrix but Spares Later Regenerative Angiogenesis.
Frontiers in immunology
Fu H,Sun Y,Shao Y,Saredy J,Cueto R,Liu L,Drummer C,Johnson C,Xu K,Lu Y,Li X,Meng S,Xue ER,Tan J,Jhala NC,Yu D,Zhou Y,Bayless KJ,Yu J,Rogers TJ,Hu W,Snyder NW,Sun J,Qin X,Jiang X,Wang H,Yang X
25-0454 was used in Flow cytometry/Cell sorting to study the role and function of IL-35 in ischemia-induced angiogenesis in peripheral artery diseases.
Fri Jul 09 00:00:00 UTC 2021
CD103+ cDC1 and endogenous CD8+ T cells are necessary for improved CD40L-overexpressing CAR T cell antitumor function.
Nature communications
Kuhn NF,Lopez AV,Li X,Cai W,Daniyan AF,Brentjens RJ
25-0454 was used in Flow cytometry/Cell sorting to demonstrate the stimulatory effect of CD40L-overexpressing CAR T cells on innate and adaptive immune cells, and provide a rationale for using CD40L-overexpressing CAR T cells to improve immunotherapy responses.
Wed Dec 02 00:00:00 UTC 2020
IFN-γ promoted exosomes from mesenchymal stem cells to attenuate colitis via miR-125a and miR-125b.
Cell death & disease
Yang R,Huang H,Cui S,Zhou Y,Zhang T,Zhou Y
25-0454-82 was used in Flow Cytometry to investigate the potential anti-inflammatory effects of bone marrow mesenchymal stem cell-derived exosomes.
Thu Jul 30 00:00:00 UTC 2020
MicroRNAs of the miR-17~9 family maintain adipose tissue macrophage homeostasis by sustaining IL-10 expression.
eLife
Zhang X,Liu J,Wu L,Hu X
25-0454 was used in Flow cytometry/Cell sorting to identifiy miR-17~92 family miRNAs as crucial regulators of the balance between pro- and anti-inflammatory cytokines and exemplify how macrophage-intrinsic regulatory circuit exerted impactful influence on general physiology.
Thu Nov 05 00:00:00 UTC 2020
The frequency of follicular T helper cells differs in acute and chronic neuroinflammation.
Scientific reports
Baniahmad A,Birkner K,Görg J,Loos J,Zipp F,Wasser B,Bittner S
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Tue Nov 24 00:00:00 UTC 2020
Long-term ex vivo haematopoietic-stem-cell expansion allows nonconditioned transplantation.
Nature
Wilkinson AC,Ishida R,Kikuchi M,Sudo K,Morita M,Crisostomo RV,Yamamoto R,Loh KM,Nakamura Y,Watanabe M,Nakauchi H,Yamazaki S
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Mon Jul 01 00:00:00 UTC 2019
Hes1 attenuates type I IFN responses via VEGF-C and WDFY1.
The Journal of experimental medicine
Ning F,Li X,Yu L,Zhang B,Zhao Y,Liu Y,Zhao B,Shang Y,Hu X
25-0454 was used in Flow cytometry/Cell sorting to identify Hes1 as a homeostatic negative regulator of type I IFNs for the maintenance of immune balance in the context of antiviral immunity and autoimmune diseases.
Mon Jun 03 00:00:00 UTC 2019
Synergized regulation of NK cell education by NKG2A and specific Ly49 family members.
Nature communications
Zhang X,Feng J,Chen S,Yang H,Dong Z
25-0454 was used in Flow cytometry/Cell sorting to provide genetic evidence supporting that NKG2A and the inhibitory members of Ly49 family receptors synergize to regulate NK cell education.
Fri Nov 01 00:00:00 UTC 2019
Loss of NF-κB1 Causes Gastric Cancer with Aberrant Inflammation and Expression of Immune Checkpoint Regulators in a STAT-1-Dependent Manner.
Immunity
O'Reilly LA,Putoczki TL,Mielke LA,Low JT,Lin A,Preaudet A,Herold MJ,Yaprianto K,Tai L,Kueh A,Pacini G,Ferrero RL,Gugasyan R,Hu Y,Christie M,Wilcox S,Grumont R,Griffin MDW,O'Connor L,Smyth GK,Ernst M,Waring P,Gerondakis S,Strasser A
25-0454-82 was used in Flow Cytometry to provide mechanistic insight into how polymorphisms that attenuate NFKB1 expression predispose humans to epithelial cancers, highlighting the pro-tumorigenic activity of STAT1 and identifying targetable vulnerabilities in gastric cancer.
Tue Mar 20 00:00:00 UTC 2018
Differential requirement of kindlin-3 for T cell progenitor homing to the non-vascularized and vascularized thymus.
eLife
Moretti FA,Klapproth S,Ruppert R,Margraf A,Weber J,Pick R,Scheiermann C,Sperandio M,Fässler R,Moser M
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Thu Sep 06 00:00:00 UTC 2018
The H3K4 methyltransferase Setd1b is essential for hematopoietic stem and progenitor cell homeostasis in mice.
eLife
Schmidt K,Zhang Q,Tasdogan A,Petzold A,Dahl A,Arneth BM,Slany R,Fehling HJ,Kranz A,Stewart AF,Anastassiadis K
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Tue Jun 19 00:00:00 UTC 2018
Heterogeneous Responses of Hematopoietic Stem Cells to Inflammatory Stimuli Are Altered with Age.
Cell reports
Mann M,Mehta A,de Boer CG,Kowalczyk MS,Lee K,Haldeman P,Rogel N,Knecht AR,Farouq D,Regev A,Baltimore D
25-0454-82 was used in Flow Cytometry to identify and isolate an LT-HSC subset that regulates myeloid versus lymphoid balance under inflammatory challenge and with age.
Tue Dec 11 00:00:00 UTC 2018
Tet1 and Tet2 maintain mesenchymal stem cell homeostasis via demethylation of the P2rX7 promoter.
Nature communications
Yang R,Yu T,Kou X,Gao X,Chen C,Liu D,Zhou Y,Shi S
25-0454 was used in Flow cytometry/Cell sorting to indicate that Tet1 and Tet2 play a role in maintaining mesenchymal stem cell and bone homeostasis through demethylation of P2rX7.
Fri Jun 01 00:00:00 UTC 2018
Egr2 and 3 control adaptive immune responses by temporally uncoupling expansion from T cell differentiation.
The Journal of experimental medicine
Miao T,Symonds ALJ,Singh R,Symonds JD,Ogbe A,Omodho B,Zhu B,Li S,Wang P
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Mon Jun 05 00:00:00 UTC 2017
Bacillus anthracis lethal toxin negatively modulates ILC3 function through perturbation of IL-23-mediated MAPK signaling.
PLoS pathogens
Seshadri S,Allan DSJ,Carlyle JR,Zenewicz LA
25-0454 was used in Flow cytometry/Cell sorting to study the effects of MAPK signalling pathways in group 3 innate immune lymphocytes using Bacillus anthracis.
Sun Oct 01 00:00:00 UTC 2017
Pneumocystis-Driven Inducible Bronchus-Associated Lymphoid Tissue Formation Requires Th2 and Th17 Immunity.
Cell reports
Eddens T,Elsegeiny W,Garcia-Hernadez ML,Castillo P,Trevejo-Nunez G,Serody K,Campfield BT,Khader SA,Chen K,Rangel-Moreno J,Kolls JK
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Tue Mar 28 00:00:00 UTC 2017
Recipient bone marrow assimilates the myeloid/lymphoid reconstitution of distinct fetal hematopoietic stem cells.
Oncotarget
Guo XL,Chu L,Ke F,Mu LL,Li Z,Cai JJ,Li HF,Hong DL
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Tue Dec 12 00:00:00 UTC 2017
Intratumoral modulation of the inducible co-stimulator ICOS by recombinant oncolytic virus promotes systemic anti-tumour immunity.
Nature communications
Zamarin D,Holmgaard RB,Ricca J,Plitt T,Palese P,Sharma P,Merghoub T,Wolchok JD,Allison JP
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Mon Feb 13 00:00:00 UTC 2017
Host-Protozoan Interactions Protect from Mucosal Infections through Activation of the Inflammasome.
Cell
Chudnovskiy A,Mortha A,Kana V,Kennard A,Ramirez JD,Rahman A,Remark R,Mogno I,Ng R,Gnjatic S,Amir ED,Solovyov A,Greenbaum B,Clemente J,Faith J,Belkaid Y,Grigg ME,Merad M
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Thu Oct 06 00:00:00 UTC 2016
The Deubiquitinase OTULIN Is an Essential Negative Regulator of Inflammation and Autoimmunity.
Cell
Damgaard RB,Walker JA,Marco-Casanova P,Morgan NV,Titheradge HL,Elliott PR,McHale D,Maher ER,McKenzie ANJ,Komander D
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Thu Aug 25 00:00:00 UTC 2016
Murine iPSC-Derived Macrophages as a Tool for Disease Modeling of Hereditary Pulmonary Alveolar Proteinosis due to Csf2rb Deficiency.
Stem cell reports
Mucci A,Kunkiel J,Suzuki T,Brennig S,Glage S,Kühnel MP,Ackermann M,Happle C,Kuhn A,Schambach A,Trapnell BC,Hansen G,Moritz T,Lachmann N
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Tue Aug 09 00:00:00 UTC 2016
Atypical chemokine receptor 1 deficiency reduces atherogenesis in ApoE-knockout mice.
Cardiovascular research
Wan W,Liu Q,Lionakis MS,Marino AP,Anderson SA,Swamydas M,Murphy PM
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Mon Jun 01 00:00:00 UTC 2015
Axitinib increases the infiltration of immune cells and reduces the suppressive capacity of monocytic MDSCs in an intracranial mouse melanoma model.
Oncoimmunology
Du Four S,Maenhout SK,De Pierre K,Renmans D,Niclou SP,Thielemans K,Neyns B,Aerts JL
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Wed Apr 01 00:00:00 UTC 2015
WASH is required for the differentiation commitment of hematopoietic stem cells in a c-Myc-dependent manner.
The Journal of experimental medicine
Xia P,Wang S,Huang G,Zhu P,Li M,Ye B,Du Y,Fan Z
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Mon Sep 22 00:00:00 UTC 2014
Leukocyte population dynamics and detection of IL-9 as a major cytokine at the mouse fetal-maternal interface.
PloS one
Habbeddine M,Verbeke P,Karaz S,Bobé P,Kanellopoulos-Langevin C
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Wed Jun 10 00:00:00 UTC 2015
Mutual expression of the transcription factors Runx3 and ThPOK regulates intestinal CD4⁺ T cell immunity.
Nature immunology
Reis BS,Rogoz A,Costa-Pinto FA,Taniuchi I,Mucida D
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Fri Mar 01 00:00:00 UTC 2013
miR-126 and miR-126* repress recruitment of mesenchymal stem cells and inflammatory monocytes to inhibit breast cancer metastasis.
Nature cell biology
Zhang Y,Yang P,Sun T,Li D,Xu X,Rui Y,Li C,Chong M,Ibrahim T,Mercatali L,Amadori D,Lu X,Xie D,Li QJ,Wang XF
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Fri Mar 01 00:00:00 UTC 2013
Arrayed lentiviral barcoding for quantification analysis of hematopoietic dynamics.
Stem cells (Dayton, Ohio)
Grosselin J,Sii-Felice K,Payen E,Chretien S,Tronik-Le Roux D,Leboulch P
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Tue Oct 01 00:00:00 UTC 2013
The cardioprotective protein apolipoprotein A1 promotes potent anti-tumorigenic effects.
The Journal of biological chemistry
Zamanian-Daryoush M,Lindner D,Tallant TC,Wang Z,Buffa J,Klipfell E,Parker Y,Hatala D,Parsons-Wingerter P,Rayman P,Yusufishaq MSS,Fisher EA,Smith JD,Finke J,DiDonato JA,Hazen SL
Published figure using CD45.2 monoclonal antibody (Product # 25-0454-82) in Flow Cytometry
Fri Jul 19 00:00:00 UTC 2013
The transcriptional regulators Id2 and Id3 control the formation of distinct memory CD8+ T cell subsets.
Nature immunology
Yang CY,Best JA,Knell J,Yang E,Sheridan AD,Jesionek AK,Li HS,Rivera RR,Lind KC,D'Cruz LM,Watowich SS,Murre C,Goldrath AW
25-0454 was used in Flow cytometry/Cell sorting to elucidate the mechanisms of regulation of terminal differentiation of CD8(+) T cells.
Sun Nov 06 00:00:00 UTC 2011