Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CD49e (Integrin alpha 5) Antibodies

          Invitrogen

          CD49e (Integrin alpha 5) Monoclonal Antibody (eBioSAM-1 (SAM-1, SAM1)), Biotin, eBioscience™

          1 Published Figure
          2 References
          View all (36) CD49e (Integrin alpha 5) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CD49e (Integrin alpha 5) Monoclonal Antibody (eBioSAM-1 (SAM-1, SAM1)), Biotin, eBioscience™

          • Antibody Testing Data (1)
          • Published Figures (1)
          CD49e (Integrin alpha 5) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD49e (Integrin alpha 5) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD49e (Integrin alpha 5) Antibody (13-0496-80) in Flow

          Normal human peripheral blood cells were stained with CD3 Monoclonal Antibody, eFluor 450 (Product # 48-0038-42) and Mouse IgG2b kappa Isotype Control, Biotin (Product # 13-4732-85) (left) or CD49e Monoclonal Antibody, Biotin (right) followed by Streptavidin PE (Product # 12-4317-87). Cells in the lymphocyte gate were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD49e (Integrin alpha 5) Antibody in Flow Cytometry (Flow)
          CD49e (Integrin alpha 5) Antibody in Flow Cytometry (Flow)

          Product Details

          13-0496-80

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.5 µg/test
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human

          Host/Isotype

          Mouse / IgG2b

          Recommended Isotype Control

          Mouse IgG2b kappa Isotype Control (eBMG2b), Biotin, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          eBioSAM-1 (SAM-1, SAM1)

          Conjugate

          Biotin Biotin Biotin
          • Unconjugated
          • Alexa Fluor 488 
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_763503

          Product Specific Information

          Description: The eBioSAM-1 monoclonal antibody reacts with human integrin alpha 5, also known as fibronectin receptor alpha chain, very late activation antigen 5 alpha, and CD49e. Integrins are composed of an alpha chain and a beta chain, which non-covalently associate to form the functional integrin. Integrin heterodimers participate in cell surface-mediated signaling and adhesion functions. Integrin alpha 5 undergoes post-translational cleavage in its extracellular domain to yield disulfide linked light and heavy chains that join with Integrin beta 1 (CD29) to form the fibronectin receptor, also known as the very late activation antigen-5 (VLA-5) complex. Integrin alpha 5 is expressed on thymocytes, T cells, monocytes, platelets, early B cells, and activated B cells.

          Applications Reported: This eBioSAM-1 (SAM-1, SAM1) antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This eBioSAM-1 (SAM-1, SAM1) antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          The product of this gene belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes the integrin alpha 5 chain. Alpha chain 5 undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CD49 antigen-like family member E; CD49e; Fibronectin receptor subunit alpha; fibronectin receptor, alpha polypeptide; fibronectin receptor, alpha subunit; integrin alpha 5; Integrin alpha-5; Integrin alpha-F; integrin, alpha 5 (fibronectin receptor, alpha polypeptide); very late activation protein 5, alpha subunit; VLA-5

          View more View less

          Gene Aliases: CD49e; FNRA; ITGA5; VLA-5; VLA5A

          View more View less

          UniProt ID: (Human) P08648

          View more View less

          Entrez Gene ID: (Human) 3678

          View more View less

          Function(s)
          virus receptor activity epidermal growth factor receptor binding platelet-derived growth factor receptor binding integrin binding protein binding vascular endothelial growth factor receptor 2 binding metal ion binding
          Process(es)
          angiogenesis cell-substrate junction assembly cell adhesion heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules leukocyte cell-cell adhesion integrin-mediated signaling pathway memory positive regulation of cell-substrate adhesion extracellular matrix organization positive regulation of cell migration positive regulation of vascular endothelial growth factor receptor signaling pathway cell-substrate adhesion cell-cell adhesion mediated by integrin heterotypic cell-cell adhesion wound healing, spreading of epidermal cells endodermal cell differentiation viral entry into host cell positive regulation of peptidyl-tyrosine phosphorylation leukocyte migration positive regulation of sprouting angiogenesis negative regulation of anoikis
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0