Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CD51/CD61 Antibodies

          NeoBiotechnologies

          CD51/Integrin V Monoclonal Antibody (ITGAV/1610)

          View all (17) CD51/CD61 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CD51/Integrin V Monoclonal Antibody (ITGAV/1610)

          • Antibody Testing Data (1)
          CD51/Integrin V Antibody in SDS-PAGE (SDS-PAGE)
          Group 53 Created with Sketch.
          CD51/Integrin V Antibody in SDS-PAGE (SDS-PAGE)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD51/Integrin V Antibody (3685-MSM1-P1ABX) in SDS-PAGE

          SDS-PAGE analysis Purified CD51/Integrin &V Mouse Monoclonal Antibody (ITGAV/1610). Confirmation of Purity and Integrity of Antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD51/Integrin V Antibody in SDS-PAGE (SDS-PAGE)

          Product Details

          3685-MSM1-P1ABX

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-2 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          1-2 µg/mL
          -

          Flow Cytometry (Flow)

          1-2 µg/10^6 cells
          -

          ELISA (ELISA)

          1-5 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          ITGAV/1610

          Immunogen

          Recombinant human full-length integrin alpha V beta 1 protein
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A/G

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          CD51 and CD61 molecules (alpha V and beta 3 integrins) form a complex. This complex binds vitronection at the RGD sequence and can also bind fibrinogen, von Willebrand factor, thrombospondin, fibronectin, osteopontin and collagen. Reacts with osteoclasts, placenta, melanoma cell lines and weakly with platelets.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: antigen CD61; antigen identified by monoclonal antibody L230; CD51; CD61; GPIIIa; integrin alpha V; integrin alpha-5 beta-3; Integrin alpha-V; integrin alphaVbeta3; integrin beta 3; Integrin beta-3; integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51); integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); Platelet membrane glycoprotein IIIa; Vitronectin receptor; Vitronectin receptor subunit alpha

          View more View less

          Gene Aliases: BDPLT16; BDPLT2; CD51; CD61; GP3A; GPIIIa; GT; ITGAV; ITGB3; MSK8; VNRA; VTNR

          View more View less

          UniProt ID: (Human) P06756, (Human) P05106

          View more View less

          Entrez Gene ID: (Human) 3685, (Human) 3690

          View more View less

          Function(s)
          virus receptor activity opsonin binding fibronectin binding protease binding protein kinase C binding voltage-gated calcium channel activity protein binding coreceptor activity fibroblast growth factor binding C-X3-C chemokine binding insulin-like growth factor I binding neuregulin binding metal ion binding transforming growth factor beta binding extracellular matrix binding extracellular matrix protein binding protein disulfide isomerase activity platelet-derived growth factor receptor binding enzyme binding identical protein binding vascular endothelial growth factor receptor 2 binding cell adhesion molecule binding
          Process(es)
          positive regulation of protein phosphorylation positive regulation of endothelial cell proliferation platelet degranulation cell-substrate junction assembly cell adhesion cell-matrix adhesion integrin-mediated signaling pathway blood coagulation positive regulation of endothelial cell migration negative regulation of macrophage derived foam cell differentiation negative regulation of lipid storage smooth muscle cell migration cell growth cell migration platelet activation extracellular matrix organization positive regulation of vascular endothelial growth factor receptor signaling pathway cell-substrate adhesion activation of protein kinase activity negative regulation of lipid transport heterotypic cell-cell adhesion substrate adhesion-dependent cell spreading tube development apolipoprotein A-I-mediated signaling pathway wound healing regulation of bone resorption negative regulation of low-density lipoprotein particle receptor biosynthetic process viral entry into host cell vascular endothelial growth factor receptor signaling pathway mesodermal cell differentiation positive regulation of peptidyl-tyrosine phosphorylation negative regulation of lipoprotein metabolic process leukocyte migration negative chemotaxis angiogenesis involved in wound healing platelet aggregation angiogenesis antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent positive regulation of cell proliferation positive regulation of cell migration positive regulation of osteoblast proliferation endodermal cell differentiation apoptotic cell clearance positive regulation of cell adhesion regulation of phagocytosis entry of symbiont into host cell by promotion of host phagocytosis ERK1 and ERK2 cascade calcium ion transmembrane transport extrinsic apoptotic signaling pathway in absence of ligand regulation of apoptotic cell clearance negative regulation of entry of bacterium into host cell negative regulation of extrinsic apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0