Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CD58 Antibodies

          Proteintech

          CD58 Monoclonal Antibody (TS2/9)

          View all (36) CD58 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CD58 Monoclonal Antibody (TS2/9)

          • Antibody Testing Data (1)
          CD58 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD58 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD58 Antibody (65193-1-IG) in Flow

          1X10^6 human peripheral blood lymphocytes were surface stained with 0.25 µg Anti-Human CD58 (Product # 65193-1-IG, Clone: TS2/9) and CoraLite®488-Conjugated AffiniPure Goat Anti-Mouse IgG(H+L) at dilution 1:1,000 (green), or stained with 0.25 µg Mouse IgG1 Isotype Control and CoraLite®488-Conjugated AffiniPure Goat Anti-Mouse IgG(H+L) at dilution 1:1,000 (black). Cells were not fixed. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD58 Antibody in Flow Cytometry (Flow)

          Product Details

          65193-1-IG

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          TS2/9

          Immunogen

          Human cytolytic T cells
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Atlantic Blue
          • CoraLite Plus 488
          • CoraLite Plus 647
          • CoraLite Plus 670
          • CoraLitePlus 405
          • CoraLitePlus 555
          • FITC Plus
          • Request custom conjugation

          Form

          Liquid

          Concentration

          500 µg/mL

          Purification

          Protein G

          Storage buffer

          PBS, pH 7.3

          Contains

          0.1% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Wet ice

          Target Information

          CD58 is a member of the immunoglobulin superfamily and exists both as a transmembrane protein and a GPI-linked extracellular protein, with cells capable of expressing both forms concurrently. Additionally, soluble CD58 has been detected in serum, urine, and synovial fluid, particularly in patients with rheumatoid arthritis. CD58 is widely expressed on non-hematopoietic and most hematopoietic cells. CD58 serves as a ligand for the CD2 protein on T lymphocytes, playing a crucial role in adhesion and activation of T cells. The interaction between CD58 and CD2 initiates a signaling pathway that includes phosphorylation and leads to cytokine release. This interaction is important for T cell activation, cytokine production, cytotoxicity, and adhesion to various cell types. The protein is localized to the plasma membrane, and alternatively spliced transcript variants have been described, indicating diverse functional roles. CD58's involvement in immune responses highlights its significance in both normal physiological processes and disease states.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Ag3; CD305; CD58; CD58 antigen, (lymphocyte function-associated antigen 3); FLJ23181; FLJ43722; LAIR-1; LAIR1; Lymphocyte function-associated antigen 3; RP5-1086K13.2; Surface glycoprotein LFA-3

          View more View less

          Gene Aliases: ag3; CD58; LFA-3; LFA3

          View more View less

          UniProt ID: (Human) P19256

          View more View less

          Entrez Gene ID: (Human) 965

          View more View less

          Function(s)
          receptor binding protein binding
          Process(es)
          cell adhesion single organismal cell-cell adhesion heterotypic cell-cell adhesion leukocyte migration cellular response to interferon-gamma cellular response to tumor necrosis factor positive regulation of interleukin-8 secretion
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
          git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline