Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CD61 (Integrin beta 3) Antibodies

          Invitrogen

          CD61 (Integrin beta 3) Monoclonal Antibody (HM beta 3.1), FITC

          View all (108) CD61 (Integrin beta 3) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CD61 (Integrin beta 3) Monoclonal Antibody (HM beta 3.1), FITC

          • Antibody Testing Data (1)
          CD61 (Integrin beta 3) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD61 (Integrin beta 3) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD61 (Integrin beta 3) Antibody (MA5-28176) in Flow

          Flow cytometry of CD61 (Integrin beta 3) in mouse peripheral blood platelets. Sample was incubated with CD61 (Integrin beta 3) monoclonal antibody (Product # MA5-28176) followed by Alexa Fluor 488. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD61 (Integrin beta 3) Antibody in Flow Cytometry (Flow)

          Product Details

          MA5-28176

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          Neat-1:10
          -
          Product Specifications

          Species Reactivity

          Mouse

          Host/Isotype

          Hamster / IgG

          Class

          Monoclonal

          Type

          Antibody

          Clone

          HM beta 3.1

          Immunogen

          Mouse alpha 5 beta 3 protein purified from the mouse hybridoma 2B4
          View immunogen

          Conjugate

          FITC FITC FITC
          • PE 
          • Request custom conjugation

          Excitation/Emission Max

          498/517 nm View spectra spectra

          Form

          Liquid

          Concentration

          100 µg/mL

          Purification

          Protein G

          Storage buffer

          PBS with 1% BSA

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2745152

          Product Specific Information

          This antibody recognizes the murine integrin beta 3 subunit (CD61), approximately a 90 kDa a type I membrane protein, expressed primarily on megakaryocytes, platelets, monocytes, macrophages, granulocytes and endothelial cells. CD61 associates with either the alpha IIb integrin (CD41) or the alpha V integrin (CD51) to form the platelet glycoprotein complex IIb/IIIa and the vitronectin receptor (VNR) respectively.

          Target Information

          CD61, also known as GPIIIa or ITGB3, is a 105 kDa glycoprotein expressed on activated T cells, granulocytes, megakaryocytes, platelets, and their precursors. It plays a crucial role in platelet aggregation and functions as a receptor for fibrinogen, fibronectin, von Willebrand factor, vitronectin, and thrombospondin. CD61 forms heterodimeric complexes by associating non-covalently with integrin alpha subunits: alphaV (CD51) to create the Vitronectin Receptor and alphaIIb (CD41) to form gpIIb/IIIa. These complexes are responsible for adhesion to extracellular matrix components, facilitating cell adhesion and cell-surface mediated signaling. CD61 is expressed on platelets and megakaryocytes in association with CD41, and on endothelial cells, monocytes, and osteoclasts in association with CD51. The protein product of CD61 is composed of an alpha chain and a beta chain, which can combine with multiple partners to form different integrins. Its involvement in cell adhesion and signaling underscores its importance in normal physiological processes. Dysfunction of CD61 is associated with diseases such as Glanzmann Thrombasthenia and Platelet type-16 Bleeding Disorder, highlighting its critical role in hemostasis and platelet function.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CD61; GPIIIa; Integrin beta-3; platelet gpIIIa; Platelet membrane glycoprotein IIIa

          View more View less

          Gene Aliases: CD61; GP3A; INGRB3; Itgb3

          View more View less

          UniProt ID: (Mouse) O54890

          View more View less

          Entrez Gene ID: (Mouse) 16416

          View more View less

          Function(s)
          fibronectin binding protease binding protein disulfide isomerase activity receptor activity integrin binding protein binding peptide binding identical protein binding vascular endothelial growth factor receptor 2 binding cell adhesion molecule binding extracellular matrix binding fibrinogen binding
          Process(es)
          positive regulation of endothelial cell proliferation positive regulation of leukocyte migration protein folding cellular calcium ion homeostasis cell-substrate junction assembly cell adhesion cell-matrix adhesion integrin-mediated signaling pathway regulation of G-protein coupled receptor protein signaling pathway positive regulation of endothelial cell migration negative regulation of macrophage derived foam cell differentiation positive regulation of fibroblast migration negative regulation of lipid storage smooth muscle cell migration positive regulation of smooth muscle cell migration cell growth cell migration platelet activation regulation of cell migration positive regulation of cell migration cell-substrate adhesion activation of protein kinase activity negative regulation of lipid transport positive regulation of cell adhesion mediated by integrin positive regulation of osteoblast proliferation heterotypic cell-cell adhesion substrate adhesion-dependent cell spreading apolipoprotein A-I-mediated signaling pathway positive regulation of MAPK cascade positive regulation of osteoclast differentiation negative regulation of low-density lipoprotein particle receptor biosynthetic process positive regulation of angiogenesis positive regulation of bone resorption viral entry into host cell positive regulation of smooth muscle cell proliferation cell projection morphogenesis positive regulation of peptidyl-tyrosine phosphorylation negative regulation of lipoprotein metabolic process negative chemotaxis regulation of release of sequestered calcium ion into cytosol angiogenesis involved in wound healing negative regulation of cell death positive regulation of ERK1 and ERK2 cascade platelet aggregation positive regulation of glomerular mesangial cell proliferation positive regulation of adenylate cyclase-inhibiting opioid receptor signaling pathway
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0