Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Description: The monoclonal antibody 135.4 recognizes ILT1, also known as CD85h, LIR7 and LILRA2. ILT1 is a member of the large family of ILT molecules (Immunologulin-like Transcripts) also known as Leukocyte Ig-like Receptors (LIRs) and Monocyte/Macrophage Ig-like Receptors (MIRs). ILT1 contains a short cytoplasmic domain. Through the cytoplasmic domain ILT1 associates with the Fc receptor gamma chain but not DAP12 resulting in a stimulatory signal. ILT1 may act as a receptor for MHC Class I antigens. Expression of ILT1 is mainly found on monocytes, macrophages, DC and weakly on granulocytes. ILT1 is thought to play a role in the TH1/Th2 balance as well as TLR signaling. The monoclonal antibody 135.4 has been reported to have activating properties upon crosslinking which result in calcium influx.
Applications Reported: This 135.4 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This 135.4 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.5 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.
Filtration: 0.2 µm post-manufacturing filtered.
Leukocyte Ig-like receptors (LIRs) are a family of immunoreceptors expressed predominantly on monocytes and B cells and at lower levels on dendritic cells and natural killer (NK) cells. All LIRs in subfamily B have an inhibitory function (LILRB1; MIM 604811). LIRs in subfamily A, with short cytoplasmic domains lacking an immunoreceptor tyrosine-based inhibitory motif (ITIM) and with trans membrane regions containing a charged arginine residue, may initiate stimulatory cascades. One member of subfamily A (LILRA3; MIM 604818) lacks a transmembrane region and is presumed to be a soluble receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: CD85 antigen-like family member H; CD85h; ILT-1; Immunoglobulin-like transcript 1; leucocyte Ig-like receptor A2; Leukocyte immunoglobulin-like receptor 7; Leukocyte immunoglobulin-like receptor subfamily A member 2; leukocyte immunoglobulin-like receptor subfamily A member 2 soluble; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 2; LIR-7
Gene Aliases: CD85H; ILT1; LILRA2; LIR-7; LIR7
UniProt ID: (Human) Q8N149
Entrez Gene ID: (Human) 11027
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support