Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CD86 Antibodies

          Invitrogen

          CD86 (B7-2) Monoclonal Antibody (IT2.2), Super Bright™ 645, eBioscience™

          16 Published Figures
          12 References
          View all (200) CD86 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CD86 (B7-2) Monoclonal Antibody (IT2.2), Super Bright™ 645, eBioscience™

          • Antibody Testing Data (1)
          • Published Figures (16)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 17

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD86 (B7-2) Antibody (64-0869-42) in Flow

          Staining of normal human peripheral blood cells with Anti-Human CD14 APC (Product # 17-0149) and Mouse IgG2b K Isotype Control Super Bright 645 (Product # 64-4732) (left) or Anti-Human CD86 (B7-2) Super Bright 645 (right). Cells in the lymphocyte and monocyte gate were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Neutralization (Neu)

          Product Details

          64-0869-42

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          5 µL (0.25 µg)/test
          View 11 publications 11 publications

          Neutralization (Neu)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG2b, kappa

          Recommended Isotype Control

          Mouse IgG2b kappa Isotype Control (eBMG2b), Super Bright™ 645, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          IT2.2

          Conjugate

          Super Bright™ 645 Super Bright™ 645 Super Bright™ 645
          • Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 647 
          • Alexa Fluor 700 
          • APC 
          • Biotin 
          • Brilliant UV 737
          • Brilliant Violet 480
          • Brilliant Violet 711
          • Functional Grade
          • NovaFluor Blue 660-120S
          • NovaFluor Blue 760
          • NovaFluor Blue 800
          • NovaFluor Red 700
          • NovaFluor Red 725
          • NovaFluor Red 755
          • NovaFluor Yellow 610
          • NovaFluor Yellow 690
          • NovaFluor Yellow 700
          • NovaFluor Yellow 755
          • NovaFluor Yellow 810
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Request custom conjugation

          Excitation/Emission Max

          414/645 nm View spectra spectra

          Form

          Liquid

          Concentration

          5 µL/Test

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2, with BSA

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2688245

          Product Specific Information

          Description: The IT2.2 monoclonal antibody reacts with human CD86, an ~80 kDa surface receptor also known as B7-2. CD86 and CD80 are members of the B7 family of costimulatory molecules. CD86 is expressed at low levels on B cells, macrophages, and dendritic cells and is upregulated on B cells through a variety of surface stimuli including the BCR complex, CD40 and some cytokine receptors. In addition to CD80 (B7-1), CD86 is a counter-receptor for the T cell surface molecules CD28 and CD152 (CTLA-4). The interaction of CD86 with its ligands plays a critical role in T-B crosstalk, T cell costimulation, autoantibody production and Th2-mediated Ig production. The kinetics of upregulation of CD86 upon stimulation supports its major contribution during the primary phase of an immune response.

          Applications Reported: This IT2.2 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This IT2.2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.25 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

          Super Bright 645 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 645 nm. We recommend using a 660/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome.

          When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information.

          Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.

          Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333) for up to 3 days in the dark at 4 degrees C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

          Excitation: 405 nm; Emission: 645 nm; Laser: Violet Laser

          Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.

          Target Information

          CD86, along with CD80, is a member of the B7 family of costimulatory molecules and plays a crucial role in T cell activation and immune response regulation. CD86 is expressed at low levels on B cells, macrophages, and dendritic cells, and its expression is upregulated on B cells through various stimuli, including the BCR complex, CD40, and certain cytokine receptors. As a type I membrane protein and member of the immunoglobulin superfamily, CD86 serves as a ligand for the T cell surface proteins CD28 and CTLA-4 (CD152). The interaction between CD86 and CD28 provides a costimulatory signal essential for T cell activation during antigen presentation, while binding with CTLA-4 negatively regulates T cell activation, diminishing the immune response. This interaction is critical for T-B cell crosstalk, T cell costimulation, autoantibody production, and Th2-mediated Ig production. The kinetics of CD86 upregulation upon stimulation suggest its significant contribution during the primary phase of an immune response. CD86 and CD80 have distinct roles in T helper cell differentiation, and insufficient co-stimulation involving these molecules can induce tolerance. Alternative splicing of CD86 results in two transcript variants encoding different isoforms, with additional variants described but not fully sequenced. Dysfunction in CD86 is associated with diseases such as gallbladder squamous cell carcinoma and myocarditis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: Activation B7-2 antigen; B lymphocyte activation antigen B72; B-lymphocyte activation antigen B7-2; B70; BU63; CD28 antigen ligand 2; CD86; CD86 antigen (CD28 antigen ligand 2, B7-2 antigen); CTLA-4 counter-receptor B7.2; Early T-cell co-stimulatory molecule 1; FUN-1; MGC34413; T-lymphocyte activation antigen CD86

          View more View less

          Gene Aliases: B7-2; B7.2; B70; CD28LG2; CD86; LAB72

          View more View less

          UniProt ID: (Human) P42081

          View more View less

          Entrez Gene ID: (Human) 942

          View more View less

          Function(s)
          virus receptor activity receptor activity receptor binding protein binding coreceptor activity phosphatidylinositol-4,5-bisphosphate 3-kinase activity
          Process(es)
          response to yeast adaptive immune response T cell proliferation involved in immune response negative regulation of T cell anergy immune response cell-cell signaling aging positive regulation of cell proliferation regulation of phosphatidylinositol 3-kinase signaling T cell costimulation toll-like receptor 3 signaling pathway response to interferon-gamma positive regulation of activated T cell proliferation T cell activation B cell activation response to drug myeloid dendritic cell differentiation positive regulation of lymphotoxin A biosynthetic process positive regulation of interleukin-2 biosynthetic process positive regulation of interleukin-4 biosynthetic process positive regulation of T-helper 2 cell differentiation positive regulation of transcription, DNA-templated viral entry into host cell phosphatidylinositol phosphorylation phosphatidylinositol-mediated signaling defense response to virus cellular response to lipopolysaccharide cellular response to metal ion cellular response to cytokine stimulus
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-2w6sl:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0