Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MSTELFSSTR EEGSSGSGPS FRSNQRKMLN LLLERDTSFT VCPDVPRTPV GKFLGDSANL SILSGGTPKC CLDLSNLSSG EITATQLTTS ADLDETGHLD SSGLQEVHLA GMNHDQHLMK CSPAQLLCST PNGLDRGHRK RDAMCSSSAN KENDNGNLVD SEMKYLGSPI TTVPKLDKNP NLGEDQAEEI SDELMEFSLK DQEAKVSRSG LYRSPSMPEN LNRPRLKQVE KFKDNTIPDK VKKKYFSGQG KLRKGLCLKK TVSLCDITIT QMLEEDSNQG HLIGDFSKVC ALPTVSGKHQ DLKYVNPETV AALLSGKFQG LIEKFYVIDC RYPYEYLGGH IQGALNLYSQ EELFNFFLKK PIVPLDTQKR IIIVFHCEFS SERGPRMCRC LREEDRSLNQ YPALYYPELY ILKGGYRDFF PEYMELCEPQ SYCPMHHQDH KTELLRCRSQ SKVQEGERQL REQIALLVKD MSP
M-phase inducer phosphatase 3is an enzyme that in humans is encoded by theCDC25Cgene. This gene is highly conserved during evolution and it plays a key role in the regulation of cell division. The encoded protein is a tyrosine phosphatase and belongs to the Cdc25 phosphatase family. It directs dephosphorylation of cyclin B-bound CDC2 (CDK1) and triggers entry into mitosis. Also, it is thought to suppress p53-induced growth arrest. Multiple alternatively spliced transcript variants of this gene have been described, however, the full-length nature of many of them is not known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dual specificity phosphatase Cdc25C; EC 3.1.3.48; M-phase inducer phosphatase 3; mitosis inducer CDC25; MPIP3; OTTHUMP00000159490; OTTHUMP00000224012; OTTHUMP00000224013; OTTHUMP00000224016; phosphotyrosine phosphatase; protein phosphatase 1, regulatory subunit 60
Gene Aliases: CDC25; CDC25C; PPP1R60
UniProt ID: (Human) Q9H2E8
Entrez Gene ID: (Human) 995
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support