Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: LVQASERLKN QSKELKDAHC QRKLAMQEFM EINERLTELH TQKQKLARHV RDKEEEVDLV MQKVESLRQE LRRTERAKKE LEVHTEALAA EASKDRKLRE
CDC42BPA (MRCKA) is a serine/threonine kinase which functions in actin cytoskeleton assembly and organization. Three independent coiled-coil domains and the N-terminal region preceding the kinase domain are responsible for intermolecular interactions leading to MRCK alpha multimerization. The transautophosphorylation process is critical for regulation of MRCK alpha catalytic activities. Binding of phorbol esters to MRCK releases its autoinhibition, allowing N-terminal dimerization and subsequent kinase activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDC42 binding protein kinase alpha (DMPK-like); CDC42 binidng protein kinase beta; CDC42-binding protein kinase alpha; CDC42-binding protein kinase alpha (DMPK-like); CDC42BPA; DKFZp686L1738; DKFZp686P1738; DMPK-like alpha; FLJ23347; MRCK alpha; Myotonic dystrophy kinase-related CDC42-binding kinase alpha; myotonic dystrophy kinase-related CDC42-binding protein kinase alpha; myotonic dystrophy protein kinase-like alpha; RP5-1087E8.4; ser-thr protein kinase PK428; ser-thr protein kinase related to the myotonic dystrophy protein kinase; Serine/threonine-protein kinase MRCK alpha
Gene Aliases: CDC42BPA; KIAA0451; MRCK; MRCKA; PK428
UniProt ID: (Human) Q5VT25
Entrez Gene ID: (Human) 8476
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support