Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: SKMISNPTNF NHVAHMGPGD GMQVLMDLPL SAVPPSQEER PGPAPTNLAR QPPSRNKPYI SWPSSGGSEP SVTVPLRSMS DPDQDFDKEP DSDSTKHSTP
CDC42BPB is a serine/threonine kinases which functions in actin cytoskeleton assembly and organization. Myotonic dystrophy kinaserelated Cdc42-binding (DMPK-like) kinases-alpha and beta (MRCK-alpha, beta) contain a cysteine-rich motif and a putative pleckstrin homology domain. MRCKs can phosphorylate nonmuscle Myosin light chain and influences Actin-Myosin contractility. MRCK-alpha can phosphorylate and activate LIM kinases downstream of Cdc42, which leads to inactivation of ADF/Cofilin and to Actin cytoskeletal reorganization. MRCK-alpha can also influence neurite outgrowth promoted by Cdc42 and Rac.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDC42 binding protein kinase beta (DMPK-like); CDC42-binding protein kinase beta; CDC42-binding protein kinase beta (DMPK-like); CDC42BP-beta; CDC42BPB; DMPK-like beta; MRCK beta; Myotonic dystrophy kinase-related CDC42-binding kinase beta; myotonic dystrophy protein kinase-like beta; Serine/threonine-protein kinase MRCK beta
Gene Aliases: CDC42BPB; KIAA1124; MRCKB
UniProt ID: (Human) Q9Y5S2
Entrez Gene ID: (Human) 9578
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support