Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: CLWKNRQQN TIQKYDPPGY LYQGSDMNGQ MVDYTTLSGA SQINGNVHGG FLTNGGLSSG YSHLHHKVPN AVNGIVNGSL NGGLYSGHSN SLTRTHVDFE HPHHLVNGGG MYTAVPQIDP LECVNCRNCR NNNRCFTKTN STFSSSPPPV VPVVAPYPQD GLEMKPLSHV KVPVCLTSAV PDCGQLPEES VKDNVEPVPT QRTCCQDIVN DVSSDGSEDP AEFSRGQEGM INLRIPDHLQ LAKSCVWEGD SCAHSETENN IVSWNALILP PVPEGCAEKT MWSPPGIPLD SPTEVLQQPR ET (987-1287 aa encoded by BC098583)
CDON and BOC are cell surface receptors of the immunoglobulin (Ig)/fibronectin type III (FNIII. CDON and BOC are coexpressed during development, form complexes with each other in a cis fashion, and are related to each other in their ectodomains, but each has a unique long cytoplasmic tail.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAM-related/down-regulated by oncogenes; Cdon homolog; Cell adhesion molecule-related/down-regulated by oncogenes; immunoglobulin superfamily member; immunoglobulin superfamily member CDO; oncogene-regulated cell adhesion molecule; surface glycoprotein, Ig superfamily member
Gene Aliases: Cd0; CDO; CDON; CDON1; HPE11; ORCAM
UniProt ID: (Human) Q4KMG0, (Mouse) Q32MD9, (Rat) O35158
Entrez Gene ID: (Human) 50937, (Mouse) 57810, (Rat) 50938
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support