Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MNYQQQLANS AAIRAEIQRF ESVHPNIYSI YELLERVEEP VLQNQIREHV IAIEDAFVNS QEWTLSRSVP ELKVGIVGNL ASGKSALVHR YLTGTYVQEE SPEGGRFKKE IVVDGQSYLL LIRDEGGPPE AQFAMWVDAV IFVFSLEGEI SFQTVYHYYS RMANYRNTSE IPLVLVGTQD AISSANPRVI DDARARKLSN DLKRCTYYET CATYGLNVER VFQDVAQKIV ATRKKQQLSI GPCKSLPNSP SHSSVCSAQV SAVHISQTSN GGGSLSDYSS SVPSTPSTSQ KELRIDVPPT ANTPTPVRKQ SKRRSNLFTS RKGSDPDKEK KGLESRADSI GSGRAIPIKQ GLPFFVLALT ASTYLRPAGA RARQSSPWPG RAEGWSDQPA LC
GTPase-activating protein for ARF1 and, to a lesser extent, ARF5. Directly and specifically regulates the adapter protein 3 (AP-3)-dependent trafficking of proteins in the endosomal-lysosomal system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AGAP-1; Arf GAP with GTP-binding protein-like, ANK repeat and PH domains 1; Arf-GAP with GTPase, ANK repeat and PH domain-containing protein 1; centaurin, gamma 2; Centaurin-gamma-2; Cnt-g2; GGAP1; GTP-binding and GTPase-activating protein 1
Gene Aliases: AGAP-1; AGAP1; CENTG2; cnt-g2; GGAP1; KIAA1099
UniProt ID: (Human) Q9UPQ3
Entrez Gene ID: (Human) 116987
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support