Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Involved in the recruitment of key centrosomal proteins to the centrosome. Provides centrosomal microtubule-nucleation activity on the gamma-tubulin ring complexes (gamma-TuRCs) and has critical roles in forming a focused bipolar spindle, which is needed for proper tension generation between sister chromatids. Required for localization of KIZ, AKAP9 and gamma-tubulin ring complexes (gamma-TuRCs).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: centrosomal protein 72 kDa; centrosomal protein 72kDa; Centrosomal protein of 72 kDa; Cep72
Gene Aliases: 2610029E11Rik; 4933440J22Rik; CEP72; KIAA1519
UniProt ID: (Human) Q9P209, (Mouse) Q9D3R3
Entrez Gene ID: (Human) 55722, (Mouse) 74470
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support