Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MHLLLFQLLV LLPLGKTTRH QDGRQNQSSL SPVLLPRNQR ELPTGNHEEA EEKPDLFVAV PHLVGTSPAG EGQRQREKML SRFGRFWKKP EREMHPSRDS DSEPFPPGTQ SLIQPIDGMK MEKSPLREEA KKFWHHFMFR KTPASQGVIL PIKSHEVHWE TCRTVPFSQT ITHEGCEKVV VQNNLCFGKC GSVHFPGAAQ HSHTSCSHCL PAKFTTMHLP LNCTELSSVI KVVMLVEECQ CKVKTEHEDG HILHAGSQDS FIPGVSA
This gene encodes a cytokine member of the cysteine knot superfamily, characterized by nine conserved cysteines and a cysteine knot region. The cerberus-related cytokines, together with Dan and DRM/Gremlin, represent a group of bone morphogenetic protein antagonists that can bind directly to BMPs and inhibit their activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cer-l; Cerberus; cerberus 1, cysteine knot superfamily, homolog; cerberus homolog; Cerberus-like protein; cerberus-related 1; Cerberus-related protein; DAN domain family member 4; MGC119894; MGC119895; MGC96951
Gene Aliases: cer-1; CER1; Cerl; Cerl1; Cerr1; DAND4
UniProt ID: (Human) O95813, (Mouse) O55233
Entrez Gene ID: (Human) 9350, (Mouse) 12622
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support