Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total CGGBP1.
CGGBP1 influences expression of the FMR1 gene (MIM 309550), which is associated with the fragile X mental retardation syndrome (MIM 300624), by specifically interacting with the 5-prime (CGG)n-3-prime repeat in its 5-prime UTR.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 20 kDa CGG-binding protein; CGG triplet repeat-binding protein 1; CGG-binding protein 1; p20-CGG binding protein; p20-CGGBP DNA-binding protein
Gene Aliases: AA960172; AL023996; AW045841; CGGBP; CGGBP1; p20-CGGBP
UniProt ID: (Human) Q9UFW8, (Mouse) Q8BHG9
Entrez Gene ID: (Human) 8545, (Mouse) 106143
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support