Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: Jurkat.
Predicted reactivity: Mouse (98%), Rat (100%), Rhesus Monkey (99%), Bovine (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CGGBP1 influences expression of the FMR1 gene (MIM 309550), which is associated with the fragile X mental retardation syndrome (MIM 300624), by specifically interacting with the 5-prime (CGG)n-3-prime repeat in its 5-prime UTR.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 20 kDa CGG-binding protein; CGG triplet repeat-binding protein 1; CGG-binding protein 1; p20-CGG binding protein; p20-CGGBP DNA-binding protein
Gene Aliases: CGGBP; CGGBP1; p20-CGGBP
UniProt ID: (Human) Q9UFW8
Entrez Gene ID: (Human) 8545
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support