Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The CGI-62 gene encodes a hypothetical protein located on chromosome 8.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 164, member A; protein FAM164A; Zinc finger C2HC domain-containing protein 1A; zinc finger, C2HC-type containing 1A
Gene Aliases: 3110050N22Rik; AI790358; AU023959; C8orf70; CGI-62; FAM164A; RGD1311970; ZC2HC1A
UniProt ID: (Human) Q96GY0, (Mouse) Q8BJH1
Entrez Gene ID: (Human) 51101, (Rat) 310244, (Mouse) 67306
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support