Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MWVFGYGSLI WKVDFPYQDK LVGYITNYSR RFWQGSTDHR GVPGKPGRVV
For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 92%; Zebrafish: 100%
CHAC2 catalyzes the cleavage glutathione into 5-oxoproline and a Cys-Gly dipeptide and acts specifically on glutathione, but not on other gamma-glutamyl peptides.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cation transport regulator-like protein 2; ChaC, cation transport regulator homolog 2; ChaC, cation transport regulator-like 2; CHAC2 (HGNC:32363); Gamma-GCG 2; gamma-GCT acting on glutathione homolog 2; Glutathione-specific gamma-glutamylcyclotransferase 2
Gene Aliases: CHAC2
UniProt ID: (Human) Q8WUX2
Entrez Gene ID: (Human) 494143
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support