Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: EFYSYYKCK LALEQQQLIC KQQTPELEPA ATMPPLPQPP LAPAAPIPPA QGAPSMDELI QQSQWNLQQQ EQHLLALRQE QVTAAVAHAV EQQMQKLLEE TQLDMNEFDN LLQPIIDTCT KDAISAGKNW MFSNAKSPPH CELMAGHLRN RITADGAHFE LRLHLIYLIN DVLHHCQRKQ ARELLAALQK VVVPIYCTSF LAVEEDKQQK IARLLQLWEK NGYFDDSIIQ QLQSPALGLG QYQATLINEY SSVVQPVQLA FQQQIQTLKT QHEEFVTSLA QQQQQQQQQQ QQLQMPQMEA EVKATPPPPA PPPAPAPAPA IPPTTQPDDS KPP (19-350 aa encoded by BC021294)
CHERP is involved in calcium homeostasis, growth and proliferation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcium homeostasis endoplasmic reticulum protein; CHERP; ERPROT 213-21; ERPROT213-21; protein with polyglutamine repeat; SR-related CTD associated factor 6; SR-related CTD-associated factor 6
Gene Aliases: 5730408I11Rik; CHERP; D8Wsu96e; DAN16; DAN26; SCAF6; SRA1
UniProt ID: (Human) Q8IWX8, (Mouse) Q8CGZ0
Entrez Gene ID: (Human) 10523, (Mouse) 27967
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support