Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LRLRLRDSS LGAVCVALDW LAFDDLLAGA AHHSQSFQLL RYPPFLPVAF HVLFASSHTP RITFPSSQQE AQNRMSQMRN LIQTLVSGIA PATRSRATPQ ALLLDALCLL LDILAPKLRP VSTQLYSTRE KQQLASLVGT MLAYSLTYRQ ERTPDGQYIY RLEPNVEELC RFPELPARKP LTYQTKQLIA REIEVEKMRR AEASARVENS PQVDGSPPGL EGLLGGIGEK GVHRPAPRNH EQRLEHIMRR AAREEQPEKD FFGRVVVRST AVPSAGDTAP EQDSVERRMG TAVGRSEVWF RFNEGVSNAV RRSLYIRDLL (657-975 aa encoded by BC018184)
CHTF18, CHTF8.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C321D2.3 (novel protein); C321D2.4 (novel protein); CHL12; Chl12 homolog; Chromosome transmission fidelity protein 18 homolog; CTF18, chromosome transmission fidelity factor 18 homolog; hCTF18; homolog of yeast CHL12; some homology with holliday junction DNA helicase RUVB like
Gene Aliases: 6030457M03Rik; C16orf41; C321D2.2; C321D2.3; C321D2.4; CHL12; CHTF18; CTF18; RUVBL
UniProt ID: (Human) Q8WVB6, (Mouse) Q8BIW9
Entrez Gene ID: (Human) 63922, (Mouse) 214901, (Rat) 287146
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support