Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Aliquoting is unnecessary for -20°C storage.
The protein encoded by this gene is a member of the calcium-binding protein family. The specific function of this protein has not yet been determined; however this protein is known to interact with DNA-dependent protein kinase and may play a role in kinase-phosphatase regulation of DNA end joining. This protein also interacts with integrin alphabeta, which may implicate this protein as a regulatory molecule for alphabeta.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium and integrin binding 1 (calmyrin); calcium and integrin binding, protein kinase interacting protein; Calcium and integrin-binding protein 1; Calcium- and integrin-binding protein; Calmyrin; CIB; CIBP; DNA-dependent protein kinase interacting protein; DNA-PK interaction protein; DNA-PKcs-interacting protein; Kinase-interacting protein; KIP; protein kinase, DNA activated, catalytic polypeptide interacting protein; SIP2-28; SNK-interacting protein 2-28; testicular secretory protein Li 9
Gene Aliases: BOS_20130; CIB; CIB1; Cibkip; CIBP; KIP; KIP1; PRKDCIP; SIP2-28
UniProt ID: (Bovine) Q17QE5, (Human) Q99828, (Rat) Q9R010, (Mouse) Q9Z0F4
Entrez Gene ID: (Bovine) 510141, (Human) 10519, (Rat) 81823, (Mouse) 23991
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support