Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: EVREQPRLFP PSADYPDLRK HNNCMAECLT PAIYAKLRNK VTPNGYTLDQ CIQTGVDNPG HPFIKTVGMV AGDEESYEVF ADLFDPVIKL RHNGYDPRVM KHTTDLDASK ITQGQFDEHY VLSSRVRTGR SIRGLSLPPA CTRAERREVE NVAITALEGL KGDLAGRYYK LSEMTEQDQQ RLIDDHFLFD K; Positive Samples: HeLa, Mouse skeletal muscle, Mouse heart, Mouse diaphragm, Rat skeletal muscle; Cellular Location: Intermembrane side, Mitochondrion inner membrane, Peripheral membrane protein
Mitochondrial creatine kinase (MtCK) is responsible for the transfer of high energy phosphate from mitochondria to the cytosolic carrier, creatine. It belongs to the creatine kinase isoenzyme family. It exists as two isoenzymes, sarcomeric MtCK and ubiquitous MtCK, encoded by separate genes. Mitochondrial creatine kinase occurs in two different oligomeric forms: dimers and octamers, in contrast to the exclusively dimeric cytosolic creatine kinase isoenzymes. Sarcomeric mitochondrial creatine kinase has 80% homology with the coding exons of ubiquitous mitochondrial creatine kinase. This gene contains sequences homologous to several motifs that are shared among some nuclear genes encoding mitochondrial proteins and thus may be essential for the coordinated activation of these genes during mitochondrial biogenesis. Three transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Basic-type mitochondrial creatine kinase; Creatine kinase S-type, mitochondrial; creatine kinase, mitochondrial 2 (sarcomeric); EC 2.7.3.2; Mib-CK; S-MtCK; Sarcomeric mitochondrial creatine kinase
Gene Aliases: 2300008A19Rik; CKMT2; ScCKmit; SMTCK
UniProt ID: (Human) P17540, (Mouse) Q6P8J7
Entrez Gene ID: (Human) 1160, (Rat) 688698, (Mouse) 76722
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support