Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MASAGMQILG VVLTLLGWVN GLVSCALPMW KVTAFIGNSI VVAQVVWEGL WMSCVVQSTG QMQCKVYDSL LALPQDLQAA RALCVIALLV ALFGLLVYLA GAKCTTCVEE KDSKARLVLT SGIVFVISGV LTLIPVCWTA HAVIRDFYNP LVAEAQKREL GASLYLGWAA SGLLLLGGGL LCCTCPSGGS QGPSHYMARY STSAPAISRG PSEYPTKNYV
Tight junctions represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. These junctions are comprised of sets of continuous networking strands in the outwardly facing cytoplasmic leaflet, with complementary grooves in the inwardly facing extracytoplasmic leaflet. This gene encodes a component of tight junction strands, which is a member of the claudin family. The protein is an integral membrane protein and is one of the entry cofactors for hepatitis C virus. The gene methylation may be involved in esophageal tumorigenesis. This gene is adjacent to another family member CLDN9 on chromosome 16.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Claudin-6; Claudin6; CLD6; Skullin
Gene Aliases: CLDN6; UNQ757/PRO1488
UniProt ID: (Human) P56747
Entrez Gene ID: (Human) 9074
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support