Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Binds polysaccharides in a Ca(2+)-independent manner with a preferentially binding to fucoidan, beta-glucans and galactans.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-type lectin domain family 18 member A; C-type lectin domain family 18, member A; mannose receptor-like 1; Mannose receptor-like protein; Mannose receptor-like protein 2; mannose receptor-like protein 3
Gene Aliases: CLEC18A; MRCL; MRCL1; Mrlp; MRLP2
UniProt ID: (Human) A5D8T8, (Mouse) Q7TSQ1
Entrez Gene ID: (Human) 348174, (Mouse) 353287
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support