Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LVTHHNFSR CKRGTGVHKL EHHAKLKCIK EKSELKSAEG STWNCCPIDW RAFQSNCYFP LTDNKTWAES ERNCSGMGAH LMTISTEAEQ NFIIQFLDRR LSYFLGLRDE NAKGQWRWVD QTPFNPRRVF WHKNEPDNSQ GENCVVLVYN QDKWAWNDVP CNFEASRICK IPGTTLN (40-215 aa encoded by BC032313)
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 8; C-type lectin domain family 4 member D; C-type lectin domain family 4, member D; C-type lectin receptor; C-type lectin superfamily member 8; C-type lectin-like receptor 6; CD368; CLEC-6; CLECSF8Mpcl; DC-associated C-type lectin 3; Dendritic cell-associated C-type lectin 3; macrophage C-type lectin; MGC40078; superfamilymember 8
Gene Aliases: CD368; CLEC-6; CLEC4D; CLEC6; CLECSF8; MCL; MPCL
UniProt ID: (Human) Q8WXI8
Entrez Gene ID: (Human) 338339
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support