Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MANEAYPCPC DIGHRLEYGG LGREVQVEHI KAYVTKSPVD AGKAVIVIQD IFGWQLPNTR YIADMISGNG YTTIVPDFFV GQEPWDPSGD WSIFPEWLKT RNAQKIDREI SAILKYLKQQ CHAQKIGIVG FCWGGTAVHH LMMKYSEFRA GVSVYGIVKD SEDIYNLKNP TLFIFAENDV VIPLKDVSLL TQKLKEHCKV EYQIKTFSGQ THGFVHRKRE DCSPADKPYI DEARRNLIEW LNKYM
CMBL is a cysteine hydrolase of the dienelactone hydrolase family that is highly expressed in liver cytosol. CMBL preferentially cleaves cyclic esters, and it activates medoxomil-ester prodrugs in which the medoxomil moiety is linked to an oxygen atom.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Carboxymethylenebutenolidase homolog; carboxymethylenebutenolidase homolog (Pseudomonas); carboxymethylenebutenolidase-like (Pseudomonas)
Gene Aliases: CMBL; JS-1
UniProt ID: (Human) Q96DG6
Entrez Gene ID: (Human) 134147
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support