Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CNKSR3 Antibodies

          Abnova

          CNKSR3 Polyclonal Antibody, MaxPab™

          View all (11) CNKSR3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CNKSR3 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (2)
          CNKSR3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CNKSR3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CNKSR3 Antibody (H00154043-B01P) in ICC/IF

          Immunofluorescence of purified MaxPab antibody to CNKSR3 on HeLa cell. Antibody concentration is 10 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CNKSR3 Antibody in Immunocytochemistry (ICC/IF)
          CNKSR3 Antibody in Western Blot (WB)

          Product Details

          H00154043-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CNKSR3 (NP_775786, 1 a.a. approximately 555 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MEPVTKWSPK QVVDWTRGLD DCLQQYVHKF EREKINGEQL LQISHQDLEE LGVTRIGHQE LVLEAVDLLC ALNYGLETDN MKNLVLKLRA SSHNLQNYIS SRRKSPAYDG NTSRKAPNEF LTSVVELIGA AKALLAWLDR APFTGITDFS VTKNKIIQLC LDLTTTVQKD CFVAEMEDKV LTVVKVLNGI CDKTIRSTTD PVMSQCACLE EVHLPNIKPG EGLGMYIKST YDGLHVITGT TENSPADRSQ KIHAGDEVIQ VNQQTVVGWQ LKNLVKKLRE NPTGVVLLLK KRPTGSFNFT PAPLKNLRWK PPLVQTSPPP ATTQSPESTM DTSLKKEKSA ILDLYIPPPP AVPYSPRDEN GSFVYGGSSK CKQPLPGPKG SESPNSFLDQ ESRRRRFTIA DSDQLPGYSV ETNILPTKMR EKTPSYGKPR PLSMPADGNW MGIVDPFARP RGHGRKGEDA LCRYFSNERI PPIIEESSSP PYRFSRPTTE RHLVRGADYI RGSRCYINSD LHSSATIPFQ EEGTKKKSGS SATKSSSTEP SLLVSWFTRL KLLTH

          Target Information

          CNKSR3 may be involved in transepithelial sodium transport. Regulates aldosterone-induced and ENaC-mediated sodium transport possibly through regulation of the ERK pathway.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CNK homolog protein 3; CNK3; CNKSR family member 3; Connector enhancer of kinase suppressor of ras 3; Connector enhancer of KSR 3; KOX14; Maguin-like protein; membrane associated guanylate kinase interacting protein-like 1; membrane associated guanylate kinase, WW and PDZ domain containing 1; membrane-associated guanylate kinase-interacting protein-like 1; ZNF21

          View more View less

          Gene Aliases: CNKSR3; MAGI1

          View more View less

          UniProt ID: (Human) Q6P9H4

          View more View less

          Entrez Gene ID: (Human) 154043

          View more View less

          Function(s)
          protein binding
          Process(es)
          positive regulation of sodium ion transport negative regulation of peptidyl-serine phosphorylation negative regulation of ERK1 and ERK2 cascade
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0