Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: NPENVTDVS LGISSNEQDQ GSDKGENEAM ESSGKRAPQC YPSSVNSART VMLFNLGSAY CLRSEYDKAR KCLHQAASMI HPKEVPPEAI LLAVYLELQN GNTQLALQII KRNQLLPAVK THSEVRKKPV FQPVHPIQPI QMSAFTTVQR K (568-717 aa encoded by BC002928)
Component of the CCR4-NOT complex which is one of the major cellular mRNA deadenylases and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation and general transcription regulation. Additional complex functions may be a consequence of its influence on mRNA expression. Is not required for association of CNOT7 to the CCR4-NOT complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCR4-NOT transcription complex subunit 10; CCR4-NOT transcription complex, subunit 10; FLJ12890; FLJ13165
Gene Aliases: 2600001P13Rik; CNOT10
UniProt ID: (Human) Q9H9A5, (Mouse) Q8BH15, (Rat) Q5XIA4
Entrez Gene ID: (Human) 25904, (Mouse) 78893, (Rat) 316034
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support