Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPSLQPVVMC VMKHLPKVPE KKLKLVMADK ELYRACAVEV KRQIWQDNQA LFGDEVSPLL KQYILEKESA LFSTELSVLH NFFSPSPKTR RQGEVVQRLT RMVGKNVKLY DMVLQFLRTL FLRTRNVHYC TLRAELLMSL HDLDVGEICT VDPCHKFTWC LDACIRERFV DSKRARELQG FLDGVKKGQE QVLGDLSMIL CDPFAINTLA LSTVRHLQEL VGQETLPRDS PDLLLLLRLL ALGQGAWDMI DSQVFKEPKM EVELITRFLP MLMSFLVDDY TFNVDQKLPA EEKAPVSYPN TLPESFTKFL QEQRMACEVG LYYVLHITKQ RNKNALLRLL PGLVETFGDL AFGDIFLHLL TGNLALLADE FALEDFCSSL FDGFFLTASP RKENVHRHAL RLLIHLHPRV APSKLEALQK ALEPTGQSGE AVKELYSQLG EKLEQLDHRK PSPAQAAETP ALELPLPSVP APAPL
NELFB is a subunit of negative elongation factor (NELF), which also includes NELFA (WHSC2), either NELFC or NELFD (TH1L), and NELFE (RDBP), a heterodimer of SPT4 (SUPT4H1), to cause transcriptional pausing of RNA polymerase II (Narita et al., 2003).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cofactor of BRCA1; DKFZp586B0519; Negative elongation factor B; negative elongation factor protein B; NELF-B; RP13-122B23.3
Gene Aliases: COBRA1; KIAA1182; NELF-B; NELFB
UniProt ID: (Human) Q8WX92
Entrez Gene ID: (Human) 25920
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support