Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: LSMILCDPFA INTLALSTVR HLQELVGQET LPRDSPDLLL LLRLLALGQG AWDMIDSQVF KEPKMEVELI TRFLPMLMSF LVDDYTFNVD QKLPAEEKAP VSYPNTLPES FTKFLQEQRM ACEVGLYYVL HITKQRNKNA LLRLLPGLVE TFGDLAFGDI FLHLLTGNLA LLADEFALED FCSSLFDGFF LTASPRKENV HRHALRLLIH LHPRVAPSKL EALQKALEPT GQSGEAVKEL YSQLGEKLEQ LDHRKPSPAQ AAETPALELP LPSVPAPAPL
NELFB is a subunit of negative elongation factor (NELF), which also includes NELFA (WHSC2), either NELFC or NELFD (TH1L), and NELFE (RDBP), a heterodimer of SPT4 (SUPT4H1), to cause transcriptional pausing of RNA polymerase II (Narita et al., 2003).
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cofactor of BRCA1; DKFZp586B0519; Negative elongation factor B; negative elongation factor complex member B, Cobra1; negative elongation factor protein B; NELF-B; ortholog of human cofactor of BRCA1 (COBRA1); RP13-122B23.3
Gene Aliases: A730008L03Rik; AB041607; AI663983; COBRA1; KIAA1182; MNCb-5210; NELF-B; NELFB
UniProt ID: (Human) Q8WX92, (Mouse) Q8C4Y3
Entrez Gene ID: (Human) 25920, (Mouse) 58202
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support