Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MATAATSPAL KRLDLRDPAA LFETHGAEEI RGLERQVRAE IEHKKEELRQ MVGERYRDLI EAADTIGQMR RCAVGLVDAV KATDQYCARL RQAGSAAPRP PRAQQPQQPS QEKFYSMAAQ IKLLLEIPEK IWSSMEASQC LHATQLYLLC CHLHSLLQLD SSSSRYSPVL SRFPILIRQV AAASHFRSTI LHESKMLLKC QGVSDQAVAE ALCSIMLLEE SSPRQALTDF LLARKATIQK LLNQPHHGAG IKAQICSLVE LLATTLKQAH ALFYTLPEGL LPDPALPCGL LFSTLETITG QHPAGKGTGV LQEEMKLCSW FKHLPASIVE FQPTLRTLAH PISQEYLKDT LQKWIHMCNE DIKNGITNLL MYVKSMKGLA GIRDAMWELL TNESTNHSWD VLCRRLLEKP LLFWEDMMQQ LFLDRLQTLT KEGFDSISSS SKELLVSALQ ELESSTSNSP SNKHIHFEYN MSLFLWSESP NDLPSDAAWV SVANRGQFAS SGLSMKAQAI SPCVQNFCSA LDSKLKVKLD DLLAYLPSDD SSLPKDVSPT QAKSSAFDRY ADAGTVQEML RTQSVACIKH IVDCIRAELQ SIEEGVQGQQ DALNSAKLHS VLFMARLCQS LGELCPHLKQ CILGKSESSE KPAREFRALR KQGKVKTQEI IPTQAKWQEV KEVLLQQSVM GYQVWSSAVV KVLIHGFTQS LLLDDAGSVL ATATSWDELE IQEEAESGSS VTSKIRLPAQ PSWYVQSFLF SLCQEINRVG GHALPKVTLQ EMLKSCMVQV VAAYEKLSEE KQIKKEGAFP VTQNRALQLL YDLRYLNIVL TAKGDEVKSG RSKPDSRIEK VTDHLEALID PFDLDVFTPH LNSNLHRLVQ RTSVLFGLVT GTENQLAPRS STFNSQEPHN ILPLASSQIR FGLLPLSMTS TRKAKSTRNI ETKAQVVPPA RSTAGDPTVP GSLFRQLVSE EDNTSAPSLF KLGWLSSMTK
The protein encoded by this gene is one of eight proteins which form a Golgi-localized complex required for normal Golgi morphology and function. It is thought that this protein is required for steps in the normal medial and trans Golgi-associated processing of glycoconjugates and plays a role in the organization of the Golgi-localized complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: COG complex subunit 1; Component of oligomeric Golgi complex 1; conserved oligomeric Golgi complex protein 1; Conserved oligomeric Golgi complex subunit 1; DKFZp762L1710; low density lipoprotein receptor defect B complementing
Gene Aliases: CDG2G; COG1; KIAA1381; LDLB
UniProt ID: (Human) Q8WTW3
Entrez Gene ID: (Human) 9382
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support