Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Collagen III Antibodies

          Abnova

          COL3A1 Polyclonal Antibody, MaxPab™

          View all (37) Collagen III antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite COL3A1 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          COL3A1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          COL3A1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          COL3A1 Antibody (H00001281-D01P) in WB

          Western Blot analysis of COL3A1 expression in transfected 293T cell line by COL3A1 MaxPab polyclonal antibody. Lane 1: COL3A1 transfected lysate(111.90 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          COL3A1 Antibody in Western Blot (WB)

          Product Details

          H00001281-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          COL3A1 (AAH28178.1, 1 a.a. approximately 1163 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MMSFVQKGSW LLLALLHPTI ILAQQEAVEG GCSHLGQSYA DRDVWKPEPC QICVCDSGSV LCDDIICDDQ ELDCPNPEIP FGECCAVCPQ PPTAPTRPPN GQGPQGPKGD PGPPGIPGRN GDPGIPGQPG SPGSPGPPGI CESCPTGPQN YSPQYDSYDV KSGVAVGGLA GYPGPAGPPG PPGPPGTSGH PGSPGSPGYQ GPPGEPGQAG PSGPPGPPGA IGPSGPAGKD GESGRPGRPG ERGLPGPPGI KGPAGIPGFP GMKGHRGFDG RNGEKGETGA PGLKGENGLP GENGAPGPMG PRGAPGERGR PGLPGAAGAR GNDGARGSDG QPGPPGPPGT AGFPGSPGAK GEVGPAGSPG SNGAPGQRGE PGPQGHAGAQ GPPGPPGING SPGGKGEMGP AGIPGAPGLM GARGPPGPAG ANGAPGLRGG AGEPGKNGAK GEPGPRGERG EAGIPGVPGA KGEDGKDGSP GEPGANGLPG AAGERGAPGF RGPAGPNGIP GEKGPAGERG APGPAGPRGA AGEPGRDGVP GGPGMRGMPG SPGGPGSDGK PGPPGSQGES GRPGPPGPSG PRGQPGVMGF PGPKGNDGAP GKNGERGGPG GPGPQGPPGK NGETGPQGPP GPTGPGGDKG DTGPPGPQGL QGLPGTGGPP GENGKPGEPG PKGDAGAPGA PGGKGDAGAP GERGPPGLAG APGLRGGAGP PGPEGGKGAA GPPGPPGAAG TPGLQGMPGE RGGLGSPGPK GDKGEPGGPG ADGVPGKDGP RGPTGPIGPP GPAGQPGDKG EGGAPGLPGI AGPRGSPGER GETGPPGPAG FPGAPGQNGE PGGKGERGAP GEKGEGGPPG VAGPPGKDGT SGHPGPIGPP GPRGNRGERG SEGSPGHPGQ PGPPGPPGAP GPCCGGVGAA AIAGIGGEKA GGFAPYYGDE PMDFKINTDE IMTSLKSVNG QIESLISPDG SRKNPARNCR DLKFCHPELK SGEYWVDPNQ GCKLDAIKVF CNMETGETCI SANPLNVPRK HWWTDSSAEK KHVWFGESMD GGFQFSYGNP ELPEDVLDVQ LAFLRLLSSR ASQNITYHCK NSIAYMDQAS GNVKKALKLM GSNEGEFKAE GNSKFTYTVL EDGCTKHTGE WSKTVFEYRT RKAVRLPIVD IAPYDIGGPD QEFGVDVGPV CFL

          Target Information

          Collagen III is a fibrillar collagen that is found in extensible connective tissues such as skin, lung, and the vascular system, frequently in association with type I Collagen. Mutations in th gene are associated with Ehlers Danlos syndrome type IV, and with aortic and arterial aneurysms. Although alternate transcripts have been detected for this gene, they are the result of mutations; these mutations alter splicing, often leading to the exclusion of multiple exons.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: alpha1 (III) collagen; COL 3; COL3; COL3 alpha 1; Collagen 3; Collagen alpha-1(III) chain; collagen, fetal; collagen, type III, alpha 1; Collagen3; Ehlers-Danlos syndrome type IV, autosomal dominant; FLJ34534; MGC93704

          View more View less

          Gene Aliases: COL3A1; EDS4A

          View more View less

          UniProt ID: (Human) P02461

          View more View less

          Entrez Gene ID: (Human) 1281

          View more View less

          Function(s)
          integrin binding extracellular matrix structural constituent protein binding SMAD binding metal ion binding platelet-derived growth factor binding
          Process(es)
          skeletal system development cell-matrix adhesion transforming growth factor beta receptor signaling pathway integrin-mediated signaling pathway heart development aging response to radiation response to mechanical stimulus peptide cross-linking cerebral cortex development platelet activation extracellular matrix organization collagen fibril organization collagen catabolic process response to cytokine positive regulation of Rho protein signal transduction wound healing extracellular fibril organization skin development digestive tract development regulation of immune response negative regulation of immune response aorta smooth muscle tissue morphogenesis cellular response to amino acid stimulus negative regulation of neuron migration
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0