Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: SSNNFNYGAY HSLEAIYHEM DNIAADFPDL ARRVKIGHSF ENRPMYVLKF STGKGVRRPA VWLNAGIHSR EWISQATAIW TARKIVSDYQ RDPAITSILE KMDIFLLPVA NPDGYVYTQT QNRLWRKTRS RNPGSSCIGA DPNRNWNASF AGKGASDNPC SEVYHGP
Carboxypeptidase A4 is a member of the carboxypeptidase A/B subfamily. Carboxypeptidases are zinc-containing exopeptidases that catalyze the release of carboxy-terminal amino acids, and are synthesized as zymogens that are activated by proteolytic cleavage. This protein could be involved in the histone hyperacetylation pathway. It is imprinted and may be a strong candidate protein for prostate cancer aggressiveness.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Carboxypeptidase A3; Carboxypeptidase A4
Gene Aliases: 1110019K20Rik; AV009555; CPA3; CPA4; RGD1563619; UNQ694/PRO1339
UniProt ID: (Human) Q9UI42, (Mouse) Q6P8K8
Entrez Gene ID: (Human) 51200, (Rat) 502736, (Mouse) 71791
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support