Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKLCSLAVLV PIVLFCEQHV FAFQSGQVLA ALPRTSRQVQ VLQNLTTTYE IVLWQPVTAD LIVKKKQVHF FVNASDVDNV KAHLNVSGIP CSVLLADVED LIQQQISNDT VSPRASASYY EQYHSLNEIY SWIEFITERH PDMLTKIHIG SSFEKYPLYV LKVSGKEQAA KNAIWIDCGI HAREWISPAF CLWFIGHITQ FYGIIGQYTN LLRLVDFYVM PVVNVDGYDY SWKKNRMWRK NRSFYANNHC IGTDLNRNFA SKHWCEEGAS SSSCSETYCG LYPESEPEVK AVASFLRRNI NQIKAYISMH SYSQHIVFPY SYTRSKSKDH EELSLVASEA VRAIEKTSKN TRYTHGHGSE TLYLAPGGGD DWIYDLGIKY SFTIELRDTG TYGFLLPERY IKPTCREAFA AVSKIAWHVI RNV
Carboxypeptidases are enzymes that hydrolyze C-terminal peptide bonds. The carboxypeptidase family includes metallo-, serine, and cysteine carboxypeptidases. According to their substrate specificity, these enzymes are referred to as carboxypeptidase A (cleaving aliphatic residues) or carboxypeptidase B (cleaving basic amino residues). CPB2 is activated by trypsin and acts on carboxypeptidase B substrates. After thrombin activation, the mature protein downregulates fibrinolysis. Polymorphisms have been described for its gene and its promoter region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: carboxypeptidase B-like protein; Carboxypeptidase B2; carboxypeptidase B2 (plasma); carboxypeptidase B2 (plasma, carboxypeptidase U); carboxypeptidase R; Carboxypeptidase U; CPU; pCPB; Plasma carboxypeptidase B; TAFI; Thrombin-activable fibrinolysis inhibitor; thrombin-activatable fibrinolysis inhibitor
Gene Aliases: CPB2; CPU; PCPB; TAFI
UniProt ID: (Human) Q96IY4
Entrez Gene ID: (Human) 1361
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support