Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CPEB3 Antibodies

          Abnova

          CPEB3 Polyclonal Antibody, MaxPab™

          View all (7) CPEB3 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CPEB3 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          CPEB3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CPEB3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CPEB3 Antibody (H00022849-B01P) in WB

          Western Blot analysis of CPEB3 expression in transfected 293T cell line by CPEB3 MaxPab polyclonal antibody. Lane 1: CPEB3 transfected lysate(76.67 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CPEB3 Antibody in Western Blot (WB)

          Product Details

          H00022849-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CPEB3 (AAH36444.1, 1 a.a. approximately 697 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MQDDLLMDKS KTQPQPQQQQ RQQQQPQPES SVSEAPSTPL SSETPKPEEN SAVPALSPAA APPAPNGPDK MQMESPLLPG LSFHQPPQQP PPPQEPAAPG ASLSPSFGST WSTGTTNAVE DSFFQGITPV NGTMLFQNFP HHVNPVFGGT FSPQIGLAQT QHHQQPPPPA PAPQPAQPAQ PPQAQPPQQR RSPASPSQAP YAQRSAAAAY GHQPIMTSKP SSSSAVAAAA AAAAASSASS SWNTHQSVNA AWSAPSNPWG GLQAGRDPRR AVGVGVGVGV GVPSPLNPIS PLKKPFSSNV IAPPKFPRAA PLTSKSWMED NAFWTDNGNN LLPFQDRSRP YDTFNLHSLE NSLMDMIRTD HEPLKGKHYP PSGPPMSFAD IMWRNHFAGR MGINFHHPGT DNIMALNNAF LDDSHGDQAL SSGLSSPTRC QNGERVERYS RKVFVGGLPP DIDEDEITAS FRRFGPLVVD WPHKAESKSY FPPKGYAFLL FQEESSVQAL IDACLEEDGK LYLCVSSPIK DKPVQIRPWN LSDSDFVMDG SQPLDPRKTI FVGGVPRPLR AVELAMIMDR LYGGVCYAGI DTDPELKYPK GAGRVAFSNQ QSYIAAISAR FVQLQHNDID KRVEVKPYVL DDQMCDECQG TRCGGKFAPF FCANVTCLQY YCEYCWASIH SRAGREFHKP LVKEGGDRPR HVPFRWS

          Target Information

          Sequence-specific RNA-binding protein which acts as a translational repressor in the basal unstimulated state but, following neuronal stimulation, acts as a translational activator. In contrast to CPEB1, does not bind to the cytoplasmic polyadenylation element (CPE), a uridine-rich sequence element within the mRNA 3'-UTR, but binds to a U-rich loop within a stem-loop structure. Required for the consolidation and maintenance of hippocampal-based long term memory. In the basal state, binds to the mRNA 3'-UTR of the glutamate receptors GRIA2/GLUR2 mRNA and negatively regulates their translation. Also represses the translation of DLG4, GRIN1, GRIN2A and GRIN2B. When activated, acts as a translational activator of GRIA1 and GRIA2. In the basal state, suppresses SUMO2 translation but activates it following neuronal stimulation. Binds to the 3'-UTR of TRPV1 mRNA and represses TRPV1 translation which is required to maintain normal thermoception. Binds actin mRNA, leading to actin translational repression in the basal state and to translational activation following neuronal stimulation. Negatively regulates target mRNA levels by binding to TOB1 which recruits CNOT7/CAF1 to a ternary complex and this leads to target mRNA deadenylation and decay (PubMed:21336257). In addition to its role in translation, binds to and inhibits the transcriptional activation activity of STAT5B without affecting its dimerization or DNA-binding activity. This, in turn, represses transcription of the STAT5B target gene EGFR which has been shown to play a role in enhancing learning and memory performance (PubMed:20639532). In contrast to CPEB1, CPEB2 and CPEB4, not required for cell cycle progression (PubMed:26398195). [UniProt]

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CPE-binding protein 3; CPE-BP3; Cytoplasmic polyadenylation element-binding protein 3; hCPEB-3

          View more View less

          Gene Aliases: CPEB3; KIAA0940

          View more View less

          UniProt ID: (Human) Q8NE35

          View more View less

          Entrez Gene ID: (Human) 22849

          View more View less

          Function(s)
          nucleotide binding translation repressor activity, nucleic acid binding RNA binding mRNA 3'-UTR binding protein binding translation factor activity, RNA binding RNA stem-loop binding mRNA 3'-UTR AU-rich region binding ribosome binding poly(A) RNA binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter translation long-term memory negative regulation of translation positive regulation of translation regulation of synaptic plasticity positive regulation of nuclear-transcribed mRNA poly(A) tail shortening regulation of dendritic spine development positive regulation of dendritic spine development 3'-UTR-mediated mRNA destabilization cellular response to amino acid stimulus positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay negative regulation of cytoplasmic translational elongation positive regulation of mRNA polyadenylation negative regulation of cytoplasmic translation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-k2m5x:80/100.66.128.182:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0