Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LSFAVESEC TFLDYIKGGT QINFTVAIDF TASNGNPSQS TSLHYMSPYQ LNAYALALTA VGEIIQHYDS DKMFPALGFG AKLPPDGRVS HEFPLNGNQE NPSCCGIDGI LEAYHRSLRT VQLYGPTNFA PVVTHVARNA AAVQDGSQYS VLLIITDGVI SDMAQTKEAI VNAAKLPMSI IIVGVGQAEF DAMVELDGDD VRISSRGKLA ERDIVQFVPF RDYVDRTGNH VLSMARLARD VLAEIPDQLV SYMKAQGIRP RPPPAAPTHS PSQSPARTPP ASPLHTHI (15-301 aa encoded by BC053872 )
Probable calcium-dependent phospholipid-binding protein that may play a role in calcium-mediated intracellular processes. Plays a role in dendrite formation by melanocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: copine 5; copine 5 protein; Copine V; Copine-5
Gene Aliases: A830083G22Rik; COPN5; CPN5; CPNE5; KIAA1599
UniProt ID: (Human) Q9HCH3, (Mouse) Q8JZW4
Entrez Gene ID: (Human) 57699, (Mouse) 240058, (Rat) 309650
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support