Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: PFLLARWGR AWGQIQTTSA NENSTVLPSS TSSSSDGNLR PEAITAIIVV FSLLAALLLA VGLALLVRKL REKRQTEGTY RPSSEEQFSH AAEARAPQDS KETVQGCLPI (15-123 aa encoded by BC018409)
Involved in the establishment of cell polarity in mammalian epithelial cells. Regulates the morphogenesis of tight junctions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: crumbs family member 3; Protein crumbs homolog 3
Gene Aliases: CRB3; UNQ588/PRO1158
UniProt ID: (Human) Q9BUF7
Entrez Gene ID: (Human) 92359
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support